Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42698242-42698515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPH2
Entrez Gene ID
5961
Gene Name
peripherin 2
Gene Aliases
AOFMD, AVMD, CACD2, DS, MDBS1, PRPH, RDS, RP7, TSPAN22, rd2
UniGene
Hs.654489
Gene Chromosome Location
Chr.6: 42694509-42723024 on Build GRCh38
Transcripts
NM_000322.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1222190 c.934G>A p.V312M Chr.6: 42698402-42698402 on Build GRCh38
6756708 c.1014C>T p.D338D Chr.6: 42698322-42698322 on Build GRCh38
5521446 c.997G>A p.E333K Chr.6: 42698339-42698339 on Build GRCh38
6502592 c.939G>A p.P313P Chr.6: 42698397-42698397 on Build GRCh38
6278016 c.1019G>A p.G340D Chr.6: 42698317-42698317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746349275 Chr.6: 42698273-42698273 on Build GRCh38
rs361524 Chr.6: 42698282-42698282 on Build GRCh38
rs61748434 Chr.6: 42698398-42698398 on Build GRCh38
rs773342762 Chr.6: 42698318-42698318 on Build GRCh38
rs751894791 Chr.6: 42698269-42698269 on Build GRCh38
rs390659 Chr.6: 42698426-42698426 on Build GRCh38
rs778093966 Chr.6: 42698458-42698458 on Build GRCh38
rs200269333 Chr.6: 42698385-42698385 on Build GRCh38
rs189476016 Chr.6: 42698270-42698270 on Build GRCh38
rs201620487 Chr.6: 42698426-42698431 on Build GRCh38
rs969800260 Chr.6: 42698350-42698350 on Build GRCh38
rs936676026 Chr.6: 42698391-42698391 on Build GRCh38
rs768773825 Chr.6: 42698289-42698289 on Build GRCh38
rs121918566 Chr.6: 42698389-42698389 on Build GRCh38
rs375325973 Chr.6: 42698490-42698490 on Build GRCh38
rs776760107 Chr.6: 42698472-42698472 on Build GRCh38
rs758660018 Chr.6: 42698457-42698457 on Build GRCh38
rs780477133 Chr.6: 42698363-42698363 on Build GRCh38
rs61748432 Chr.6: 42698422-42698422 on Build GRCh38
rs61748429 Chr.6: 42698438-42698439 on Build GRCh38
rs903826110 Chr.6: 42698335-42698335 on Build GRCh38
rs764833128 Chr.6: 42698325-42698325 on Build GRCh38
rs193921105 Chr.6: 42698339-42698339 on Build GRCh38
rs375326474 Chr.6: 42698434-42698434 on Build GRCh38
rs886043500 Chr.6: 42698320-42698320 on Build GRCh38
rs62645939 Chr.6: 42698470-42698470 on Build GRCh38
rs760687443 Chr.6: 42698321-42698321 on Build GRCh38
rs761108586 Chr.6: 42698425-42698426 on Build GRCh38
rs750796514 Chr.6: 42698425-42698425 on Build GRCh38
rs781604214 Chr.6: 42698347-42698347 on Build GRCh38
rs62645938 Chr.6: 42698481-42698481 on Build GRCh38
rs281865374 Chr.6: 42698414-42698422 on Build GRCh38
rs767883031 Chr.6: 42698323-42698324 on Build GRCh38
rs1011239238 Chr.6: 42698331-42698331 on Build GRCh38
rs137853904 Chr.6: 42698403-42698403 on Build GRCh38
rs370994796 Chr.6: 42698486-42698486 on Build GRCh38
rs61748430 Chr.6: 42698432-42698432 on Build GRCh38
rs62645938 Chr.6: 42698481-42698481 on Build GRCh38
rs771868894 Chr.6: 42698309-42698309 on Build GRCh38
rs759011231 Chr.6: 42698409-42698409 on Build GRCh38
rs756613766 Chr.6: 42698361-42698361 on Build GRCh38
rs749295756 Chr.6: 42698287-42698287 on Build GRCh38
rs202230698 Chr.6: 42698390-42698390 on Build GRCh38
rs62645939 Chr.6: 42698470-42698470 on Build GRCh38
rs955930526 Chr.6: 42698327-42698327 on Build GRCh38
rs763666304 Chr.6: 42698418-42698418 on Build GRCh38
rs180775924 Chr.6: 42698275-42698275 on Build GRCh38
rs766811082 Chr.6: 42698426-42698427 on Build GRCh38
rs368337679 Chr.6: 42698271-42698271 on Build GRCh38
rs749391375 Chr.6: 42698375-42698375 on Build GRCh38
rs774584357 Chr.6: 42698291-42698291 on Build GRCh38
rs183714869 Chr.6: 42698448-42698448 on Build GRCh38
rs746742888 Chr.6: 42698399-42698399 on Build GRCh38
rs281865375 Chr.6: 42698345-42698345 on Build GRCh38
rs762308246 Chr.6: 42698493-42698493 on Build GRCh38
rs434102 Chr.6: 42698323-42698323 on Build GRCh38
rs779048042 Chr.6: 42698447-42698447 on Build GRCh38
rs775059514 Chr.6: 42698412-42698412 on Build GRCh38
rs142381301 Chr.6: 42698334-42698334 on Build GRCh38
rs61748433 Chr.6: 42698416-42698416 on Build GRCh38
rs373560002 Chr.6: 42698424-42698424 on Build GRCh38
rs1039119330 Chr.6: 42698454-42698454 on Build GRCh38
rs202109541 Chr.6: 42698322-42698322 on Build GRCh38
rs750992392 Chr.6: 42698352-42698352 on Build GRCh38
rs144111167 Chr.6: 42698427-42698427 on Build GRCh38
rs745807357 Chr.6: 42698484-42698484 on Build GRCh38
rs762023485 Chr.6: 42698322-42698323 on Build GRCh38
rs1052492384 Chr.6: 42698397-42698397 on Build GRCh38
rs754337687 Chr.6: 42698433-42698433 on Build GRCh38
rs752365478 Chr.6: 42698328-42698328 on Build GRCh38
rs139329966 Chr.6: 42698381-42698381 on Build GRCh38
rs746836512 Chr.6: 42698469-42698469 on Build GRCh38
rs1016933713 Chr.6: 42698473-42698473 on Build GRCh38
rs761542005 Chr.6: 42698292-42698292 on Build GRCh38
rs764785171 Chr.6: 42698411-42698411 on Build GRCh38
rs762660751 Chr.6: 42698413-42698413 on Build GRCh38
rs771015384 Chr.6: 42698402-42698402 on Build GRCh38
rs962365465 Chr.6: 42698485-42698485 on Build GRCh38
rs771829987 Chr.6: 42698394-42698394 on Build GRCh38
rs425876 Chr.6: 42698407-42698407 on Build GRCh38
rs750992655 Chr.6: 42698404-42698406 on Build GRCh38
rs774283806 Chr.6: 42698318-42698323 on Build GRCh38

View Full Product Details