Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41515765-41516008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT15
Entrez Gene ID
3866
Gene Name
keratin 15
Gene Aliases
CK15, K15, K1CO
UniGene
Hs.654570
Gene Chromosome Location
Chr.17: 41513745-41522413 on Build GRCh38
Transcripts
NM_002275.3 XM_011524784.2 XM_017024614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1256077 c.982A>G p.T328A Chr.17: 41515929-41515929 on Build GRCh38
5016827 c.973C>A p.L325M Chr.17: 41515938-41515938 on Build GRCh38
1750030 c.975G>C p.L325L Chr.17: 41515936-41515936 on Build GRCh38
3517442 c.946C>T p.Q316* Chr.17: 41515965-41515965 on Build GRCh38
3517441 c.963G>A p.E321E Chr.17: 41515948-41515948 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768767508 Chr.17: 41515927-41515927 on Build GRCh38
rs369009119 Chr.17: 41515851-41515851 on Build GRCh38
rs368789787 Chr.17: 41515878-41515878 on Build GRCh38
rs748429113 Chr.17: 41515939-41515939 on Build GRCh38
rs375136118 Chr.17: 41515877-41515877 on Build GRCh38
rs776947737 Chr.17: 41515888-41515888 on Build GRCh38
rs541758777 Chr.17: 41515792-41515792 on Build GRCh38
rs965743156 Chr.17: 41515900-41515900 on Build GRCh38
rs747416124 Chr.17: 41515957-41515957 on Build GRCh38
rs758225696 Chr.17: 41515852-41515852 on Build GRCh38
rs369832431 Chr.17: 41515836-41515836 on Build GRCh38
rs771153001 Chr.17: 41515978-41515978 on Build GRCh38
rs755047067 Chr.17: 41515868-41515868 on Build GRCh38
rs372848775 Chr.17: 41515867-41515867 on Build GRCh38
rs758981626 Chr.17: 41515840-41515840 on Build GRCh38
rs540200615 Chr.17: 41515931-41515931 on Build GRCh38
rs563494383 Chr.17: 41515834-41515834 on Build GRCh38
rs752298242 Chr.17: 41515850-41515850 on Build GRCh38
rs929857507 Chr.17: 41515880-41515880 on Build GRCh38
rs776189615 Chr.17: 41515837-41515837 on Build GRCh38
rs964083632 Chr.17: 41515862-41515862 on Build GRCh38
rs151089400 Chr.17: 41515928-41515928 on Build GRCh38
rs374991384 Chr.17: 41515843-41515843 on Build GRCh38
rs974853892 Chr.17: 41515908-41515908 on Build GRCh38
rs575388820 Chr.17: 41515853-41515853 on Build GRCh38
rs775166862 Chr.17: 41515919-41515919 on Build GRCh38
rs755136685 Chr.17: 41515933-41515933 on Build GRCh38
rs757208858 Chr.17: 41515864-41515864 on Build GRCh38
rs149918613 Chr.17: 41515932-41515932 on Build GRCh38
rs200152929 Chr.17: 41515938-41515938 on Build GRCh38
rs759069758 Chr.17: 41515889-41515889 on Build GRCh38
rs369652117 Chr.17: 41515943-41515943 on Build GRCh38
rs780062822 Chr.17: 41515866-41515866 on Build GRCh38
rs949153676 Chr.17: 41515909-41515909 on Build GRCh38
rs764883400 Chr.17: 41515841-41515841 on Build GRCh38
rs373081379 Chr.17: 41515952-41515952 on Build GRCh38
rs779296116 Chr.17: 41515872-41515872 on Build GRCh38
rs145005480 Chr.17: 41515951-41515951 on Build GRCh38
rs762576421 Chr.17: 41515923-41515923 on Build GRCh38
rs771474796 Chr.17: 41515884-41515884 on Build GRCh38
rs200702391 Chr.17: 41515873-41515873 on Build GRCh38
rs564196717 Chr.17: 41515902-41515902 on Build GRCh38
rs763856197 Chr.17: 41515924-41515924 on Build GRCh38

View Full Product Details