Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3228403-3228658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRGPRE
Entrez Gene ID
116534
Gene Name
MAS related GPR family member E
Gene Aliases
GPR167, MGRF, MRGE
UniGene
Hs.745512
Gene Chromosome Location
Chr.11: 3227811-3232386 on Build GRCh38
Transcripts
NM_001039165.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5625734 c.187G>T p.D63Y Chr.11: 3228610-3228610 on Build GRCh38
4032383 c.189C>T p.D63D Chr.11: 3228608-3228608 on Build GRCh38
4032384 c.188A>G p.D63G Chr.11: 3228609-3228609 on Build GRCh38
4032385 c.171C>T p.F57F Chr.11: 3228626-3228626 on Build GRCh38
5685904 c.273G>A p.P91P Chr.11: 3228524-3228524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770052986 Chr.11: 3228578-3228578 on Build GRCh38
rs769389632 Chr.11: 3228442-3228442 on Build GRCh38
rs371485588 Chr.11: 3228475-3228475 on Build GRCh38
rs369200596 Chr.11: 3228560-3228560 on Build GRCh38
rs758199266 Chr.11: 3228427-3228427 on Build GRCh38
rs1054039176 Chr.11: 3228559-3228559 on Build GRCh38
rs751476188 Chr.11: 3228525-3228525 on Build GRCh38
rs559487164 Chr.11: 3228480-3228480 on Build GRCh38
rs199688365 Chr.11: 3228537-3228537 on Build GRCh38
rs899337770 Chr.11: 3228534-3228534 on Build GRCh38
rs770550122 Chr.11: 3228429-3228429 on Build GRCh38
rs368971331 Chr.11: 3228493-3228493 on Build GRCh38
rs371522732 Chr.11: 3228428-3228428 on Build GRCh38
rs780741481 Chr.11: 3228476-3228476 on Build GRCh38
rs749454603 Chr.11: 3228632-3228632 on Build GRCh38
rs760513724 Chr.11: 3228607-3228607 on Build GRCh38
rs893534302 Chr.11: 3228571-3228571 on Build GRCh38
rs201376741 Chr.11: 3228626-3228626 on Build GRCh38
rs34568544 Chr.11: 3228596-3228597 on Build GRCh38
rs867930119 Chr.11: 3228569-3228569 on Build GRCh38
rs764008025 Chr.11: 3228473-3228473 on Build GRCh38
rs775661694 Chr.11: 3228590-3228590 on Build GRCh38
rs770107975 Chr.11: 3228631-3228631 on Build GRCh38
rs771200300 Chr.11: 3228556-3228556 on Build GRCh38
rs776726917 Chr.11: 3228507-3228507 on Build GRCh38
rs765444406 Chr.11: 3228516-3228516 on Build GRCh38
rs11026040 Chr.11: 3228599-3228599 on Build GRCh38
rs200957230 Chr.11: 3228554-3228554 on Build GRCh38
rs570815823 Chr.11: 3228572-3228572 on Build GRCh38
rs200110994 Chr.11: 3228597-3228597 on Build GRCh38
rs201651483 Chr.11: 3228538-3228538 on Build GRCh38
rs757025534 Chr.11: 3228526-3228526 on Build GRCh38
rs146856033 Chr.11: 3228456-3228456 on Build GRCh38
rs200462560 Chr.11: 3228451-3228451 on Build GRCh38
rs773736351 Chr.11: 3228497-3228497 on Build GRCh38
rs374090732 Chr.11: 3228455-3228455 on Build GRCh38
rs374764339 Chr.11: 3228478-3228478 on Build GRCh38
rs750293023 Chr.11: 3228602-3228602 on Build GRCh38
rs375800804 Chr.11: 3228518-3228518 on Build GRCh38
rs776871095 Chr.11: 3228450-3228450 on Build GRCh38
rs767285183 Chr.11: 3228527-3228527 on Build GRCh38
rs371926242 Chr.11: 3228452-3228452 on Build GRCh38
rs780877112 Chr.11: 3228433-3228433 on Build GRCh38
rs763899695 Chr.11: 3228484-3228486 on Build GRCh38
rs755887832 Chr.11: 3228533-3228533 on Build GRCh38
rs534877746 Chr.11: 3228596-3228596 on Build GRCh38
rs750238036 Chr.11: 3228531-3228531 on Build GRCh38
rs950696908 Chr.11: 3228437-3228437 on Build GRCh38
rs771276116 Chr.11: 3228501-3228501 on Build GRCh38
rs763980999 Chr.11: 3228524-3228524 on Build GRCh38
rs748793044 Chr.11: 3228492-3228492 on Build GRCh38
rs529843091 Chr.11: 3228517-3228517 on Build GRCh38
rs778416441 Chr.11: 3228619-3228619 on Build GRCh38
rs139403822 Chr.11: 3228553-3228553 on Build GRCh38
rs766546646 Chr.11: 3228448-3228448 on Build GRCh38
rs754862294 Chr.11: 3228621-3228621 on Build GRCh38
rs200334859 Chr.11: 3228598-3228598 on Build GRCh38
rs202197260 Chr.11: 3228612-3228612 on Build GRCh38
rs751405991 Chr.11: 3228569-3228586 on Build GRCh38
rs187488196 Chr.11: 3228622-3228622 on Build GRCh38
rs766159353 Chr.11: 3228608-3228608 on Build GRCh38
rs745412549 Chr.11: 3228434-3228434 on Build GRCh38
rs958889057 Chr.11: 3228623-3228623 on Build GRCh38
rs761077384 Chr.11: 3228447-3228447 on Build GRCh38
rs376921803 Chr.11: 3228629-3228629 on Build GRCh38
rs753489871 Chr.11: 3228609-3228609 on Build GRCh38
rs372934659 Chr.11: 3228498-3228498 on Build GRCh38
rs201409610 Chr.11: 3228513-3228513 on Build GRCh38
rs757845019 Chr.11: 3228625-3228625 on Build GRCh38
rs374547754 Chr.11: 3228446-3228446 on Build GRCh38
rs779775924 Chr.11: 3228482-3228482 on Build GRCh38

View Full Product Details