Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8649430-8649663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFAP5
Entrez Gene ID
8076
Gene Name
microfibrillar associated protein 5
Gene Aliases
AAT9, MAGP-2, MAGP2, MFAP-5, MP25
UniGene
Hs.512842
Gene Chromosome Location
Chr.12: 8645943-8662888 on Build GRCh38
Transcripts
NM_001297709.1 NM_001297710.1 NM_001297711.1 NM_001297712.1 NM_003480.3 NR_123733.1 NR_123734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
944186 c.390G>T p.K130N Chr.12: 8649520-8649520 on Build GRCh38
1994695 c.340C>T p.R114* Chr.12: 8649570-8649570 on Build GRCh38
3786118 c.355G>A p.V119I Chr.12: 8649555-8649555 on Build GRCh38
1994693 c.344G>A p.R115H Chr.12: 8649566-8649566 on Build GRCh38
3980168 c.403A>T p.M135L Chr.12: 8649507-8649507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376830567 Chr.12: 8649570-8649570 on Build GRCh38
rs752036635 Chr.12: 8649554-8649554 on Build GRCh38
rs866733609 Chr.12: 8649513-8649513 on Build GRCh38
rs763031215 Chr.12: 8649490-8649490 on Build GRCh38
rs780574974 Chr.12: 8649551-8649551 on Build GRCh38
rs927906237 Chr.12: 8649528-8649528 on Build GRCh38
rs775163759 Chr.12: 8649577-8649577 on Build GRCh38
rs192822432 Chr.12: 8649555-8649555 on Build GRCh38
rs752124786 Chr.12: 8649589-8649589 on Build GRCh38
rs752465187 Chr.12: 8649524-8649524 on Build GRCh38
rs777242064 Chr.12: 8649532-8649532 on Build GRCh38
rs770619503 Chr.12: 8649558-8649558 on Build GRCh38
rs767358373 Chr.12: 8649517-8649517 on Build GRCh38
rs374890052 Chr.12: 8649515-8649515 on Build GRCh38
rs540639689 Chr.12: 8649534-8649534 on Build GRCh38
rs760474089 Chr.12: 8649582-8649582 on Build GRCh38
rs865910671 Chr.12: 8649533-8649533 on Build GRCh38
rs753493759 Chr.12: 8649585-8649585 on Build GRCh38
rs774489509 Chr.12: 8649569-8649569 on Build GRCh38
rs762855396 Chr.12: 8649465-8649465 on Build GRCh38
rs753315025 Chr.12: 8649548-8649548 on Build GRCh38
rs769921511 Chr.12: 8649562-8649562 on Build GRCh38
rs1015841546 Chr.12: 8649590-8649590 on Build GRCh38
rs752770204 Chr.12: 8649615-8649616 on Build GRCh38
rs945068341 Chr.12: 8649459-8649459 on Build GRCh38
rs756712055 Chr.12: 8649550-8649550 on Build GRCh38
rs117523212 Chr.12: 8649583-8649583 on Build GRCh38
rs767408914 Chr.12: 8649521-8649521 on Build GRCh38
rs778887600 Chr.12: 8649483-8649483 on Build GRCh38
rs755761395 Chr.12: 8649542-8649542 on Build GRCh38
rs778001824 Chr.12: 8649563-8649563 on Build GRCh38
rs764711127 Chr.12: 8649588-8649588 on Build GRCh38
rs771981445 Chr.12: 8649571-8649571 on Build GRCh38
rs781681464 Chr.12: 8649611-8649611 on Build GRCh38
rs373421193 Chr.12: 8649566-8649566 on Build GRCh38
rs199578843 Chr.12: 8649514-8649514 on Build GRCh38
rs781404692 Chr.12: 8649556-8649556 on Build GRCh38
rs748795143 Chr.12: 8649567-8649567 on Build GRCh38
rs769980641 Chr.12: 8649475-8649475 on Build GRCh38
rs1050283078 Chr.12: 8649625-8649625 on Build GRCh38
rs201597525 Chr.12: 8649610-8649610 on Build GRCh38

View Full Product Details