Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42349892-42350165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLB
Entrez Gene ID
5423
Gene Name
DNA polymerase beta
Gene Aliases
-
UniGene
Hs.654484
Gene Chromosome Location
Chr.8: 42338452-42371813 on Build GRCh38
Transcripts
NM_002690.2 XM_005273535.3 XM_005273536.3 XM_005273537.3 XM_005273538.2 XM_005273539.2 XM_005273540.4 XM_006716353.2 XM_017013583.1 XM_017013584.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3900286 c.319G>T p.G107C Chr.8: 42350064-42350064 on Build GRCh38
4899777 c.273T>G p.D91E Chr.8: 42350018-42350018 on Build GRCh38
6549301 c.284C>T p.S95L Chr.8: 42350029-42350029 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759532567 Chr.8: 42350012-42350012 on Build GRCh38
rs758558251 Chr.8: 42349990-42349990 on Build GRCh38
rs771233353 Chr.8: 42350093-42350094 on Build GRCh38
rs373934594 Chr.8: 42350095-42350095 on Build GRCh38
rs760360725 Chr.8: 42350030-42350030 on Build GRCh38
rs753569815 Chr.8: 42350047-42350047 on Build GRCh38
rs749841076 Chr.8: 42350089-42350089 on Build GRCh38
rs776830689 Chr.8: 42350099-42350102 on Build GRCh38
rs1030232850 Chr.8: 42350076-42350076 on Build GRCh38
rs747318428 Chr.8: 42349967-42349967 on Build GRCh38
rs778568185 Chr.8: 42350085-42350085 on Build GRCh38
rs768988358 Chr.8: 42349980-42349980 on Build GRCh38
rs3817667 Chr.8: 42349936-42349936 on Build GRCh38
rs763514354 Chr.8: 42349951-42349951 on Build GRCh38
rs369177937 Chr.8: 42350093-42350093 on Build GRCh38
rs767746911 Chr.8: 42350136-42350136 on Build GRCh38
rs377121458 Chr.8: 42350023-42350023 on Build GRCh38
rs781486026 Chr.8: 42350100-42350100 on Build GRCh38
rs3136745 Chr.8: 42349963-42349963 on Build GRCh38
rs568996287 Chr.8: 42349943-42349943 on Build GRCh38
rs55953487 Chr.8: 42350070-42350070 on Build GRCh38
rs748183535 Chr.8: 42349989-42349990 on Build GRCh38
rs776801874 Chr.8: 42349988-42349988 on Build GRCh38
rs774433675 Chr.8: 42350050-42350050 on Build GRCh38
rs900301099 Chr.8: 42350045-42350045 on Build GRCh38
rs769946450 Chr.8: 42350116-42350116 on Build GRCh38
rs778063657 Chr.8: 42350049-42350049 on Build GRCh38
rs371956560 Chr.8: 42350011-42350011 on Build GRCh38
rs769895566 Chr.8: 42350007-42350007 on Build GRCh38
rs148674351 Chr.8: 42350038-42350038 on Build GRCh38
rs768985449 Chr.8: 42350096-42350096 on Build GRCh38
rs773737449 Chr.8: 42349944-42349944 on Build GRCh38
rs377568594 Chr.8: 42350115-42350115 on Build GRCh38
rs773211075 Chr.8: 42350010-42350010 on Build GRCh38
rs187073233 Chr.8: 42349992-42349992 on Build GRCh38
rs746271125 Chr.8: 42350104-42350105 on Build GRCh38
rs752608576 Chr.8: 42350024-42350024 on Build GRCh38
rs747183821 Chr.8: 42350088-42350089 on Build GRCh38
rs753891293 Chr.8: 42350128-42350128 on Build GRCh38

View Full Product Details