Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73480606-73480879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTO1
Entrez Gene ID
25821
Gene Name
mitochondrial tRNA translation optimization 1
Gene Aliases
CGI-02, COXPD10
UniGene
Hs.347614
Gene Chromosome Location
Chr.6: 73461727-73501456 on Build GRCh38
Transcripts
NM_001123226.1 NM_012123.3 NM_133645.2 XM_006715444.2 XM_006715445.2 XM_006715446.3 XM_011535723.1 XM_011535724.1 XM_011535725.2 XM_017010697.1 XM_017010698.1 XM_017010699.1 XM_017010700.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
451779 c.1292G>A p.G431E Chr.6: 73480762-73480762 on Build GRCh38
6346939 c.1238G>T p.R413L Chr.6: 73480708-73480708 on Build GRCh38
1445845 c.1289C>T p.A430V Chr.6: 73480759-73480759 on Build GRCh38
7377875 c.1147G>A p.D383N Chr.6: 73480692-73480692 on Build GRCh38
5554190 c.1282T>G p.F428V Chr.6: 73480752-73480752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029241914 Chr.6: 73480841-73480841 on Build GRCh38
rs968331050 Chr.6: 73480644-73480644 on Build GRCh38
rs141477592 Chr.6: 73480682-73480682 on Build GRCh38
rs766326436 Chr.6: 73480781-73480781 on Build GRCh38
rs777067523 Chr.6: 73480649-73480650 on Build GRCh38
rs771732801 Chr.6: 73480833-73480834 on Build GRCh38
rs201848621 Chr.6: 73480635-73480635 on Build GRCh38
rs935181583 Chr.6: 73480811-73480815 on Build GRCh38
rs398122419 Chr.6: 73480777-73480777 on Build GRCh38
rs972218527 Chr.6: 73480653-73480653 on Build GRCh38
rs761538034 Chr.6: 73480830-73480831 on Build GRCh38
rs767029283 Chr.6: 73480771-73480771 on Build GRCh38
rs925019140 Chr.6: 73480801-73480801 on Build GRCh38
rs377702561 Chr.6: 73480717-73480717 on Build GRCh38
rs200518440 Chr.6: 73480703-73480703 on Build GRCh38
rs746293185 Chr.6: 73480680-73480680 on Build GRCh38
rs887091614 Chr.6: 73480774-73480774 on Build GRCh38
rs754467660 Chr.6: 73480804-73480804 on Build GRCh38
rs773800597 Chr.6: 73480813-73480813 on Build GRCh38
rs947813456 Chr.6: 73480708-73480708 on Build GRCh38
rs750271885 Chr.6: 73480639-73480639 on Build GRCh38
rs775623164 Chr.6: 73480746-73480746 on Build GRCh38
rs779895347 Chr.6: 73480679-73480679 on Build GRCh38
rs943998709 Chr.6: 73480780-73480780 on Build GRCh38
rs772209676 Chr.6: 73480739-73480739 on Build GRCh38
rs746493463 Chr.6: 73480680-73480680 on Build GRCh38
rs2036039 Chr.6: 73480725-73480725 on Build GRCh38
rs777334217 Chr.6: 73480838-73480838 on Build GRCh38
rs772554295 Chr.6: 73480845-73480845 on Build GRCh38
rs377325620 Chr.6: 73480647-73480647 on Build GRCh38
rs751612754 Chr.6: 73480784-73480784 on Build GRCh38
rs774190783 Chr.6: 73480826-73480827 on Build GRCh38
rs539835237 Chr.6: 73480707-73480707 on Build GRCh38
rs145163085 Chr.6: 73480692-73480692 on Build GRCh38
rs759603080 Chr.6: 73480727-73480727 on Build GRCh38
rs147521446 Chr.6: 73480836-73480836 on Build GRCh38
rs780859448 Chr.6: 73480822-73480822 on Build GRCh38
rs752345992 Chr.6: 73480825-73480825 on Build GRCh38
rs923594911 Chr.6: 73480669-73480669 on Build GRCh38
rs770281584 Chr.6: 73480823-73480824 on Build GRCh38
rs747803200 Chr.6: 73480697-73480697 on Build GRCh38
rs749645528 Chr.6: 73480840-73480840 on Build GRCh38
rs149407452 Chr.6: 73480767-73480767 on Build GRCh38
rs760900956 Chr.6: 73480747-73480747 on Build GRCh38
rs771440116 Chr.6: 73480724-73480724 on Build GRCh38

View Full Product Details