Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 94593467-94593724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105369438
Entrez Gene ID
105369438
Gene Name
uncharacterized LOC105369438
Gene Aliases
-
UniGene
Hs.568921
Gene Chromosome Location
Chr.11: 94545332-94740355 on Build GRCh38
Transcripts
NR_135093.1 NR_135094.1 NR_135096.1 NM_152431.2

Gene Symbol
PIWIL4
Entrez Gene ID
143689
Gene Name
piwi like RNA-mediated gene silencing 4
Gene Aliases
HIWI2, MIWI2
UniGene
Hs.660188
Gene Chromosome Location
Chr.11: 94567308-94621421 on Build GRCh38
Transcripts
NR_135093.1 NR_135094.1 NR_135096.1 NM_152431.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
933512 c.1091G>T p.R364I Chr.11: 94593582-94593582 on Build GRCh38
3737567 c.1064T>C p.M355T Chr.11: 94593555-94593555 on Build GRCh38
246607 c.1042G>A p.V348I Chr.11: 94593533-94593533 on Build GRCh38
3454789 c.1116C>T p.L372L Chr.11: 94593607-94593607 on Build GRCh38
3752820 c.1108G>C p.A370P Chr.11: 94593599-94593599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760299910 Chr.11: 94593674-94593674 on Build GRCh38
rs757435710 Chr.11: 94593571-94593573 on Build GRCh38
rs773613135 Chr.11: 94593668-94593668 on Build GRCh38
rs761066054 Chr.11: 94593519-94593519 on Build GRCh38
rs758423897 Chr.11: 94593608-94593608 on Build GRCh38
rs766933935 Chr.11: 94593671-94593671 on Build GRCh38
rs148862743 Chr.11: 94593632-94593632 on Build GRCh38
rs747227272 Chr.11: 94593645-94593645 on Build GRCh38
rs34492831 Chr.11: 94593554-94593554 on Build GRCh38
rs546251499 Chr.11: 94593670-94593670 on Build GRCh38
rs748563763 Chr.11: 94593662-94593662 on Build GRCh38
rs1026616871 Chr.11: 94593502-94593502 on Build GRCh38
rs745915474 Chr.11: 94593607-94593607 on Build GRCh38
rs752357680 Chr.11: 94593551-94593551 on Build GRCh38
rs746230327 Chr.11: 94593657-94593658 on Build GRCh38
rs57607909 Chr.11: 94593599-94593599 on Build GRCh38
rs111738403 Chr.11: 94593489-94593489 on Build GRCh38
rs779072088 Chr.11: 94593656-94593658 on Build GRCh38
rs7119735 Chr.11: 94593544-94593544 on Build GRCh38
rs147602287 Chr.11: 94593533-94593533 on Build GRCh38
rs780257909 Chr.11: 94593637-94593637 on Build GRCh38
rs7120750 Chr.11: 94593700-94593700 on Build GRCh38
rs145301752 Chr.11: 94593529-94593529 on Build GRCh38
rs747285253 Chr.11: 94593686-94593686 on Build GRCh38
rs369955037 Chr.11: 94593540-94593540 on Build GRCh38
rs369047221 Chr.11: 94593679-94593679 on Build GRCh38
rs772302203 Chr.11: 94593508-94593508 on Build GRCh38
rs775601221 Chr.11: 94593510-94593510 on Build GRCh38
rs755832586 Chr.11: 94593553-94593553 on Build GRCh38
rs11020852 Chr.11: 94593682-94593682 on Build GRCh38
rs575492946 Chr.11: 94593538-94593538 on Build GRCh38
rs745979781 Chr.11: 94593499-94593499 on Build GRCh38
rs757188432 Chr.11: 94593596-94593596 on Build GRCh38
rs769054444 Chr.11: 94593646-94593646 on Build GRCh38
rs753682337 Chr.11: 94593587-94593587 on Build GRCh38
rs545301118 Chr.11: 94593496-94593496 on Build GRCh38
rs901844302 Chr.11: 94593669-94593669 on Build GRCh38
rs757027836 Chr.11: 94593688-94593688 on Build GRCh38
rs903807599 Chr.11: 94593699-94593699 on Build GRCh38
rs777109850 Chr.11: 94593652-94593652 on Build GRCh38
rs373486887 Chr.11: 94593543-94593543 on Build GRCh38
rs375761037 Chr.11: 94593664-94593664 on Build GRCh38
rs185092971 Chr.11: 94593537-94593537 on Build GRCh38
rs762392714 Chr.11: 94593535-94593535 on Build GRCh38

View Full Product Details