Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7698615-7698872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OVCH2
Entrez Gene ID
341277
Gene Name
ovochymase 2 (gene/pseudogene)
Gene Aliases
OVTN
UniGene
Hs.532475
Gene Chromosome Location
Chr.11: 7678623-7706894 on Build GRCh38
Transcripts
NM_198185.4 XM_017017659.1

SNP Detail

SNP ID Chromosome Location
rs184379352 Chr.11: 7698658-7698658 on Build GRCh38
rs765643024 Chr.11: 7698756-7698756 on Build GRCh38
rs564274084 Chr.11: 7698640-7698640 on Build GRCh38
rs750755009 Chr.11: 7698820-7698820 on Build GRCh38
rs752271233 Chr.11: 7698772-7698772 on Build GRCh38
rs961140933 Chr.11: 7698737-7698737 on Build GRCh38
rs887734113 Chr.11: 7698715-7698715 on Build GRCh38
rs764088511 Chr.11: 7698794-7698794 on Build GRCh38
rs769229247 Chr.11: 7698746-7698746 on Build GRCh38
rs747418646 Chr.11: 7698732-7698732 on Build GRCh38
rs371302518 Chr.11: 7698825-7698825 on Build GRCh38
rs776941277 Chr.11: 7698750-7698750 on Build GRCh38
rs773982264 Chr.11: 7698767-7698767 on Build GRCh38
rs549130597 Chr.11: 7698819-7698819 on Build GRCh38
rs759301822 Chr.11: 7698768-7698768 on Build GRCh38
rs73400743 Chr.11: 7698791-7698791 on Build GRCh38
rs547233634 Chr.11: 7698665-7698665 on Build GRCh38
rs766878465 Chr.11: 7698769-7698769 on Build GRCh38
rs971233763 Chr.11: 7698754-7698754 on Build GRCh38
rs566885018 Chr.11: 7698782-7698782 on Build GRCh38
rs569199568 Chr.11: 7698826-7698826 on Build GRCh38
rs746149384 Chr.11: 7698714-7698714 on Build GRCh38
rs762138231 Chr.11: 7698751-7698751 on Build GRCh38
rs368901937 Chr.11: 7698818-7698818 on Build GRCh38
rs757104381 Chr.11: 7698805-7698805 on Build GRCh38
rs567095797 Chr.11: 7698822-7698822 on Build GRCh38
rs779254900 Chr.11: 7698705-7698705 on Build GRCh38
rs546805271 Chr.11: 7698698-7698698 on Build GRCh38
rs983844819 Chr.11: 7698670-7698670 on Build GRCh38
rs867359609 Chr.11: 7698749-7698749 on Build GRCh38
rs955761667 Chr.11: 7698838-7698838 on Build GRCh38
rs775645915 Chr.11: 7698722-7698722 on Build GRCh38
rs753873959 Chr.11: 7698800-7698800 on Build GRCh38
rs370844182 Chr.11: 7698745-7698745 on Build GRCh38
rs986805479 Chr.11: 7698801-7698801 on Build GRCh38

View Full Product Details