Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113319560-113319793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMP1
Entrez Gene ID
3916
Gene Name
lysosomal associated membrane protein 1
Gene Aliases
CD107a, LAMPA, LGP120
UniGene
Hs.494419
Gene Chromosome Location
Chr.13: 113297154-113323431 on Build GRCh38
Transcripts
NM_005561.3 XM_011537494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4919783 c.705C>T p.S235S Chr.13: 113319611-113319611 on Build GRCh38
3718826 c.747_750+35del39 p.? Chr.13: 113319653-113319691 on Build GRCh38
7233332 c.670G>T p.V224L Chr.13: 113319576-113319576 on Build GRCh38
945586 c.744C>T p.D248D Chr.13: 113319650-113319650 on Build GRCh38
35824 c.750+1G>A p.? Chr.13: 113319657-113319657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761898603 Chr.13: 113319583-113319583 on Build GRCh38
rs902957085 Chr.13: 113319731-113319731 on Build GRCh38
rs1009065022 Chr.13: 113319718-113319718 on Build GRCh38
rs781761301 Chr.13: 113319654-113319654 on Build GRCh38
rs761179281 Chr.13: 113319666-113319666 on Build GRCh38
rs763038744 Chr.13: 113319596-113319596 on Build GRCh38
rs768386237 Chr.13: 113319692-113319692 on Build GRCh38
rs1041417535 Chr.13: 113319717-113319717 on Build GRCh38
rs926829981 Chr.13: 113319674-113319674 on Build GRCh38
rs1005413564 Chr.13: 113319588-113319588 on Build GRCh38
rs369885554 Chr.13: 113319695-113319695 on Build GRCh38
rs201866480 Chr.13: 113319667-113319667 on Build GRCh38
rs778525208 Chr.13: 113319668-113319668 on Build GRCh38
rs200017606 Chr.13: 113319677-113319677 on Build GRCh38
rs771598801 Chr.13: 113319681-113319681 on Build GRCh38
rs776754723 Chr.13: 113319582-113319582 on Build GRCh38
rs908531092 Chr.13: 113319642-113319642 on Build GRCh38
rs567560779 Chr.13: 113319643-113319643 on Build GRCh38
rs761779089 Chr.13: 113319691-113319691 on Build GRCh38
rs753196309 Chr.13: 113319662-113319662 on Build GRCh38
rs752826527 Chr.13: 113319752-113319752 on Build GRCh38
rs746772656 Chr.13: 113319690-113319690 on Build GRCh38
rs147481309 Chr.13: 113319746-113319746 on Build GRCh38
rs565799075 Chr.13: 113319716-113319716 on Build GRCh38
rs563136954 Chr.13: 113319653-113319691 on Build GRCh38
rs548996606 Chr.13: 113319586-113319586 on Build GRCh38
rs755576074 Chr.13: 113319653-113319653 on Build GRCh38
rs750678018 Chr.13: 113319590-113319590 on Build GRCh38
rs377759076 Chr.13: 113319694-113319694 on Build GRCh38
rs191041858 Chr.13: 113319689-113319689 on Build GRCh38
rs760712195 Chr.13: 113319581-113319581 on Build GRCh38
rs143656034 Chr.13: 113319736-113319736 on Build GRCh38
rs369669372 Chr.13: 113319634-113319634 on Build GRCh38
rs745553838 Chr.13: 113319670-113319670 on Build GRCh38
rs533358425 Chr.13: 113319661-113319661 on Build GRCh38
rs570143580 Chr.13: 113319739-113319739 on Build GRCh38
rs539527803 Chr.13: 113319735-113319735 on Build GRCh38

View Full Product Details