Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21726895-21727144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF100
Entrez Gene ID
163227
Gene Name
zinc finger protein 100
Gene Aliases
-
UniGene
Hs.365142
Gene Chromosome Location
Chr.19: 21724041-21767628 on Build GRCh38
Transcripts
NM_173531.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6827822 c.1276G>T p.E426* Chr.19: 21727036-21727036 on Build GRCh38
5635139 c.1385G>A p.C462Y Chr.19: 21726927-21726927 on Build GRCh38
993708 c.1262G>A p.R421K Chr.19: 21727050-21727050 on Build GRCh38
6827823 c.1279A>T p.K427* Chr.19: 21727033-21727033 on Build GRCh38
6827825 c.1284C>T p.P428P Chr.19: 21727028-21727028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749261986 Chr.19: 21726925-21726933 on Build GRCh38
rs764975995 Chr.19: 21726946-21726946 on Build GRCh38
rs773752537 Chr.19: 21727002-21727002 on Build GRCh38
rs748335273 Chr.19: 21727026-21727027 on Build GRCh38
rs748435196 Chr.19: 21727059-21727059 on Build GRCh38
rs768269272 Chr.19: 21726955-21726956 on Build GRCh38
rs779269607 Chr.19: 21727056-21727056 on Build GRCh38
rs768043951 Chr.19: 21726918-21726918 on Build GRCh38
rs530787118 Chr.19: 21727047-21727047 on Build GRCh38
rs371546506 Chr.19: 21727103-21727103 on Build GRCh38
rs753431110 Chr.19: 21726939-21726939 on Build GRCh38
rs780236096 Chr.19: 21727052-21727052 on Build GRCh38
rs759138843 Chr.19: 21727021-21727021 on Build GRCh38
rs755146922 Chr.19: 21727055-21727055 on Build GRCh38
rs561185850 Chr.19: 21727053-21727053 on Build GRCh38
rs763809624 Chr.19: 21727041-21727041 on Build GRCh38
rs774761289 Chr.19: 21726987-21726987 on Build GRCh38
rs773453580 Chr.19: 21727068-21727068 on Build GRCh38
rs543112236 Chr.19: 21727050-21727050 on Build GRCh38
rs748490199 Chr.19: 21726994-21726994 on Build GRCh38
rs769025706 Chr.19: 21726984-21726984 on Build GRCh38
rs761719206 Chr.19: 21727093-21727093 on Build GRCh38
rs372931531 Chr.19: 21726980-21726980 on Build GRCh38
rs202172761 Chr.19: 21726949-21726949 on Build GRCh38
rs546924864 Chr.19: 21727080-21727080 on Build GRCh38
rs751337504 Chr.19: 21727092-21727092 on Build GRCh38
rs759044950 Chr.19: 21727071-21727071 on Build GRCh38
rs777170062 Chr.19: 21727070-21727070 on Build GRCh38
rs72479598 Chr.19: 21726931-21726931 on Build GRCh38
rs772403996 Chr.19: 21727049-21727050 on Build GRCh38
rs765848309 Chr.19: 21727011-21727011 on Build GRCh38
rs879149939 Chr.19: 21726995-21726995 on Build GRCh38
rs758169696 Chr.19: 21726955-21726955 on Build GRCh38
rs115256934 Chr.19: 21727073-21727073 on Build GRCh38
rs199672005 Chr.19: 21727086-21727086 on Build GRCh38
rs775066535 Chr.19: 21727074-21727074 on Build GRCh38
rs147769781 Chr.19: 21727094-21727094 on Build GRCh38
rs878905947 Chr.19: 21726974-21726974 on Build GRCh38
rs772418956 Chr.19: 21727064-21727064 on Build GRCh38
rs772382628 Chr.19: 21726996-21726996 on Build GRCh38
rs762516981 Chr.19: 21727076-21727076 on Build GRCh38
rs746977407 Chr.19: 21726965-21726965 on Build GRCh38
rs368185825 Chr.19: 21727015-21727015 on Build GRCh38
rs544889931 Chr.19: 21726978-21726978 on Build GRCh38
rs773610867 Chr.19: 21727063-21727069 on Build GRCh38
rs879023817 Chr.19: 21727001-21727001 on Build GRCh38
rs533592689 Chr.19: 21726932-21726932 on Build GRCh38
rs747509710 Chr.19: 21727069-21727069 on Build GRCh38
rs78722705 Chr.19: 21726990-21726990 on Build GRCh38
rs754628199 Chr.19: 21726942-21726942 on Build GRCh38
rs775186467 Chr.19: 21727028-21727028 on Build GRCh38
rs752633317 Chr.19: 21726972-21726973 on Build GRCh38
rs185406570 Chr.19: 21727026-21727026 on Build GRCh38
rs532890971 Chr.19: 21727113-21727113 on Build GRCh38
rs756161010 Chr.19: 21726975-21726975 on Build GRCh38
rs374252867 Chr.19: 21727005-21727005 on Build GRCh38

View Full Product Details