Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58645423-58645696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT1
Entrez Gene ID
51339
Gene Name
dishevelled binding antagonist of beta catenin 1
Gene Aliases
DAPPER, DAPPER1, DPR1, FRODO, HDPR1, THYEX3
UniGene
Hs.48950
Gene Chromosome Location
Chr.14: 58634068-58648321 on Build GRCh38
Transcripts
NM_001079520.1 NM_016651.5 NR_046093.1 NR_046095.1 XM_006720167.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6639878 c.963G>A p.P321P Chr.14: 58645586-58645586 on Build GRCh38
6028128 c.1042delA p.T350fs*4 Chr.14: 58645665-58645665 on Build GRCh38
956584 c.803G>A p.R268H Chr.14: 58645426-58645426 on Build GRCh38
7242435 c.908G>A p.S303N Chr.14: 58645531-58645531 on Build GRCh38
1370277 c.1048A>G p.T350A Chr.14: 58645671-58645671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752793412 Chr.14: 58645533-58645533 on Build GRCh38
rs749119286 Chr.14: 58645562-58645562 on Build GRCh38
rs951091466 Chr.14: 58645592-58645592 on Build GRCh38
rs751970499 Chr.14: 58645495-58645495 on Build GRCh38
rs763171388 Chr.14: 58645493-58645493 on Build GRCh38
rs369545745 Chr.14: 58645463-58645463 on Build GRCh38
rs141135250 Chr.14: 58645551-58645551 on Build GRCh38
rs1002410963 Chr.14: 58645526-58645526 on Build GRCh38
rs574353920 Chr.14: 58645586-58645586 on Build GRCh38
rs892486399 Chr.14: 58645484-58645484 on Build GRCh38
rs751930178 Chr.14: 58645607-58645607 on Build GRCh38
rs760167392 Chr.14: 58645585-58645585 on Build GRCh38
rs756469227 Chr.14: 58645543-58645543 on Build GRCh38
rs776202159 Chr.14: 58645595-58645595 on Build GRCh38
rs952965134 Chr.14: 58645565-58645565 on Build GRCh38
rs772018481 Chr.14: 58645579-58645579 on Build GRCh38
rs373250796 Chr.14: 58645476-58645476 on Build GRCh38
rs76254314 Chr.14: 58645458-58645458 on Build GRCh38
rs766603971 Chr.14: 58645599-58645599 on Build GRCh38
rs147388110 Chr.14: 58645445-58645445 on Build GRCh38
rs768312424 Chr.14: 58645593-58645593 on Build GRCh38
rs760001060 Chr.14: 58645612-58645612 on Build GRCh38
rs77323631 Chr.14: 58645503-58645503 on Build GRCh38
rs961450224 Chr.14: 58645594-58645594 on Build GRCh38
rs760365379 Chr.14: 58645478-58645478 on Build GRCh38
rs756201454 Chr.14: 58645658-58645658 on Build GRCh38
rs139223977 Chr.14: 58645664-58645664 on Build GRCh38
rs201127318 Chr.14: 58645580-58645580 on Build GRCh38
rs375963448 Chr.14: 58645494-58645494 on Build GRCh38
rs778143201 Chr.14: 58645659-58645659 on Build GRCh38
rs770542487 Chr.14: 58645447-58645447 on Build GRCh38
rs768131365 Chr.14: 58645614-58645614 on Build GRCh38
rs147190910 Chr.14: 58645670-58645670 on Build GRCh38
rs138519520 Chr.14: 58645456-58645456 on Build GRCh38
rs752881298 Chr.14: 58645637-58645637 on Build GRCh38
rs139175989 Chr.14: 58645577-58645577 on Build GRCh38
rs763406526 Chr.14: 58645597-58645597 on Build GRCh38
rs1019368076 Chr.14: 58645531-58645531 on Build GRCh38
rs763734181 Chr.14: 58645483-58645483 on Build GRCh38
rs201587878 Chr.14: 58645467-58645467 on Build GRCh38
rs371662684 Chr.14: 58645486-58645486 on Build GRCh38
rs781470708 Chr.14: 58645529-58645529 on Build GRCh38
rs150721565 Chr.14: 58645563-58645563 on Build GRCh38

View Full Product Details