Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 116435797-116436022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSE
Entrez Gene ID
29940
Gene Name
dermatan sulfate epimerase
Gene Aliases
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2
UniGene
Hs.458358
Gene Chromosome Location
Chr.6: 116254152-116441261 on Build GRCh38
Transcripts
NM_001080976.2 NM_001322937.1 NM_001322938.1 NM_001322939.1 NM_001322940.1 NM_001322941.1 NM_001322943.1 NM_001322944.1 NM_013352.3 NR_136524.1 XM_017010795.1 XM_017010796.1 XM_017010797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6053456 c.1476G>T p.K492N Chr.6: 116435944-116435944 on Build GRCh38
328890 c.1541A>T p.K514M Chr.6: 116436009-116436009 on Build GRCh38
6053457 c.1533G>T p.K511N Chr.6: 116435944-116435944 on Build GRCh38
7349491 c.1498G>C p.G500R Chr.6: 116435966-116435966 on Build GRCh38
7102307 c.1599G>C p.K533N Chr.6: 116436010-116436010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750974183 Chr.6: 116435920-116435920 on Build GRCh38
rs149181929 Chr.6: 116435899-116435899 on Build GRCh38
rs867058537 Chr.6: 116435897-116435897 on Build GRCh38
rs753845781 Chr.6: 116435900-116435900 on Build GRCh38
rs758790510 Chr.6: 116435926-116435926 on Build GRCh38
rs376824800 Chr.6: 116435946-116435946 on Build GRCh38
rs181711422 Chr.6: 116435849-116435849 on Build GRCh38
rs750453583 Chr.6: 116435968-116435968 on Build GRCh38
rs751463171 Chr.6: 116435847-116435847 on Build GRCh38
rs374010235 Chr.6: 116435887-116435887 on Build GRCh38
rs950301129 Chr.6: 116435833-116435833 on Build GRCh38
rs780931991 Chr.6: 116435974-116435974 on Build GRCh38
rs200485959 Chr.6: 116435851-116435851 on Build GRCh38
rs766127141 Chr.6: 116435852-116435852 on Build GRCh38
rs757356537 Chr.6: 116435905-116435905 on Build GRCh38
rs773305945 Chr.6: 116435850-116435850 on Build GRCh38
rs267600774 Chr.6: 116435928-116435928 on Build GRCh38
rs144957600 Chr.6: 116435856-116435856 on Build GRCh38
rs780344963 Chr.6: 116435940-116435940 on Build GRCh38
rs768130032 Chr.6: 116435959-116435959 on Build GRCh38
rs779340056 Chr.6: 116435914-116435914 on Build GRCh38
rs760862778 Chr.6: 116435862-116435862 on Build GRCh38
rs200018749 Chr.6: 116435980-116435980 on Build GRCh38

View Full Product Details