Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203128687-203128946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA1
Entrez Gene ID
134
Gene Name
adenosine A1 receptor
Gene Aliases
RDC7
UniGene
Hs.77867
Gene Chromosome Location
Chr.1: 203127705-203167405 on Build GRCh38
Transcripts
NM_000674.2 NM_001048230.1 XM_005244901.1 XM_005244902.4 XM_017000342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6595311 c.55A>G p.I19V Chr.1: 203128896-203128896 on Build GRCh38
3976871 c.81C>A p.N27K Chr.1: 203128922-203128922 on Build GRCh38
3976872 c.82G>A p.V28M Chr.1: 203128923-203128923 on Build GRCh38
6595306 c.3G>A p.M1I Chr.1: 203128844-203128844 on Build GRCh38
5106011 c.4C>T p.P2S Chr.1: 203128845-203128845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762019552 Chr.1: 203128840-203128840 on Build GRCh38
rs774277573 Chr.1: 203128839-203128839 on Build GRCh38
rs201436867 Chr.1: 203128811-203128811 on Build GRCh38
rs778264891 Chr.1: 203128798-203128798 on Build GRCh38
rs770874262 Chr.1: 203128808-203128808 on Build GRCh38
rs752310008 Chr.1: 203128800-203128800 on Build GRCh38
rs202220505 Chr.1: 203128848-203128848 on Build GRCh38
rs200737401 Chr.1: 203128739-203128739 on Build GRCh38
rs148895280 Chr.1: 203128872-203128872 on Build GRCh38
rs201916217 Chr.1: 203128752-203128752 on Build GRCh38
rs201792430 Chr.1: 203128804-203128804 on Build GRCh38
rs766701231 Chr.1: 203128850-203128850 on Build GRCh38
rs1037615563 Chr.1: 203128738-203128738 on Build GRCh38
rs149147396 Chr.1: 203128801-203128801 on Build GRCh38
rs749081064 Chr.1: 203128862-203128862 on Build GRCh38
rs1002805942 Chr.1: 203128815-203128815 on Build GRCh38
rs760024197 Chr.1: 203128877-203128877 on Build GRCh38
rs752086617 Chr.1: 203128916-203128916 on Build GRCh38
rs777489656 Chr.1: 203128803-203128803 on Build GRCh38
rs898990633 Chr.1: 203128771-203128771 on Build GRCh38
rs766811799 Chr.1: 203128794-203128794 on Build GRCh38
rs11547177 Chr.1: 203128849-203128849 on Build GRCh38
rs755519862 Chr.1: 203128797-203128797 on Build GRCh38
rs754376012 Chr.1: 203128795-203128795 on Build GRCh38
rs201138442 Chr.1: 203128923-203128923 on Build GRCh38
rs1019929241 Chr.1: 203128845-203128845 on Build GRCh38
rs199912976 Chr.1: 203128814-203128814 on Build GRCh38
rs1032621076 Chr.1: 203128882-203128882 on Build GRCh38
rs199986518 Chr.1: 203128922-203128922 on Build GRCh38
rs369808639 Chr.1: 203128838-203128838 on Build GRCh38
rs892803005 Chr.1: 203128743-203128743 on Build GRCh38
rs1001404620 Chr.1: 203128787-203128787 on Build GRCh38
rs978339398 Chr.1: 203128871-203128871 on Build GRCh38
rs940576915 Chr.1: 203128725-203128725 on Build GRCh38
rs61731145 Chr.1: 203128847-203128847 on Build GRCh38
rs765840235 Chr.1: 203128881-203128881 on Build GRCh38
rs374400699 Chr.1: 203128853-203128853 on Build GRCh38

View Full Product Details