Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 19569351-19569624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf62
Entrez Gene ID
57020
Gene Name
chromosome 16 open reading frame 62
Gene Aliases
-
UniGene
Hs.654964
Gene Chromosome Location
Chr.16: 19555415-19701163 on Build GRCh38
Transcripts
NM_001300743.1 NM_020314.5 XM_005255435.3 XM_006721065.3 XM_011545896.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3888037 c.504C>T p.L168L Chr.16: 19569543-19569543 on Build GRCh38
3370337 c.516A>C p.A172A Chr.16: 19569555-19569555 on Build GRCh38
967911 c.504C>A p.L168L Chr.16: 19569543-19569543 on Build GRCh38
5450729 c.214G>A p.V72I Chr.16: 19569520-19569520 on Build GRCh38
6555467 c.195G>T p.V65V Chr.16: 19569501-19569501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113403724 Chr.16: 19569422-19569422 on Build GRCh38
rs121235 Chr.16: 19569377-19569377 on Build GRCh38
rs766144664 Chr.16: 19569499-19569499 on Build GRCh38
rs1031735086 Chr.16: 19569599-19569599 on Build GRCh38
rs774809331 Chr.16: 19569454-19569454 on Build GRCh38
rs751276255 Chr.16: 19569402-19569402 on Build GRCh38
rs919102879 Chr.16: 19569431-19569431 on Build GRCh38
rs772383799 Chr.16: 19569469-19569469 on Build GRCh38
rs751422794 Chr.16: 19569501-19569501 on Build GRCh38
rs369135032 Chr.16: 19569555-19569555 on Build GRCh38
rs750292712 Chr.16: 19569478-19569480 on Build GRCh38
rs755766433 Chr.16: 19569514-19569514 on Build GRCh38
rs764737854 Chr.16: 19569475-19569476 on Build GRCh38
rs774148780 Chr.16: 19569605-19569605 on Build GRCh38
rs776736181 Chr.16: 19569564-19569564 on Build GRCh38
rs750366124 Chr.16: 19569392-19569392 on Build GRCh38
rs768830249 Chr.16: 19569562-19569562 on Build GRCh38
rs146118670 Chr.16: 19569498-19569498 on Build GRCh38
rs760821274 Chr.16: 19569477-19569477 on Build GRCh38
rs370569405 Chr.16: 19569571-19569571 on Build GRCh38
rs775686394 Chr.16: 19569471-19569471 on Build GRCh38
rs758333386 Chr.16: 19569600-19569600 on Build GRCh38
rs746313792 Chr.16: 19569526-19569526 on Build GRCh38
rs777349163 Chr.16: 19569447-19569447 on Build GRCh38
rs553777201 Chr.16: 19569386-19569386 on Build GRCh38
rs373022657 Chr.16: 19569396-19569396 on Build GRCh38
rs753521367 Chr.16: 19569394-19569394 on Build GRCh38
rs747254310 Chr.16: 19569557-19569557 on Build GRCh38
rs746186181 Chr.16: 19569455-19569455 on Build GRCh38
rs764172168 Chr.16: 19569497-19569497 on Build GRCh38
rs1057422420 Chr.16: 19569472-19569472 on Build GRCh38
rs554713753 Chr.16: 19569509-19569509 on Build GRCh38
rs375300632 Chr.16: 19569519-19569519 on Build GRCh38
rs767341890 Chr.16: 19569510-19569510 on Build GRCh38
rs755678441 Chr.16: 19569442-19569442 on Build GRCh38
rs754703041 Chr.16: 19569416-19569416 on Build GRCh38
rs749822818 Chr.16: 19569448-19569448 on Build GRCh38
rs772328728 Chr.16: 19569543-19569543 on Build GRCh38
rs938489146 Chr.16: 19569446-19569446 on Build GRCh38
rs201207645 Chr.16: 19569388-19569388 on Build GRCh38
rs747715306 Chr.16: 19569437-19569437 on Build GRCh38
rs761780737 Chr.16: 19569570-19569570 on Build GRCh38
rs757822505 Chr.16: 19569523-19569523 on Build GRCh38
rs756709751 Chr.16: 19569421-19569422 on Build GRCh38
rs748706506 Chr.16: 19569520-19569520 on Build GRCh38
rs779264667 Chr.16: 19569525-19569525 on Build GRCh38
rs938526744 Chr.16: 19569529-19569529 on Build GRCh38

View Full Product Details