Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49893985-49894228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF6
Entrez Gene ID
382
Gene Name
ADP ribosylation factor 6
Gene Aliases
-
UniGene
Hs.525330
Gene Chromosome Location
Chr.14: 49893018-49897054 on Build GRCh38
Transcripts
NM_001663.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5680498 c.284G>T p.R95L Chr.14: 49894020-49894020 on Build GRCh38
955980 c.415C>A p.L139M Chr.14: 49894151-49894151 on Build GRCh38
7275625 c.313C>T p.R105C Chr.14: 49894049-49894049 on Build GRCh38
1748860 c.462C>T p.S154S Chr.14: 49894198-49894198 on Build GRCh38
4848300 c.433C>T p.R145W Chr.14: 49894169-49894169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749233938 Chr.14: 49894090-49894090 on Build GRCh38
rs751003183 Chr.14: 49894051-49894051 on Build GRCh38
rs3100898 Chr.14: 49894043-49894043 on Build GRCh38
rs61755571 Chr.14: 49894136-49894136 on Build GRCh38
rs61755574 Chr.14: 49894188-49894188 on Build GRCh38
rs376018460 Chr.14: 49894138-49894138 on Build GRCh38
rs201401465 Chr.14: 49894093-49894093 on Build GRCh38
rs575053502 Chr.14: 49894075-49894075 on Build GRCh38
rs61755569 Chr.14: 49894078-49894078 on Build GRCh38
rs61755567 Chr.14: 49894034-49894034 on Build GRCh38
rs61755572 Chr.14: 49894163-49894163 on Build GRCh38
rs754637279 Chr.14: 49894114-49894114 on Build GRCh38
rs766708308 Chr.14: 49894087-49894087 on Build GRCh38
rs761927063 Chr.14: 49894022-49894022 on Build GRCh38
rs760817062 Chr.14: 49894009-49894009 on Build GRCh38
rs61755570 Chr.14: 49894097-49894097 on Build GRCh38
rs61755568 Chr.14: 49894050-49894050 on Build GRCh38
rs765630104 Chr.14: 49894028-49894028 on Build GRCh38
rs757158549 Chr.14: 49894186-49894186 on Build GRCh38
rs61755573 Chr.14: 49894170-49894170 on Build GRCh38
rs749309161 Chr.14: 49894157-49894157 on Build GRCh38
rs767302736 Chr.14: 49894117-49894117 on Build GRCh38
rs755693781 Chr.14: 49894132-49894132 on Build GRCh38
rs117373659 Chr.14: 49894063-49894063 on Build GRCh38
rs752233598 Chr.14: 49894124-49894124 on Build GRCh38
rs776640440 Chr.14: 49894021-49894021 on Build GRCh38
rs1026625998 Chr.14: 49894036-49894036 on Build GRCh38
rs138905564 Chr.14: 49894207-49894207 on Build GRCh38

View Full Product Details