Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 30445239-30445496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF536
Entrez Gene ID
9745
Gene Name
zinc finger protein 536
Gene Aliases
-
UniGene
Hs.378901
Gene Chromosome Location
Chr.19: 30224236-30713538 on Build GRCh38
Transcripts
NM_014717.2 XM_011527554.2 XM_011527555.2 XM_011527557.2 XM_011527558.2 XM_011527560.2 XM_017027527.1 XM_017027528.1 XM_017027529.1 XM_017027530.1 XM_017027531.1 XM_017027532.1 XM_017027533.1 XM_017027534.1 XM_017027535.1 XM_017027536.1 XM_017027537.1 XM_017027538.1 XM_017027539.1 XM_017027540.1 XM_017027541.1 XM_017027542.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1712110 c.1816G>A p.D606N Chr.19: 30445378-30445378 on Build GRCh38
6084714 c.1832G>T p.G611V Chr.19: 30445394-30445394 on Build GRCh38
3532056 c.1728G>A p.Q576Q Chr.19: 30445290-30445290 on Build GRCh38
3532058 c.1889C>T p.P630L Chr.19: 30445451-30445451 on Build GRCh38
77138 c.1819T>A p.F607I Chr.19: 30445381-30445381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201261663 Chr.19: 30445455-30445455 on Build GRCh38
rs199990247 Chr.19: 30445419-30445419 on Build GRCh38
rs7508667 Chr.19: 30445465-30445465 on Build GRCh38
rs201947801 Chr.19: 30445271-30445271 on Build GRCh38
rs781335953 Chr.19: 30445332-30445332 on Build GRCh38
rs375381239 Chr.19: 30445404-30445404 on Build GRCh38
rs777011363 Chr.19: 30445397-30445397 on Build GRCh38
rs762517451 Chr.19: 30445450-30445450 on Build GRCh38
rs368393123 Chr.19: 30445433-30445433 on Build GRCh38
rs780920726 Chr.19: 30445323-30445323 on Build GRCh38
rs752321746 Chr.19: 30445301-30445301 on Build GRCh38
rs771230573 Chr.19: 30445392-30445392 on Build GRCh38
rs767274722 Chr.19: 30445475-30445475 on Build GRCh38
rs761713867 Chr.19: 30445474-30445474 on Build GRCh38
rs748156582 Chr.19: 30445357-30445357 on Build GRCh38
rs866522049 Chr.19: 30445378-30445378 on Build GRCh38
rs764740243 Chr.19: 30445299-30445299 on Build GRCh38
rs763902544 Chr.19: 30445470-30445470 on Build GRCh38
rs141248439 Chr.19: 30445375-30445375 on Build GRCh38
rs757114293 Chr.19: 30445324-30445324 on Build GRCh38
rs758984861 Chr.19: 30445287-30445287 on Build GRCh38
rs751244018 Chr.19: 30445473-30445473 on Build GRCh38
rs969199848 Chr.19: 30445463-30445463 on Build GRCh38
rs747436182 Chr.19: 30445391-30445391 on Build GRCh38
rs762643337 Chr.19: 30445309-30445309 on Build GRCh38
rs998131158 Chr.19: 30445434-30445434 on Build GRCh38
rs771494276 Chr.19: 30445279-30445279 on Build GRCh38
rs750316142 Chr.19: 30445334-30445334 on Build GRCh38
rs953908363 Chr.19: 30445477-30445477 on Build GRCh38
rs866274492 Chr.19: 30445297-30445297 on Build GRCh38
rs150100779 Chr.19: 30445342-30445342 on Build GRCh38
rs763812644 Chr.19: 30445312-30445312 on Build GRCh38
rs992023772 Chr.19: 30445399-30445399 on Build GRCh38
rs61746682 Chr.19: 30445389-30445389 on Build GRCh38
rs779003416 Chr.19: 30445351-30445351 on Build GRCh38
rs925189920 Chr.19: 30445476-30445476 on Build GRCh38
rs777186073 Chr.19: 30445284-30445284 on Build GRCh38
rs769207359 Chr.19: 30445414-30445414 on Build GRCh38
rs760051175 Chr.19: 30445398-30445398 on Build GRCh38
rs1051849078 Chr.19: 30445269-30445269 on Build GRCh38

View Full Product Details