Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 37472513-37472744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK1A
Entrez Gene ID
1859
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1A
Gene Aliases
DYRK, DYRK1, HP86, MNB, MNBH, MRD7
UniGene
Hs.368240
Gene Chromosome Location
Chr.21: 37365790-37515376 on Build GRCh38
Transcripts
NM_001396.3 NM_101395.2 NM_130436.2 NM_130438.2 XM_006723976.3 XM_006723977.3 XM_006723978.3 XM_006723979.3 XM_011529482.2 XM_011529483.2 XM_011529484.2 XM_011529485.2 XM_017028284.1 XM_017028285.1 XM_017028286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5217729 c.46C>T p.R16W Chr.21: 37472719-37472719 on Build GRCh38
725180 c.11G>A p.G4E Chr.21: 37472684-37472684 on Build GRCh38
5834027 c.46C>T p.R16W Chr.21: 37472719-37472719 on Build GRCh38
444439 c.46C>T p.R16W Chr.21: 37472719-37472719 on Build GRCh38
122743 c.56C>T p.P19L Chr.21: 37472729-37472729 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369277619 Chr.21: 37472667-37472667 on Build GRCh38
rs749897117 Chr.21: 37472665-37472665 on Build GRCh38
rs778105063 Chr.21: 37472674-37472674 on Build GRCh38
rs1044452102 Chr.21: 37472632-37472632 on Build GRCh38
rs756356211 Chr.21: 37472613-37472613 on Build GRCh38
rs746036251 Chr.21: 37472590-37472590 on Build GRCh38
rs926715780 Chr.21: 37472630-37472630 on Build GRCh38
rs560378772 Chr.21: 37472673-37472673 on Build GRCh38
rs747715722 Chr.21: 37472680-37472681 on Build GRCh38
rs771919737 Chr.21: 37472713-37472713 on Build GRCh38
rs745895020 Chr.21: 37472712-37472712 on Build GRCh38
rs1057175147 Chr.21: 37472720-37472720 on Build GRCh38
rs776496374 Chr.21: 37472649-37472649 on Build GRCh38
rs972368714 Chr.21: 37472605-37472605 on Build GRCh38
rs541259005 Chr.21: 37472657-37472657 on Build GRCh38
rs537018575 Chr.21: 37472644-37472644 on Build GRCh38
rs778892348 Chr.21: 37472707-37472707 on Build GRCh38
rs923507635 Chr.21: 37472565-37472565 on Build GRCh38
rs778246194 Chr.21: 37472661-37472661 on Build GRCh38
rs181142211 Chr.21: 37472597-37472597 on Build GRCh38
rs929589627 Chr.21: 37472578-37472578 on Build GRCh38
rs764698320 Chr.21: 37472656-37472656 on Build GRCh38
rs373777907 Chr.21: 37472548-37472548 on Build GRCh38
rs930850981 Chr.21: 37472626-37472626 on Build GRCh38
rs548118886 Chr.21: 37472627-37472627 on Build GRCh38
rs761522106 Chr.21: 37472654-37472654 on Build GRCh38
rs201209586 Chr.21: 37472564-37472564 on Build GRCh38
rs757452222 Chr.21: 37472689-37472689 on Build GRCh38
rs780200883 Chr.21: 37472716-37472716 on Build GRCh38
rs1049753 Chr.21: 37472718-37472718 on Build GRCh38
rs754108504 Chr.21: 37472680-37472680 on Build GRCh38
rs753071130 Chr.21: 37472672-37472672 on Build GRCh38

View Full Product Details