Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89280420-89280671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD11
Entrez Gene ID
29123
Gene Name
ankyrin repeat domain 11
Gene Aliases
ANCO-1, ANCO1, LZ16, T13
UniGene
Hs.335003
Gene Chromosome Location
Chr.16: 89267619-89490561 on Build GRCh38
Transcripts
NM_001256182.1 NM_001256183.1 NM_013275.5 XM_006721181.1 XM_006721184.2 XM_011523051.2 XM_011523053.2 XM_011523054.2 XM_011523055.2 XM_011523056.2 XM_011523057.2 XM_017023182.1 XM_017023183.1 XM_017023184.1 XM_017023185.1 XM_017023186.1 XM_017023187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3691266 c.6085G>A p.V2029I Chr.16: 89280457-89280457 on Build GRCh38
6969756 c.6047C>T p.P2016L Chr.16: 89280495-89280495 on Build GRCh38
3889143 c.5998C>T p.L2000F Chr.16: 89280544-89280544 on Build GRCh38
3889144 c.5962C>T p.P1988S Chr.16: 89280580-89280580 on Build GRCh38
6259714 c.5961C>A p.F1987L Chr.16: 89280581-89280581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565960089 Chr.16: 89280571-89280571 on Build GRCh38
rs763152116 Chr.16: 89280603-89280603 on Build GRCh38
rs772203828 Chr.16: 89280476-89280477 on Build GRCh38
rs759965356 Chr.16: 89280608-89280608 on Build GRCh38
rs761151327 Chr.16: 89280617-89280617 on Build GRCh38
rs1005592913 Chr.16: 89280633-89280633 on Build GRCh38
rs745685018 Chr.16: 89280504-89280504 on Build GRCh38
rs758181637 Chr.16: 89280585-89280585 on Build GRCh38
rs746078253 Chr.16: 89280473-89280481 on Build GRCh38
rs757540026 Chr.16: 89280439-89280439 on Build GRCh38
rs746355740 Chr.16: 89280447-89280447 on Build GRCh38
rs747661700 Chr.16: 89280454-89280454 on Build GRCh38
rs780570915 Chr.16: 89280478-89280478 on Build GRCh38
rs746928698 Chr.16: 89280554-89280554 on Build GRCh38
rs778167926 Chr.16: 89280438-89280438 on Build GRCh38
rs1053257756 Chr.16: 89280531-89280531 on Build GRCh38
rs780892836 Chr.16: 89280648-89280648 on Build GRCh38
rs1032995945 Chr.16: 89280542-89280542 on Build GRCh38
rs773403872 Chr.16: 89280565-89280565 on Build GRCh38
rs763207005 Chr.16: 89280568-89280568 on Build GRCh38
rs766783032 Chr.16: 89280529-89280529 on Build GRCh38
rs113264953 Chr.16: 89280547-89280547 on Build GRCh38
rs748086945 Chr.16: 89280601-89280601 on Build GRCh38
rs767818765 Chr.16: 89280611-89280611 on Build GRCh38
rs375457115 Chr.16: 89280599-89280599 on Build GRCh38
rs774653782 Chr.16: 89280607-89280607 on Build GRCh38
rs377471988 Chr.16: 89280506-89280506 on Build GRCh38
rs756815181 Chr.16: 89280546-89280546 on Build GRCh38
rs755603365 Chr.16: 89280484-89280484 on Build GRCh38
rs780624241 Chr.16: 89280449-89280449 on Build GRCh38
rs768098760 Chr.16: 89280578-89280578 on Build GRCh38
rs976983352 Chr.16: 89280521-89280521 on Build GRCh38
rs750094695 Chr.16: 89280582-89280582 on Build GRCh38
rs776509679 Chr.16: 89280556-89280556 on Build GRCh38
rs771142694 Chr.16: 89280604-89280604 on Build GRCh38
rs373218212 Chr.16: 89280477-89280477 on Build GRCh38
rs549967314 Chr.16: 89280515-89280515 on Build GRCh38
rs151329092 Chr.16: 89280514-89280514 on Build GRCh38
rs547762539 Chr.16: 89280533-89280533 on Build GRCh38
rs748892371 Chr.16: 89280494-89280494 on Build GRCh38
rs761021002 Chr.16: 89280551-89280551 on Build GRCh38
rs200531141 Chr.16: 89280522-89280522 on Build GRCh38
rs777385823 Chr.16: 89280485-89280485 on Build GRCh38
rs542077760 Chr.16: 89280470-89280470 on Build GRCh38
rs564238731 Chr.16: 89280481-89280481 on Build GRCh38
rs953261599 Chr.16: 89280488-89280488 on Build GRCh38
rs765328496 Chr.16: 89280561-89280561 on Build GRCh38
rs140586593 Chr.16: 89280644-89280644 on Build GRCh38
rs368118594 Chr.16: 89280558-89280558 on Build GRCh38
rs199785661 Chr.16: 89280457-89280457 on Build GRCh38
rs760859183 Chr.16: 89280471-89280471 on Build GRCh38
rs775544970 Chr.16: 89280510-89280510 on Build GRCh38
rs534220503 Chr.16: 89280580-89280580 on Build GRCh38
rs747612277 Chr.16: 89280480-89280480 on Build GRCh38
rs779884158 Chr.16: 89280635-89280635 on Build GRCh38
rs60520302 Chr.16: 89280475-89280475 on Build GRCh38
rs560402669 Chr.16: 89280473-89280473 on Build GRCh38
rs199737737 Chr.16: 89280631-89280631 on Build GRCh38
rs929624389 Chr.16: 89280581-89280581 on Build GRCh38
rs774332859 Chr.16: 89280472-89280473 on Build GRCh38
rs752781169 Chr.16: 89280466-89280466 on Build GRCh38
rs189297725 Chr.16: 89280458-89280458 on Build GRCh38
rs771037147 Chr.16: 89280471-89280479 on Build GRCh38
rs759486921 Chr.16: 89280463-89280463 on Build GRCh38
rs768438701 Chr.16: 89280594-89280594 on Build GRCh38
rs755371317 Chr.16: 89280577-89280577 on Build GRCh38
rs775221712 Chr.16: 89280482-89280490 on Build GRCh38
rs753215685 Chr.16: 89280615-89280615 on Build GRCh38
rs1046804092 Chr.16: 89280482-89280482 on Build GRCh38
rs994275260 Chr.16: 89280620-89280620 on Build GRCh38
rs768652993 Chr.16: 89280555-89280555 on Build GRCh38
rs111596062 Chr.16: 89280455-89280455 on Build GRCh38
rs775491352 Chr.16: 89280553-89280553 on Build GRCh38
rs375589721 Chr.16: 89280518-89280518 on Build GRCh38
rs773948438 Chr.16: 89280501-89280501 on Build GRCh38
rs370272709 Chr.16: 89280552-89280552 on Build GRCh38
rs372396960 Chr.16: 89280590-89280590 on Build GRCh38
rs867065526 Chr.16: 89280612-89280612 on Build GRCh38
rs748832367 Chr.16: 89280545-89280545 on Build GRCh38
rs1001824505 Chr.16: 89280557-89280557 on Build GRCh38
rs770628197 Chr.16: 89280498-89280498 on Build GRCh38
rs749868016 Chr.16: 89280466-89280467 on Build GRCh38
rs202044071 Chr.16: 89280634-89280634 on Build GRCh38
rs779372391 Chr.16: 89280445-89280445 on Build GRCh38
rs779935496 Chr.16: 89280588-89280588 on Build GRCh38
rs563584451 Chr.16: 89280512-89280512 on Build GRCh38
rs766728029 Chr.16: 89280570-89280570 on Build GRCh38
rs745564262 Chr.16: 89280548-89280548 on Build GRCh38
rs767562546 Chr.16: 89280465-89280465 on Build GRCh38
rs777086823 Chr.16: 89280544-89280544 on Build GRCh38
rs753973951 Chr.16: 89280474-89280474 on Build GRCh38
rs769888687 Chr.16: 89280602-89280602 on Build GRCh38
rs761984613 Chr.16: 89280560-89280560 on Build GRCh38
rs1025761111 Chr.16: 89280626-89280626 on Build GRCh38
rs751266006 Chr.16: 89280643-89280643 on Build GRCh38
rs879253753 Chr.16: 89280526-89280527 on Build GRCh38
rs863223321 Chr.16: 89280588-89280589 on Build GRCh38
rs933089649 Chr.16: 89280525-89280525 on Build GRCh38

View Full Product Details