Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139053904-139054159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZC3HAV1
Entrez Gene ID
56829
Gene Name
zinc finger CCCH-type containing, antiviral 1
Gene Aliases
ARTD13, FLB6421, PARP13, ZAP, ZC3H2, ZC3HDC2
UniGene
Hs.133512
Gene Chromosome Location
Chr.7: 139043519-139109720 on Build GRCh38
Transcripts
NM_020119.3 XM_005250501.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2860496 c.2253G>A p.M751I Chr.7: 139054030-139054030 on Build GRCh38
7211231 c.2229A>C p.V743V Chr.7: 139054054-139054054 on Build GRCh38
1086084 c.2309A>G p.K770R Chr.7: 139053974-139053974 on Build GRCh38
7081847 c.2301C>T p.L767L Chr.7: 139053982-139053982 on Build GRCh38
4402502 c.2207T>G p.L736R Chr.7: 139054076-139054076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749789154 Chr.7: 139054077-139054077 on Build GRCh38
rs990654313 Chr.7: 139053998-139053998 on Build GRCh38
rs759318595 Chr.7: 139054121-139054121 on Build GRCh38
rs765093497 Chr.7: 139053956-139053956 on Build GRCh38
rs775926377 Chr.7: 139054073-139054073 on Build GRCh38
rs752678086 Chr.7: 139053975-139053975 on Build GRCh38
rs375707741 Chr.7: 139054119-139054119 on Build GRCh38
rs757301345 Chr.7: 139053977-139053977 on Build GRCh38
rs766475912 Chr.7: 139053943-139053943 on Build GRCh38
rs370576444 Chr.7: 139054016-139054016 on Build GRCh38
rs770225433 Chr.7: 139053933-139053933 on Build GRCh38
rs765069409 Chr.7: 139054123-139054123 on Build GRCh38
rs774380058 Chr.7: 139054101-139054101 on Build GRCh38
rs754750196 Chr.7: 139053955-139053955 on Build GRCh38
rs977453602 Chr.7: 139054088-139054088 on Build GRCh38
rs997358689 Chr.7: 139053969-139053969 on Build GRCh38
rs768665806 Chr.7: 139054078-139054078 on Build GRCh38
rs779096298 Chr.7: 139053984-139053984 on Build GRCh38
rs561451564 Chr.7: 139053948-139053948 on Build GRCh38
rs201452196 Chr.7: 139054013-139054013 on Build GRCh38
rs367910457 Chr.7: 139053947-139053947 on Build GRCh38
rs770127819 Chr.7: 139054059-139054059 on Build GRCh38
rs776007931 Chr.7: 139053935-139053935 on Build GRCh38
rs1003122409 Chr.7: 139054052-139054052 on Build GRCh38
rs753661827 Chr.7: 139053952-139053952 on Build GRCh38
rs201721838 Chr.7: 139054015-139054015 on Build GRCh38
rs777402962 Chr.7: 139054010-139054010 on Build GRCh38
rs753889325 Chr.7: 139053946-139053946 on Build GRCh38
rs761837124 Chr.7: 139054109-139054109 on Build GRCh38
rs757908127 Chr.7: 139053997-139053997 on Build GRCh38
rs746250062 Chr.7: 139054038-139054038 on Build GRCh38
rs780945445 Chr.7: 139054035-139054035 on Build GRCh38
rs767746007 Chr.7: 139054113-139054113 on Build GRCh38

View Full Product Details