Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 20407858-20408123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT3
Entrez Gene ID
10196
Gene Name
protein arginine methyltransferase 3
Gene Aliases
HRMT1L3
UniGene
Hs.152337
Gene Chromosome Location
Chr.11: 20387530-20510861 on Build GRCh38
Transcripts
NM_001145166.1 NM_001145167.1 NM_005788.3 XM_011519836.2 XM_017017081.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
925903 c.862G>T p.E288* Chr.11: 20408001-20408001 on Build GRCh38
925901 c.833C>T p.A278V Chr.11: 20407972-20407972 on Build GRCh38
1353170 c.772G>A p.V258I Chr.11: 20407911-20407911 on Build GRCh38
925902 c.834G>A p.A278A Chr.11: 20407973-20407973 on Build GRCh38
377834 c.794G>T p.G265V Chr.11: 20407933-20407933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144727080 Chr.11: 20407973-20407973 on Build GRCh38
rs746776660 Chr.11: 20407905-20407905 on Build GRCh38
rs769317675 Chr.11: 20408034-20408034 on Build GRCh38
rs762608745 Chr.11: 20407930-20407930 on Build GRCh38
rs761331262 Chr.11: 20407917-20407917 on Build GRCh38
rs1055102115 Chr.11: 20408087-20408087 on Build GRCh38
rs755342137 Chr.11: 20407890-20407891 on Build GRCh38
rs757368857 Chr.11: 20407996-20407996 on Build GRCh38
rs768084873 Chr.11: 20408074-20408074 on Build GRCh38
rs747986110 Chr.11: 20408032-20408032 on Build GRCh38
rs759917326 Chr.11: 20408064-20408064 on Build GRCh38
rs10741836 Chr.11: 20407908-20407908 on Build GRCh38
rs748393411 Chr.11: 20408043-20408044 on Build GRCh38
rs752994465 Chr.11: 20408077-20408077 on Build GRCh38
rs749368890 Chr.11: 20407995-20407995 on Build GRCh38
rs779069393 Chr.11: 20407943-20407944 on Build GRCh38
rs756571970 Chr.11: 20408084-20408084 on Build GRCh38
rs776508007 Chr.11: 20407913-20407913 on Build GRCh38
rs370807624 Chr.11: 20408060-20408060 on Build GRCh38
rs1016891323 Chr.11: 20407900-20407900 on Build GRCh38
rs368162185 Chr.11: 20408049-20408049 on Build GRCh38
rs201331276 Chr.11: 20407950-20407950 on Build GRCh38
rs746901328 Chr.11: 20408010-20408010 on Build GRCh38
rs766985909 Chr.11: 20407935-20407935 on Build GRCh38
rs779944972 Chr.11: 20408004-20408004 on Build GRCh38
rs755343797 Chr.11: 20407959-20407959 on Build GRCh38
rs775473835 Chr.11: 20407897-20407897 on Build GRCh38
rs978701143 Chr.11: 20408023-20408023 on Build GRCh38
rs139576705 Chr.11: 20407990-20407990 on Build GRCh38
rs767722630 Chr.11: 20407972-20407972 on Build GRCh38
rs768671334 Chr.11: 20408019-20408019 on Build GRCh38
rs144295044 Chr.11: 20408028-20408028 on Build GRCh38
rs377173644 Chr.11: 20407890-20407890 on Build GRCh38
rs756235169 Chr.11: 20407980-20407980 on Build GRCh38
rs917420324 Chr.11: 20408037-20408037 on Build GRCh38
rs764946301 Chr.11: 20407927-20407927 on Build GRCh38
rs772788055 Chr.11: 20407928-20407928 on Build GRCh38

View Full Product Details