Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72206822-72207049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKM
Entrez Gene ID
5315
Gene Name
pyruvate kinase, muscle
Gene Aliases
CTHBP, HEL-S-30, OIP3, PK3, PKM2, TCB, THBP1
UniGene
Hs.534770
Gene Chromosome Location
Chr.15: 72199029-72231624 on Build GRCh38
Transcripts
NM_001206796.2 NM_001206797.2 NM_001206798.2 NM_001206799.1 NM_001316318.1 NM_002654.5 NM_182470.3 NM_182471.3 XM_005254443.1 XM_005254445.4 XM_006720570.1 XM_011521670.2 XM_017022313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6747433 c.991C>T p.L331L Chr.15: 72206877-72206877 on Build GRCh38
6747434 c.946C>T p.L316L Chr.15: 72206877-72206877 on Build GRCh38
6747436 c.1024C>T p.R342W Chr.15: 72206844-72206844 on Build GRCh38
6747437 c.979C>T p.R327W Chr.15: 72206844-72206844 on Build GRCh38
5762613 c.988-5C>T p.? Chr.15: 72206885-72206885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759567784 Chr.15: 72206861-72206861 on Build GRCh38
rs755560581 Chr.15: 72206926-72206927 on Build GRCh38
rs956262421 Chr.15: 72207027-72207027 on Build GRCh38
rs750860797 Chr.15: 72206999-72207000 on Build GRCh38
rs878910008 Chr.15: 72206851-72206852 on Build GRCh38
rs1022637577 Chr.15: 72206896-72206896 on Build GRCh38
rs879228992 Chr.15: 72206884-72206884 on Build GRCh38
rs1012537263 Chr.15: 72206843-72206843 on Build GRCh38
rs776264536 Chr.15: 72206894-72206894 on Build GRCh38
rs565473191 Chr.15: 72206990-72206990 on Build GRCh38
rs769295120 Chr.15: 72206906-72206906 on Build GRCh38
rs1032241008 Chr.15: 72206913-72206913 on Build GRCh38
rs2959910 Chr.15: 72206852-72206852 on Build GRCh38
rs904324887 Chr.15: 72206966-72206966 on Build GRCh38
rs749574324 Chr.15: 72206846-72206846 on Build GRCh38
rs761579675 Chr.15: 72206857-72206857 on Build GRCh38
rs971136544 Chr.15: 72206902-72206902 on Build GRCh38
rs769086093 Chr.15: 72206849-72206849 on Build GRCh38
rs775234394 Chr.15: 72206908-72206908 on Build GRCh38
rs745380726 Chr.15: 72206903-72206903 on Build GRCh38
rs765420055 Chr.15: 72206869-72206876 on Build GRCh38
rs748631492 Chr.15: 72206883-72206883 on Build GRCh38
rs749700037 Chr.15: 72206847-72206847 on Build GRCh38
rs3021075 Chr.15: 72206851-72206851 on Build GRCh38
rs762565660 Chr.15: 72206910-72206910 on Build GRCh38
rs772769486 Chr.15: 72206890-72206890 on Build GRCh38
rs8192417 Chr.15: 72206911-72206911 on Build GRCh38
rs1034663548 Chr.15: 72207008-72207008 on Build GRCh38

View Full Product Details