Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121632001-121632240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCLS1
Entrez Gene ID
3059
Gene Name
hematopoietic cell-specific Lyn substrate 1
Gene Aliases
CTTNL, HS1, lckBP1, p75
UniGene
Hs.14601
Gene Chromosome Location
Chr.3: 121631398-121660948 on Build GRCh38
Transcripts
NM_001292041.1 NM_005335.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
727954 c.1323A>T p.G441G Chr.3: 121632102-121632102 on Build GRCh38
1418165 c.1300G>A p.V434M Chr.3: 121632125-121632125 on Build GRCh38
6544706 c.1257G>A p.P419P Chr.3: 121632168-121632168 on Build GRCh38
4054948 c.1256C>T p.P419L Chr.3: 121632169-121632169 on Build GRCh38
6579706 c.1324+4G>A p.? Chr.3: 121632097-121632097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199584417 Chr.3: 121632145-121632145 on Build GRCh38
rs971130288 Chr.3: 121632038-121632038 on Build GRCh38
rs761917549 Chr.3: 121632203-121632203 on Build GRCh38
rs774362322 Chr.3: 121632073-121632073 on Build GRCh38
rs41271409 Chr.3: 121632123-121632123 on Build GRCh38
rs751796699 Chr.3: 121632149-121632150 on Build GRCh38
rs778248397 Chr.3: 121632142-121632142 on Build GRCh38
rs748139355 Chr.3: 121632151-121632151 on Build GRCh38
rs147125943 Chr.3: 121632125-121632125 on Build GRCh38
rs200847413 Chr.3: 121632120-121632120 on Build GRCh38
rs112494135 Chr.3: 121632141-121632146 on Build GRCh38
rs748894989 Chr.3: 121632135-121632135 on Build GRCh38
rs371393467 Chr.3: 121632096-121632096 on Build GRCh38
rs945532274 Chr.3: 121632070-121632070 on Build GRCh38
rs9869984 Chr.3: 121632119-121632119 on Build GRCh38
rs774338969 Chr.3: 121632196-121632196 on Build GRCh38
rs768270380 Chr.3: 121632069-121632069 on Build GRCh38
rs528332003 Chr.3: 121632089-121632089 on Build GRCh38
rs768608716 Chr.3: 121632056-121632056 on Build GRCh38
rs756278135 Chr.3: 121632082-121632082 on Build GRCh38
rs748878546 Chr.3: 121632062-121632062 on Build GRCh38
rs181543689 Chr.3: 121632083-121632083 on Build GRCh38
rs775692474 Chr.3: 121632175-121632175 on Build GRCh38
rs375238578 Chr.3: 121632049-121632049 on Build GRCh38
rs767979007 Chr.3: 121632207-121632207 on Build GRCh38
rs763032774 Chr.3: 121632114-121632114 on Build GRCh38
rs201382649 Chr.3: 121632165-121632165 on Build GRCh38
rs763119132 Chr.3: 121632185-121632185 on Build GRCh38
rs375804712 Chr.3: 121632211-121632211 on Build GRCh38
rs368506277 Chr.3: 121632137-121632137 on Build GRCh38
rs779824969 Chr.3: 121632061-121632061 on Build GRCh38
rs757104479 Chr.3: 121632145-121632146 on Build GRCh38
rs371041353 Chr.3: 121632055-121632055 on Build GRCh38
rs553563399 Chr.3: 121632191-121632191 on Build GRCh38
rs781165290 Chr.3: 121632077-121632077 on Build GRCh38
rs754541179 Chr.3: 121632136-121632136 on Build GRCh38
rs753752592 Chr.3: 121632140-121632141 on Build GRCh38
rs376245237 Chr.3: 121632103-121632103 on Build GRCh38
rs374456259 Chr.3: 121632079-121632079 on Build GRCh38
rs147742489 Chr.3: 121632169-121632169 on Build GRCh38
rs777761340 Chr.3: 121632168-121632168 on Build GRCh38
rs764011681 Chr.3: 121632117-121632117 on Build GRCh38

View Full Product Details