Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49559349-49559596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCRS1
Entrez Gene ID
10445
Gene Name
microspherule protein 1
Gene Aliases
ICP22BP, INO80Q, MCRS2, MSP58, P78
UniGene
Hs.25313
Gene Chromosome Location
Chr.12: 49558294-49572157 on Build GRCh38
Transcripts
NM_001012300.1 NM_001278341.1 NM_006337.4 XM_005268572.3 XM_011537760.2 XM_017018689.1 XM_017018690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4782686 c.1064G>A p.R355Q Chr.12: 49559475-49559475 on Build GRCh38
1362124 c.1091G>A p.G364D Chr.12: 49559487-49559487 on Build GRCh38
6710069 c.1043-1G>T p.? Chr.12: 49559536-49559536 on Build GRCh38
5565718 c.1088G>A p.R363Q Chr.12: 49559490-49559490 on Build GRCh38
4785915 c.1052G>A p.G351D Chr.12: 49559487-49559487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374501190 Chr.12: 49559566-49559566 on Build GRCh38
rs753895615 Chr.12: 49559404-49559404 on Build GRCh38
rs766714316 Chr.12: 49559425-49559427 on Build GRCh38
rs746343401 Chr.12: 49559457-49559457 on Build GRCh38
rs908366750 Chr.12: 49559420-49559420 on Build GRCh38
rs201759154 Chr.12: 49559433-49559433 on Build GRCh38
rs755123283 Chr.12: 49559407-49559407 on Build GRCh38
rs1011547904 Chr.12: 49559507-49559507 on Build GRCh38
rs768368026 Chr.12: 49559475-49559475 on Build GRCh38
rs746455035 Chr.12: 49559559-49559559 on Build GRCh38
rs145685896 Chr.12: 49559523-49559523 on Build GRCh38
rs983948694 Chr.12: 49559406-49559406 on Build GRCh38
rs778940779 Chr.12: 49559414-49559414 on Build GRCh38
rs368024584 Chr.12: 49559434-49559434 on Build GRCh38
rs1002954405 Chr.12: 49559437-49559437 on Build GRCh38
rs774037141 Chr.12: 49559484-49559484 on Build GRCh38
rs907379205 Chr.12: 49559432-49559432 on Build GRCh38
rs61745830 Chr.12: 49559459-49559459 on Build GRCh38
rs771376185 Chr.12: 49559439-49559439 on Build GRCh38
rs372654661 Chr.12: 49559568-49559568 on Build GRCh38
rs751302296 Chr.12: 49559379-49559379 on Build GRCh38
rs752393849 Chr.12: 49559570-49559570 on Build GRCh38
rs764049768 Chr.12: 49559377-49559377 on Build GRCh38
rs372192833 Chr.12: 49559503-49559503 on Build GRCh38
rs761670646 Chr.12: 49559382-49559382 on Build GRCh38
rs200647259 Chr.12: 49559543-49559543 on Build GRCh38
rs1049767164 Chr.12: 49559393-49559393 on Build GRCh38
rs375912255 Chr.12: 49559577-49559577 on Build GRCh38
rs764340496 Chr.12: 49559545-49559545 on Build GRCh38
rs961236357 Chr.12: 49559421-49559421 on Build GRCh38
rs919869145 Chr.12: 49559430-49559430 on Build GRCh38
rs757693972 Chr.12: 49559548-49559548 on Build GRCh38
rs748954581 Chr.12: 49559470-49559470 on Build GRCh38
rs762682514 Chr.12: 49559370-49559370 on Build GRCh38
rs894453498 Chr.12: 49559480-49559480 on Build GRCh38
rs773249513 Chr.12: 49559515-49559515 on Build GRCh38
rs762746583 Chr.12: 49559392-49559392 on Build GRCh38
rs767440095 Chr.12: 49559402-49559402 on Build GRCh38
rs891947056 Chr.12: 49559554-49559554 on Build GRCh38
rs775135641 Chr.12: 49559464-49559464 on Build GRCh38
rs781745064 Chr.12: 49559440-49559440 on Build GRCh38
rs921602406 Chr.12: 49559384-49559384 on Build GRCh38
rs781400888 Chr.12: 49559553-49559553 on Build GRCh38
rs557620508 Chr.12: 49559570-49559570 on Build GRCh38
rs752971057 Chr.12: 49559418-49559418 on Build GRCh38
rs145122526 Chr.12: 49559424-49559424 on Build GRCh38
rs767766239 Chr.12: 49559516-49559516 on Build GRCh38
rs774469744 Chr.12: 49559408-49559408 on Build GRCh38
rs761808156 Chr.12: 49559511-49559511 on Build GRCh38
rs199947971 Chr.12: 49559547-49559547 on Build GRCh38
rs765364425 Chr.12: 49559518-49559518 on Build GRCh38

View Full Product Details