Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153975709-153975982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JTB
Entrez Gene ID
10899
Gene Name
jumping translocation breakpoint
Gene Aliases
HJTB, HSPC222, PAR, hJT
UniGene
Hs.6396
Gene Chromosome Location
Chr.1: 153974269-153977975 on Build GRCh38
Transcripts
NM_006694.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
423965 c.272delA p.N91fs*10 Chr.1: 153975838-153975838 on Build GRCh38
1667990 c.232G>T p.G78* Chr.1: 153975878-153975878 on Build GRCh38
5843758 c.233G>A p.G78E Chr.1: 153975877-153975877 on Build GRCh38
6901277 c.284+1G>T p.? Chr.1: 153975825-153975825 on Build GRCh38
3418092 c.274G>T p.E92* Chr.1: 153975836-153975836 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs893221509 Chr.1: 153975787-153975787 on Build GRCh38
rs756044036 Chr.1: 153975900-153975900 on Build GRCh38
rs547980518 Chr.1: 153975761-153975761 on Build GRCh38
rs945382686 Chr.1: 153975774-153975774 on Build GRCh38
rs201165351 Chr.1: 153975874-153975874 on Build GRCh38
rs372182903 Chr.1: 153975850-153975850 on Build GRCh38
rs912624870 Chr.1: 153975755-153975755 on Build GRCh38
rs762289192 Chr.1: 153975788-153975788 on Build GRCh38
rs373905373 Chr.1: 153975780-153975780 on Build GRCh38
rs566096106 Chr.1: 153975762-153975762 on Build GRCh38
rs748929536 Chr.1: 153975782-153975782 on Build GRCh38
rs775535090 Chr.1: 153975823-153975824 on Build GRCh38
rs775922210 Chr.1: 153975843-153975843 on Build GRCh38
rs772666276 Chr.1: 153975930-153975930 on Build GRCh38
rs778778863 Chr.1: 153975925-153975925 on Build GRCh38
rs372293933 Chr.1: 153975864-153975864 on Build GRCh38
rs7527963 Chr.1: 153975745-153975745 on Build GRCh38
rs1013736125 Chr.1: 153975920-153975920 on Build GRCh38
rs763085971 Chr.1: 153975884-153975884 on Build GRCh38
rs368694080 Chr.1: 153975846-153975846 on Build GRCh38
rs370674490 Chr.1: 153975922-153975922 on Build GRCh38
rs747502800 Chr.1: 153975952-153975952 on Build GRCh38
rs1003180750 Chr.1: 153975876-153975876 on Build GRCh38
rs143327674 Chr.1: 153975862-153975862 on Build GRCh38
rs986861335 Chr.1: 153975746-153975746 on Build GRCh38
rs537869728 Chr.1: 153975891-153975891 on Build GRCh38
rs995348255 Chr.1: 153975928-153975928 on Build GRCh38
rs745505224 Chr.1: 153975940-153975940 on Build GRCh38
rs377729852 Chr.1: 153975800-153975800 on Build GRCh38
rs777016179 Chr.1: 153975858-153975858 on Build GRCh38
rs771707949 Chr.1: 153975821-153975821 on Build GRCh38
rs760197674 Chr.1: 153975943-153975943 on Build GRCh38
rs1026768633 Chr.1: 153975941-153975941 on Build GRCh38
rs753617013 Chr.1: 153975918-153975918 on Build GRCh38
rs777456448 Chr.1: 153975911-153975911 on Build GRCh38
rs906916047 Chr.1: 153975942-153975942 on Build GRCh38
rs982467294 Chr.1: 153975958-153975958 on Build GRCh38
rs376498621 Chr.1: 153975932-153975932 on Build GRCh38
rs533702289 Chr.1: 153975763-153975763 on Build GRCh38
rs750504921 Chr.1: 153975878-153975878 on Build GRCh38
rs377514205 Chr.1: 153975897-153975897 on Build GRCh38
rs146392390 Chr.1: 153975887-153975887 on Build GRCh38
rs1015716920 Chr.1: 153975963-153975963 on Build GRCh38
rs745315790 Chr.1: 153975819-153975819 on Build GRCh38
rs779587667 Chr.1: 153975949-153975949 on Build GRCh38
rs958751102 Chr.1: 153975895-153975895 on Build GRCh38

View Full Product Details