Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 248295488-248295713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR2T12
Entrez Gene ID
127064
Gene Name
olfactory receptor family 2 subfamily T member 12
Gene Aliases
OR1-57
UniGene
Hs.553582
Gene Chromosome Location
Chr.1: 248294616-248295578 on Build GRCh38
Transcripts
NM_001004692.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1719784 c.52C>T p.H18Y Chr.1: 248295527-248295527 on Build GRCh38
5886084 c.40G>A p.G14R Chr.1: 248295539-248295539 on Build GRCh38
5846135 c.47T>A p.F16Y Chr.1: 248295532-248295532 on Build GRCh38
3943804 c.22C>A p.P8T Chr.1: 248295557-248295557 on Build GRCh38
4709840 c.69A>T p.Q23H Chr.1: 248295510-248295510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759100628 Chr.1: 248295575-248295575 on Build GRCh38
rs145359340 Chr.1: 248295528-248295528 on Build GRCh38
rs754367817 Chr.1: 248295540-248295540 on Build GRCh38
rs11339452 Chr.1: 248295574-248295574 on Build GRCh38
rs762104902 Chr.1: 248295562-248295562 on Build GRCh38
rs1023923138 Chr.1: 248295588-248295588 on Build GRCh38
rs796678700 Chr.1: 248295672-248295673 on Build GRCh38
rs766225405 Chr.1: 248295572-248295573 on Build GRCh38
rs771636236 Chr.1: 248295624-248295624 on Build GRCh38
rs745755236 Chr.1: 248295623-248295623 on Build GRCh38
rs758602974 Chr.1: 248295554-248295554 on Build GRCh38
rs972197299 Chr.1: 248295636-248295636 on Build GRCh38
rs374328676 Chr.1: 248295598-248295598 on Build GRCh38
rs200775570 Chr.1: 248295512-248295512 on Build GRCh38
rs779008430 Chr.1: 248295549-248295549 on Build GRCh38
rs780333423 Chr.1: 248295555-248295555 on Build GRCh38
rs774842761 Chr.1: 248295628-248295628 on Build GRCh38
rs370289766 Chr.1: 248295600-248295600 on Build GRCh38
rs777749915 Chr.1: 248295611-248295611 on Build GRCh38
rs764494820 Chr.1: 248295590-248295590 on Build GRCh38
rs113201762 Chr.1: 248295597-248295597 on Build GRCh38
rs961042615 Chr.1: 248295691-248295691 on Build GRCh38
rs140902647 Chr.1: 248295543-248295543 on Build GRCh38
rs115699723 Chr.1: 248295629-248295629 on Build GRCh38
rs149438533 Chr.1: 248295545-248295545 on Build GRCh38
rs756476760 Chr.1: 248295526-248295526 on Build GRCh38
rs368296827 Chr.1: 248295586-248295586 on Build GRCh38
rs773091021 Chr.1: 248295565-248295565 on Build GRCh38
rs773791152 Chr.1: 248295571-248295571 on Build GRCh38
rs775240833 Chr.1: 248295579-248295579 on Build GRCh38
rs140217475 Chr.1: 248295523-248295523 on Build GRCh38
rs986547266 Chr.1: 248295665-248295665 on Build GRCh38
rs779954848 Chr.1: 248295606-248295606 on Build GRCh38
rs770792464 Chr.1: 248295617-248295617 on Build GRCh38
rs867087384 Chr.1: 248295518-248295518 on Build GRCh38
rs751428300 Chr.1: 248295573-248295573 on Build GRCh38
rs370473498 Chr.1: 248295559-248295559 on Build GRCh38
rs755227064 Chr.1: 248295609-248295609 on Build GRCh38
rs536654792 Chr.1: 248295674-248295674 on Build GRCh38
rs770960178 Chr.1: 248295569-248295569 on Build GRCh38
rs552823206 Chr.1: 248295664-248295664 on Build GRCh38
rs779271580 Chr.1: 248295573-248295574 on Build GRCh38
rs201895737 Chr.1: 248295578-248295578 on Build GRCh38
rs747130777 Chr.1: 248295557-248295557 on Build GRCh38
rs749090410 Chr.1: 248295614-248295614 on Build GRCh38
rs74153116 Chr.1: 248295673-248295673 on Build GRCh38
rs557679170 Chr.1: 248295552-248295552 on Build GRCh38
rs376253948 Chr.1: 248295620-248295620 on Build GRCh38
rs902999585 Chr.1: 248295558-248295558 on Build GRCh38
rs933372898 Chr.1: 248295660-248295660 on Build GRCh38
rs11204626 Chr.1: 248295672-248295672 on Build GRCh38
rs1035116042 Chr.1: 248295690-248295690 on Build GRCh38
rs747255627 Chr.1: 248295608-248295608 on Build GRCh38
rs148984043 Chr.1: 248295659-248295659 on Build GRCh38

View Full Product Details