Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41916732-41917001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED20
Entrez Gene ID
9477
Gene Name
mediator complex subunit 20
Gene Aliases
PRO0213, TRFP
UniGene
Hs.278434
Gene Chromosome Location
Chr.6: 41905353-41921147 on Build GRCh38
Transcripts
NM_001305455.1 NM_001305456.1 NM_001305457.1 NM_004275.4 NR_131160.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7188089 c.112A>G p.T38A Chr.6: 41916842-41916842 on Build GRCh38
5615690 c.165C>G p.S55R Chr.6: 41916789-41916789 on Build GRCh38
3874045 c.66G>A p.E22E Chr.6: 41916888-41916888 on Build GRCh38
5570042 c.143C>T p.T48M Chr.6: 41916811-41916811 on Build GRCh38
3874044 c.144G>A p.T48T Chr.6: 41916810-41916810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923195549 Chr.6: 41916828-41916828 on Build GRCh38
rs745370482 Chr.6: 41916904-41916905 on Build GRCh38
rs373021133 Chr.6: 41916787-41916787 on Build GRCh38
rs771489139 Chr.6: 41916815-41916815 on Build GRCh38
rs762795138 Chr.6: 41916763-41916763 on Build GRCh38
rs948298931 Chr.6: 41916954-41916954 on Build GRCh38
rs767564473 Chr.6: 41916880-41916880 on Build GRCh38
rs778167430 Chr.6: 41916810-41916810 on Build GRCh38
rs778379107 Chr.6: 41916915-41916915 on Build GRCh38
rs746015751 Chr.6: 41916853-41916853 on Build GRCh38
rs150677099 Chr.6: 41916918-41916918 on Build GRCh38
rs201163704 Chr.6: 41916872-41916872 on Build GRCh38
rs758738114 Chr.6: 41916808-41916808 on Build GRCh38
rs769852552 Chr.6: 41916859-41916859 on Build GRCh38
rs775739222 Chr.6: 41916862-41916862 on Build GRCh38
rs148167617 Chr.6: 41916878-41916878 on Build GRCh38
rs767311771 Chr.6: 41916782-41916782 on Build GRCh38
rs145617243 Chr.6: 41916877-41916877 on Build GRCh38
rs763208484 Chr.6: 41916866-41916866 on Build GRCh38
rs985517128 Chr.6: 41916756-41916756 on Build GRCh38
rs750378983 Chr.6: 41916886-41916886 on Build GRCh38
rs774367486 Chr.6: 41916966-41916966 on Build GRCh38
rs769905909 Chr.6: 41916934-41916934 on Build GRCh38
rs775686255 Chr.6: 41916762-41916762 on Build GRCh38
rs747294349 Chr.6: 41916811-41916811 on Build GRCh38
rs746353113 Chr.6: 41916927-41916927 on Build GRCh38
rs751495058 Chr.6: 41916772-41916772 on Build GRCh38
rs781707850 Chr.6: 41916926-41916926 on Build GRCh38
rs773328304 Chr.6: 41916982-41916982 on Build GRCh38
rs986025920 Chr.6: 41916870-41916870 on Build GRCh38
rs761731450 Chr.6: 41916974-41916974 on Build GRCh38
rs748931812 Chr.6: 41916807-41916807 on Build GRCh38
rs781051637 Chr.6: 41916814-41916816 on Build GRCh38
rs140526856 Chr.6: 41916806-41916806 on Build GRCh38
rs41271269 Chr.6: 41916962-41916962 on Build GRCh38
rs369921112 Chr.6: 41916950-41916950 on Build GRCh38
rs777057932 Chr.6: 41916840-41916840 on Build GRCh38
rs142006963 Chr.6: 41916889-41916889 on Build GRCh38
rs376069545 Chr.6: 41916894-41916894 on Build GRCh38
rs201541518 Chr.6: 41916766-41916766 on Build GRCh38
rs754688891 Chr.6: 41916895-41916895 on Build GRCh38
rs765965809 Chr.6: 41916893-41916893 on Build GRCh38
rs763930938 Chr.6: 41916770-41916770 on Build GRCh38
rs747406107 Chr.6: 41916917-41916917 on Build GRCh38
rs757253766 Chr.6: 41916775-41916775 on Build GRCh38
rs771975922 Chr.6: 41916978-41916978 on Build GRCh38
rs769230044 Chr.6: 41916935-41916938 on Build GRCh38

View Full Product Details