Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153310500-153310773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGLYRP3
Entrez Gene ID
114771
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
PGLYRPIalpha, PGRP-Ialpha, PGRPIA
UniGene
Hs.348266
Gene Chromosome Location
Chr.1: 153297589-153312975 on Build GRCh38
Transcripts
NM_052891.2 XM_011509118.1 XM_011509119.1 XM_011509120.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6744660 c.41G>C p.G14A Chr.1: 153310625-153310625 on Build GRCh38
7302201 c.26C>G p.A9G Chr.1: 153310640-153310640 on Build GRCh38
5950552 c.42T>C p.G14G Chr.1: 153310624-153310624 on Build GRCh38
1472670 c.45C>G p.L15L Chr.1: 153310621-153310621 on Build GRCh38
5436141 c.12G>A p.L4L Chr.1: 153310654-153310654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374684250 Chr.1: 153310699-153310699 on Build GRCh38
rs201112896 Chr.1: 153310652-153310652 on Build GRCh38
rs368151243 Chr.1: 153310650-153310650 on Build GRCh38
rs773554069 Chr.1: 153310692-153310692 on Build GRCh38
rs374274244 Chr.1: 153310717-153310717 on Build GRCh38
rs765652972 Chr.1: 153310572-153310572 on Build GRCh38
rs372264715 Chr.1: 153310689-153310689 on Build GRCh38
rs79228057 Chr.1: 153310658-153310658 on Build GRCh38
rs752392126 Chr.1: 153310559-153310559 on Build GRCh38
rs775329352 Chr.1: 153310675-153310675 on Build GRCh38
rs188086445 Chr.1: 153310681-153310681 on Build GRCh38
rs201683514 Chr.1: 153310558-153310558 on Build GRCh38
rs373935468 Chr.1: 153310712-153310712 on Build GRCh38
rs765079377 Chr.1: 153310534-153310534 on Build GRCh38
rs755230803 Chr.1: 153310632-153310632 on Build GRCh38
rs751029974 Chr.1: 153310688-153310688 on Build GRCh38
rs866857208 Chr.1: 153310693-153310693 on Build GRCh38
rs146790202 Chr.1: 153310654-153310654 on Build GRCh38
rs376486442 Chr.1: 153310686-153310686 on Build GRCh38
rs556030768 Chr.1: 153310746-153310746 on Build GRCh38
rs821423 Chr.1: 153310528-153310528 on Build GRCh38
rs148920280 Chr.1: 153310700-153310700 on Build GRCh38
rs771713172 Chr.1: 153310662-153310662 on Build GRCh38
rs751784425 Chr.1: 153310621-153310621 on Build GRCh38
rs375220818 Chr.1: 153310611-153310611 on Build GRCh38
rs778100694 Chr.1: 153310649-153310650 on Build GRCh38
rs369707602 Chr.1: 153310687-153310687 on Build GRCh38
rs376472085 Chr.1: 153310715-153310715 on Build GRCh38
rs764811111 Chr.1: 153310690-153310690 on Build GRCh38
rs201878205 Chr.1: 153310565-153310565 on Build GRCh38
rs41264634 Chr.1: 153310602-153310602 on Build GRCh38
rs1043411312 Chr.1: 153310702-153310702 on Build GRCh38
rs548249878 Chr.1: 153310701-153310701 on Build GRCh38
rs201396205 Chr.1: 153310685-153310685 on Build GRCh38
rs200831497 Chr.1: 153310616-153310616 on Build GRCh38
rs200469609 Chr.1: 153310657-153310657 on Build GRCh38
rs960509222 Chr.1: 153310529-153310529 on Build GRCh38
rs3006473 Chr.1: 153310703-153310703 on Build GRCh38

View Full Product Details