Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79236761-79237008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMY
Entrez Gene ID
133746
Gene Name
junction mediating and regulatory protein, p53 cofactor
Gene Aliases
WHAMM2, WHDC1L3
UniGene
Hs.482605
Gene Chromosome Location
Chr.5: 79236102-79327215 on Build GRCh38
Transcripts
NM_152405.4 XM_005248430.2 XM_011543155.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4988008 c.253G>A p.A85T Chr.5: 79236903-79236903 on Build GRCh38
3765213 c.307G>C p.G103R Chr.5: 79236957-79236957 on Build GRCh38
6860073 c.135C>T p.T45T Chr.5: 79236785-79236785 on Build GRCh38
5466562 c.122A>C p.K41T Chr.5: 79236772-79236772 on Build GRCh38
6020241 c.349C>G p.R117G Chr.5: 79236999-79236999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769196049 Chr.5: 79236817-79236817 on Build GRCh38
rs893183562 Chr.5: 79236973-79236973 on Build GRCh38
rs760988203 Chr.5: 79236905-79236905 on Build GRCh38
rs772331132 Chr.5: 79236844-79236844 on Build GRCh38
rs891841077 Chr.5: 79236892-79236892 on Build GRCh38
rs901658140 Chr.5: 79236881-79236881 on Build GRCh38
rs1010206229 Chr.5: 79236893-79236893 on Build GRCh38
rs1044347644 Chr.5: 79236982-79236982 on Build GRCh38
rs548000037 Chr.5: 79236830-79236830 on Build GRCh38
rs3733893 Chr.5: 79236835-79236835 on Build GRCh38
rs919051852 Chr.5: 79236801-79236801 on Build GRCh38
rs1040313635 Chr.5: 79236848-79236848 on Build GRCh38
rs984420677 Chr.5: 79236806-79236806 on Build GRCh38
rs960144929 Chr.5: 79236925-79236925 on Build GRCh38
rs1011626029 Chr.5: 79236977-79236977 on Build GRCh38
rs528007143 Chr.5: 79236784-79236784 on Build GRCh38
rs942234251 Chr.5: 79236832-79236832 on Build GRCh38
rs1052279148 Chr.5: 79236861-79236861 on Build GRCh38
rs904473060 Chr.5: 79236900-79236903 on Build GRCh38
rs867809452 Chr.5: 79236810-79236810 on Build GRCh38
rs1053612004 Chr.5: 79236964-79236964 on Build GRCh38
rs983756277 Chr.5: 79236796-79236796 on Build GRCh38
rs930382426 Chr.5: 79236802-79236802 on Build GRCh38
rs773238664 Chr.5: 79236897-79236897 on Build GRCh38
rs539491871 Chr.5: 79236840-79236840 on Build GRCh38
rs1043015223 Chr.5: 79236899-79236899 on Build GRCh38
rs1014052045 Chr.5: 79236957-79236957 on Build GRCh38
rs1034223045 Chr.5: 79236921-79236921 on Build GRCh38

View Full Product Details