Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6556102-6556347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLDC
Entrez Gene ID
2731
Gene Name
glycine decarboxylase
Gene Aliases
GCE, GCSP, HYGN1
UniGene
Hs.584238
Gene Chromosome Location
Chr.9: 6532464-6645692 on Build GRCh38
Transcripts
NM_000170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5479870 c.2148C>G p.L716L Chr.9: 6556207-6556207 on Build GRCh38
2777840 c.2177T>G p.L726R Chr.9: 6556178-6556178 on Build GRCh38
2777844 c.2066A>T p.K689M Chr.9: 6556289-6556289 on Build GRCh38
223315 c.2077G>A p.A693T Chr.9: 6556278-6556278 on Build GRCh38
6242213 c.2078C>A p.A693E Chr.9: 6556277-6556277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372470287 Chr.9: 6556318-6556318 on Build GRCh38
rs912659426 Chr.9: 6556218-6556218 on Build GRCh38
rs763579283 Chr.9: 6556306-6556306 on Build GRCh38
rs769158507 Chr.9: 6556210-6556210 on Build GRCh38
rs966649046 Chr.9: 6556132-6556132 on Build GRCh38
rs140352381 Chr.9: 6556282-6556282 on Build GRCh38
rs774067413 Chr.9: 6556217-6556217 on Build GRCh38
rs1010614777 Chr.9: 6556295-6556295 on Build GRCh38
rs778267718 Chr.9: 6556197-6556197 on Build GRCh38
rs147275962 Chr.9: 6556242-6556242 on Build GRCh38
rs575709546 Chr.9: 6556187-6556187 on Build GRCh38
rs368418946 Chr.9: 6556147-6556147 on Build GRCh38
rs149694787 Chr.9: 6556258-6556258 on Build GRCh38
rs758841885 Chr.9: 6556195-6556195 on Build GRCh38
rs778298626 Chr.9: 6556247-6556247 on Build GRCh38
rs1017391774 Chr.9: 6556273-6556273 on Build GRCh38
rs386833538 Chr.9: 6556275-6556275 on Build GRCh38
rs201599889 Chr.9: 6556130-6556130 on Build GRCh38
rs386833539 Chr.9: 6556257-6556257 on Build GRCh38
rs145547090 Chr.9: 6556270-6556270 on Build GRCh38
rs371447782 Chr.9: 6556269-6556269 on Build GRCh38
rs762786233 Chr.9: 6556215-6556215 on Build GRCh38
rs752086205 Chr.9: 6556313-6556313 on Build GRCh38
rs1014629026 Chr.9: 6556136-6556136 on Build GRCh38
rs187370615 Chr.9: 6556134-6556134 on Build GRCh38
rs140877566 Chr.9: 6556307-6556307 on Build GRCh38
rs779434645 Chr.9: 6556188-6556188 on Build GRCh38
rs376968366 Chr.9: 6556314-6556314 on Build GRCh38
rs763854673 Chr.9: 6556163-6556163 on Build GRCh38
rs753855821 Chr.9: 6556183-6556183 on Build GRCh38
rs367988640 Chr.9: 6556302-6556302 on Build GRCh38
rs746722279 Chr.9: 6556205-6556205 on Build GRCh38
rs753895623 Chr.9: 6556228-6556228 on Build GRCh38
rs386833541 Chr.9: 6556200-6556202 on Build GRCh38
rs753301368 Chr.9: 6556241-6556241 on Build GRCh38
rs147951756 Chr.9: 6556180-6556180 on Build GRCh38
rs386833543 Chr.9: 6556169-6556169 on Build GRCh38
rs386833544 Chr.9: 6556159-6556159 on Build GRCh38
rs769404917 Chr.9: 6556260-6556260 on Build GRCh38
rs867370329 Chr.9: 6556172-6556172 on Build GRCh38
rs751926697 Chr.9: 6556249-6556249 on Build GRCh38
rs761555221 Chr.9: 6556277-6556277 on Build GRCh38
rs386833540 Chr.9: 6556250-6556250 on Build GRCh38
rs759659824 Chr.9: 6556292-6556292 on Build GRCh38
rs993732861 Chr.9: 6556231-6556231 on Build GRCh38
rs765177412 Chr.9: 6556144-6556144 on Build GRCh38
rs765205689 Chr.9: 6556240-6556240 on Build GRCh38
rs545419197 Chr.9: 6556164-6556164 on Build GRCh38
rs763944767 Chr.9: 6556216-6556216 on Build GRCh38
rs577906178 Chr.9: 6556145-6556145 on Build GRCh38
rs376946439 Chr.9: 6556293-6556293 on Build GRCh38
rs780029956 Chr.9: 6556274-6556274 on Build GRCh38
rs781398668 Chr.9: 6556316-6556316 on Build GRCh38
rs150943866 Chr.9: 6556229-6556229 on Build GRCh38
rs1055782169 Chr.9: 6556204-6556204 on Build GRCh38
rs367750112 Chr.9: 6556207-6556207 on Build GRCh38
rs748569405 Chr.9: 6556192-6556192 on Build GRCh38
rs117460214 Chr.9: 6556206-6556206 on Build GRCh38
rs866506579 Chr.9: 6556173-6556174 on Build GRCh38
rs757004829 Chr.9: 6556253-6556253 on Build GRCh38
rs770620676 Chr.9: 6556133-6556133 on Build GRCh38
rs145018304 Chr.9: 6556208-6556208 on Build GRCh38
rs775547766 Chr.9: 6556213-6556213 on Build GRCh38
rs386833542 Chr.9: 6556173-6556173 on Build GRCh38
rs200451792 Chr.9: 6556289-6556289 on Build GRCh38
rs779427031 Chr.9: 6556261-6556261 on Build GRCh38
rs192045547 Chr.9: 6556174-6556174 on Build GRCh38
rs978127778 Chr.9: 6556230-6556230 on Build GRCh38

View Full Product Details