Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3576116-3576383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMG20B
Entrez Gene ID
10362
Gene Name
high mobility group 20B
Gene Aliases
BRAF25, BRAF35, HMGX2, HMGXB2, PP7706, SMARCE1r, SOXL, pp8857
UniGene
Hs.406534
Gene Chromosome Location
Chr.19: 3572938-3579083 on Build GRCh38
Transcripts
NM_006339.2 XM_017026144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6273301 c.473-5G>T p.? Chr.19: 3576256-3576256 on Build GRCh38
1740909 c.519+9_519+11delCCT p.? Chr.19: 3576316-3576318 on Build GRCh38
7263376 c.473-5G>A p.? Chr.19: 3576256-3576256 on Build GRCh38
459987 c.473-1G>C p.? Chr.19: 3576260-3576260 on Build GRCh38
5722786 c.510T>G p.N170K Chr.19: 3576298-3576298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930249416 Chr.19: 3576181-3576181 on Build GRCh38
rs1041914501 Chr.19: 3576263-3576263 on Build GRCh38
rs3217053 Chr.19: 3576321-3576323 on Build GRCh38
rs778831830 Chr.19: 3576340-3576340 on Build GRCh38
rs761157973 Chr.19: 3576234-3576235 on Build GRCh38
rs746521917 Chr.19: 3576257-3576257 on Build GRCh38
rs752632746 Chr.19: 3576247-3576247 on Build GRCh38
rs777434905 Chr.19: 3576249-3576249 on Build GRCh38
rs767001490 Chr.19: 3576293-3576293 on Build GRCh38
rs571180513 Chr.19: 3576238-3576238 on Build GRCh38
rs758692175 Chr.19: 3576349-3576349 on Build GRCh38
rs201729575 Chr.19: 3576321-3576321 on Build GRCh38
rs892309735 Chr.19: 3576336-3576336 on Build GRCh38
rs754453310 Chr.19: 3576342-3576343 on Build GRCh38
rs764039257 Chr.19: 3576334-3576334 on Build GRCh38
rs949875127 Chr.19: 3576184-3576184 on Build GRCh38
rs962067934 Chr.19: 3576177-3576177 on Build GRCh38
rs753713892 Chr.19: 3576338-3576338 on Build GRCh38
rs1039419586 Chr.19: 3576215-3576215 on Build GRCh38
rs201615672 Chr.19: 3576313-3576313 on Build GRCh38
rs756074413 Chr.19: 3576248-3576248 on Build GRCh38
rs754445812 Chr.19: 3576223-3576223 on Build GRCh38
rs750709800 Chr.19: 3576218-3576226 on Build GRCh38
rs757572351 Chr.19: 3576256-3576256 on Build GRCh38
rs757155458 Chr.19: 3576339-3576339 on Build GRCh38
rs772329634 Chr.19: 3576267-3576267 on Build GRCh38
rs372498541 Chr.19: 3576268-3576268 on Build GRCh38
rs542196630 Chr.19: 3576201-3576201 on Build GRCh38
rs377530592 Chr.19: 3576273-3576273 on Build GRCh38
rs369096744 Chr.19: 3576213-3576213 on Build GRCh38
rs35766767 Chr.19: 3576317-3576319 on Build GRCh38
rs371631206 Chr.19: 3576149-3576149 on Build GRCh38
rs200252527 Chr.19: 3576348-3576348 on Build GRCh38
rs927182456 Chr.19: 3576209-3576209 on Build GRCh38
rs776874277 Chr.19: 3576280-3576280 on Build GRCh38
rs1047325378 Chr.19: 3576187-3576187 on Build GRCh38
rs945726712 Chr.19: 3576255-3576255 on Build GRCh38
rs918525036 Chr.19: 3576178-3576178 on Build GRCh38
rs1054154668 Chr.19: 3576231-3576231 on Build GRCh38
rs375024663 Chr.19: 3576188-3576188 on Build GRCh38
rs545961533 Chr.19: 3576143-3576143 on Build GRCh38
rs917490427 Chr.19: 3576161-3576161 on Build GRCh38
rs202025208 Chr.19: 3576292-3576292 on Build GRCh38
rs1029573798 Chr.19: 3576325-3576325 on Build GRCh38
rs751093335 Chr.19: 3576221-3576221 on Build GRCh38
rs766949834 Chr.19: 3576318-3576320 on Build GRCh38
rs531866633 Chr.19: 3576200-3576200 on Build GRCh38
rs769141226 Chr.19: 3576275-3576275 on Build GRCh38
rs756013998 Chr.19: 3576330-3576330 on Build GRCh38
rs746054754 Chr.19: 3576259-3576259 on Build GRCh38
rs993446605 Chr.19: 3576142-3576142 on Build GRCh38

View Full Product Details