Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 96334313-96334572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2F2
Entrez Gene ID
7026
Gene Name
nuclear receptor subfamily 2 group F member 2
Gene Aliases
ARP1, CHTD4, COUPTFB, COUPTFII, NF-E3, NR2F1, SVP40, TFCOUP2
UniGene
Hs.347991
Gene Chromosome Location
Chr.15: 96325928-96340263 on Build GRCh38
Transcripts
NM_001145155.1 NM_001145156.1 NM_001145157.1 NM_021005.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7207712 c.859G>A p.A287T Chr.15: 96334492-96334492 on Build GRCh38
4255124 c.891G>T p.E297D Chr.15: 96334524-96334524 on Build GRCh38
4255111 c.303C>T p.P101P Chr.15: 96334335-96334335 on Build GRCh38
4505664 c.699C>T p.F233F Chr.15: 96334332-96334332 on Build GRCh38
4255113 c.333G>C p.L111L Chr.15: 96334365-96334365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899050920 Chr.15: 96334474-96334474 on Build GRCh38
rs759562272 Chr.15: 96334488-96334488 on Build GRCh38
rs774068976 Chr.15: 96334460-96334460 on Build GRCh38
rs146156153 Chr.15: 96334513-96334513 on Build GRCh38
rs1006491294 Chr.15: 96334476-96334476 on Build GRCh38
rs764508071 Chr.15: 96334543-96334543 on Build GRCh38
rs878855066 Chr.15: 96334342-96334342 on Build GRCh38
rs745316011 Chr.15: 96334434-96334434 on Build GRCh38
rs550114728 Chr.15: 96334545-96334545 on Build GRCh38
rs376446626 Chr.15: 96334552-96334552 on Build GRCh38
rs929257505 Chr.15: 96334485-96334485 on Build GRCh38
rs933038285 Chr.15: 96334374-96334374 on Build GRCh38
rs755719702 Chr.15: 96334437-96334437 on Build GRCh38
rs781768648 Chr.15: 96334431-96334431 on Build GRCh38
rs775659509 Chr.15: 96334512-96334512 on Build GRCh38
rs764406477 Chr.15: 96334404-96334404 on Build GRCh38
rs775539296 Chr.15: 96334377-96334377 on Build GRCh38
rs773164116 Chr.15: 96334479-96334479 on Build GRCh38
rs929629529 Chr.15: 96334356-96334356 on Build GRCh38
rs886041730 Chr.15: 96334489-96334490 on Build GRCh38
rs530322110 Chr.15: 96334452-96334452 on Build GRCh38
rs561184433 Chr.15: 96334440-96334440 on Build GRCh38
rs749297542 Chr.15: 96334376-96334376 on Build GRCh38
rs150869148 Chr.15: 96334371-96334371 on Build GRCh38
rs747987863 Chr.15: 96334463-96334463 on Build GRCh38
rs76163938 Chr.15: 96334449-96334449 on Build GRCh38
rs758770000 Chr.15: 96334387-96334387 on Build GRCh38
rs765401593 Chr.15: 96334335-96334335 on Build GRCh38
rs199886454 Chr.15: 96334467-96334467 on Build GRCh38
rs757765199 Chr.15: 96334428-96334428 on Build GRCh38
rs751816358 Chr.15: 96334551-96334551 on Build GRCh38
rs1037849009 Chr.15: 96334489-96334489 on Build GRCh38
rs143013583 Chr.15: 96334509-96334509 on Build GRCh38
rs752103768 Chr.15: 96334425-96334425 on Build GRCh38
rs886657413 Chr.15: 96334468-96334468 on Build GRCh38

View Full Product Details