Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17152429-17152702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYO9B
Entrez Gene ID
4650
Gene Name
myosin IXB
Gene Aliases
CELIAC4, MYR5
UniGene
Hs.123198
Gene Chromosome Location
Chr.19: 17075781-17213295 on Build GRCh38
Transcripts
NM_001130065.1 NM_004145.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4836942 c.961G>C p.E321Q Chr.19: 17152669-17152669 on Build GRCh38
6237848 c.968C>A p.S323Y Chr.19: 17152676-17152676 on Build GRCh38
417711 c.955C>G p.L319V Chr.19: 17152663-17152663 on Build GRCh38
6379763 c.938C>A p.A313D Chr.19: 17152646-17152646 on Build GRCh38
6341746 c.970C>A p.R324S Chr.19: 17152678-17152678 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs965862218 Chr.19: 17152454-17152454 on Build GRCh38
rs773618591 Chr.19: 17152619-17152619 on Build GRCh38
rs991146581 Chr.19: 17152464-17152464 on Build GRCh38
rs979881377 Chr.19: 17152481-17152481 on Build GRCh38
rs927068072 Chr.19: 17152564-17152564 on Build GRCh38
rs571786593 Chr.19: 17152650-17152650 on Build GRCh38
rs948226975 Chr.19: 17152523-17152523 on Build GRCh38
rs1018783667 Chr.19: 17152453-17152453 on Build GRCh38
rs555742114 Chr.19: 17152537-17152537 on Build GRCh38
rs762053205 Chr.19: 17152594-17152594 on Build GRCh38
rs375021161 Chr.19: 17152587-17152587 on Build GRCh38
rs886113804 Chr.19: 17152584-17152584 on Build GRCh38
rs199684217 Chr.19: 17152563-17152564 on Build GRCh38
rs958367713 Chr.19: 17152463-17152463 on Build GRCh38
rs758512069 Chr.19: 17152452-17152453 on Build GRCh38
rs571240460 Chr.19: 17152533-17152533 on Build GRCh38
rs756813644 Chr.19: 17152677-17152677 on Build GRCh38
rs368337581 Chr.19: 17152651-17152651 on Build GRCh38
rs558277353 Chr.19: 17152631-17152631 on Build GRCh38
rs538238945 Chr.19: 17152565-17152565 on Build GRCh38
rs977312425 Chr.19: 17152578-17152578 on Build GRCh38
rs767840946 Chr.19: 17152616-17152616 on Build GRCh38
rs752530375 Chr.19: 17152648-17152648 on Build GRCh38
rs375200567 Chr.19: 17152678-17152678 on Build GRCh38
rs1033892786 Chr.19: 17152452-17152452 on Build GRCh38
rs917044506 Chr.19: 17152493-17152493 on Build GRCh38
rs77145711 Chr.19: 17152563-17152563 on Build GRCh38
rs956890121 Chr.19: 17152499-17152499 on Build GRCh38
rs532091264 Chr.19: 17152549-17152549 on Build GRCh38
rs567567082 Chr.19: 17152522-17152522 on Build GRCh38
rs879094527 Chr.19: 17152582-17152582 on Build GRCh38
rs765116095 Chr.19: 17152638-17152638 on Build GRCh38
rs992426914 Chr.19: 17152517-17152517 on Build GRCh38
rs949823413 Chr.19: 17152573-17152573 on Build GRCh38
rs34703734 Chr.19: 17152453-17152453 on Build GRCh38
rs1000968152 Chr.19: 17152667-17152667 on Build GRCh38
rs550974285 Chr.19: 17152477-17152477 on Build GRCh38
rs995941572 Chr.19: 17152458-17152458 on Build GRCh38
rs924884291 Chr.19: 17152555-17152555 on Build GRCh38
rs199639787 Chr.19: 17152653-17152653 on Build GRCh38
rs745558830 Chr.19: 17152679-17152679 on Build GRCh38
rs775293241 Chr.19: 17152516-17152516 on Build GRCh38

View Full Product Details