Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73570185-73570458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPTN
Entrez Gene ID
27020
Gene Name
neuroplastin
Gene Aliases
GP55, GP65, SDFR1, SDR1, np55, np65
UniGene
Hs.744867
Gene Chromosome Location
Chr.15: 73560003-73633412 on Build GRCh38
Transcripts
NM_001161363.1 NM_001161364.1 NM_012428.3 NM_017455.3 XM_011521452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5849731 c.1061T>G p.V354G Chr.15: 73570203-73570203 on Build GRCh38
5828276 c.1025C>A p.P342H Chr.15: 73570239-73570239 on Build GRCh38
6728928 c.860G>A p.G287D Chr.15: 73570404-73570404 on Build GRCh38
3887443 c.1025C>T p.P342L Chr.15: 73570239-73570239 on Build GRCh38
5712691 c.982G>T p.V328F Chr.15: 73570282-73570282 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577403495 Chr.15: 73570340-73570340 on Build GRCh38
rs148367676 Chr.15: 73570225-73570225 on Build GRCh38
rs571333983 Chr.15: 73570302-73570302 on Build GRCh38
rs1039383369 Chr.15: 73570311-73570311 on Build GRCh38
rs374225987 Chr.15: 73570341-73570341 on Build GRCh38
rs549983480 Chr.15: 73570292-73570292 on Build GRCh38
rs565798901 Chr.15: 73570409-73570409 on Build GRCh38
rs75660208 Chr.15: 73570426-73570426 on Build GRCh38
rs932962854 Chr.15: 73570344-73570344 on Build GRCh38
rs368630333 Chr.15: 73570370-73570370 on Build GRCh38
rs768364843 Chr.15: 73570235-73570235 on Build GRCh38
rs906089317 Chr.15: 73570247-73570247 on Build GRCh38
rs768150791 Chr.15: 73570396-73570396 on Build GRCh38
rs371300963 Chr.15: 73570333-73570333 on Build GRCh38
rs767422988 Chr.15: 73570291-73570291 on Build GRCh38
rs753969241 Chr.15: 73570304-73570304 on Build GRCh38
rs774063385 Chr.15: 73570263-73570263 on Build GRCh38
rs778390383 Chr.15: 73570360-73570360 on Build GRCh38
rs779373081 Chr.15: 73570379-73570379 on Build GRCh38
rs372732045 Chr.15: 73570390-73570390 on Build GRCh38
rs766431527 Chr.15: 73570303-73570303 on Build GRCh38
rs35100473 Chr.15: 73570265-73570265 on Build GRCh38
rs774040967 Chr.15: 73570397-73570397 on Build GRCh38
rs111525661 Chr.15: 73570425-73570425 on Build GRCh38
rs188693216 Chr.15: 73570264-73570264 on Build GRCh38
rs1006218315 Chr.15: 73570214-73570214 on Build GRCh38

View Full Product Details