Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50154393-50154640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IZUMO2
Entrez Gene ID
126123
Gene Name
IZUMO family member 2
Gene Aliases
C19orf41, PLAL6978, PRO21961, SCRL
UniGene
Hs.414175
Gene Chromosome Location
Chr.19: 50152548-50163281 on Build GRCh38
Transcripts
NM_001321449.1 NM_152358.2 XM_011526436.2 XM_011526437.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3537095 c.612C>T p.V204V Chr.19: 50154611-50154611 on Build GRCh38
1743589 c.601G>A p.V201I Chr.19: 50154622-50154622 on Build GRCh38
999589 c.607G>A p.V203M Chr.19: 50154616-50154616 on Build GRCh38
6346654 c.593C>A p.A198D Chr.19: 50154630-50154630 on Build GRCh38
3537094 c.623C>T p.S208L Chr.19: 50154600-50154600 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756441076 Chr.19: 50154585-50154585 on Build GRCh38
rs189762069 Chr.19: 50154607-50154607 on Build GRCh38
rs570421316 Chr.19: 50154574-50154574 on Build GRCh38
rs1000524192 Chr.19: 50154421-50154421 on Build GRCh38
rs1052749095 Chr.19: 50154499-50154499 on Build GRCh38
rs537960096 Chr.19: 50154582-50154582 on Build GRCh38
rs398079933 Chr.19: 50154532-50154532 on Build GRCh38
rs372232855 Chr.19: 50154464-50154464 on Build GRCh38
rs398034962 Chr.19: 50154533-50154533 on Build GRCh38
rs764725550 Chr.19: 50154569-50154569 on Build GRCh38
rs66651303 Chr.19: 50154530-50154530 on Build GRCh38
rs892896062 Chr.19: 50154537-50154537 on Build GRCh38
rs964437521 Chr.19: 50154505-50154505 on Build GRCh38
rs574717532 Chr.19: 50154616-50154616 on Build GRCh38
rs760480367 Chr.19: 50154553-50154553 on Build GRCh38
rs373238643 Chr.19: 50154567-50154567 on Build GRCh38
rs761800283 Chr.19: 50154568-50154568 on Build GRCh38
rs558524733 Chr.19: 50154550-50154550 on Build GRCh38
rs960962806 Chr.19: 50154606-50154606 on Build GRCh38
rs202151172 Chr.19: 50154530-50154530 on Build GRCh38
rs533728246 Chr.19: 50154485-50154485 on Build GRCh38
rs745913193 Chr.19: 50154608-50154608 on Build GRCh38
rs376223447 Chr.19: 50154575-50154575 on Build GRCh38
rs570788034 Chr.19: 50154599-50154599 on Build GRCh38
rs974027122 Chr.19: 50154615-50154615 on Build GRCh38
rs998946265 Chr.19: 50154469-50154470 on Build GRCh38
rs975804104 Chr.19: 50154509-50154509 on Build GRCh38
rs780257716 Chr.19: 50154617-50154617 on Build GRCh38
rs754391801 Chr.19: 50154586-50154586 on Build GRCh38
rs1030256419 Chr.19: 50154528-50154528 on Build GRCh38
rs766580853 Chr.19: 50154584-50154584 on Build GRCh38
rs1026277967 Chr.19: 50154614-50154614 on Build GRCh38
rs955532620 Chr.19: 50154538-50154538 on Build GRCh38
rs763081741 Chr.19: 50154541-50154541 on Build GRCh38
rs566466721 Chr.19: 50154437-50154437 on Build GRCh38
rs901538126 Chr.19: 50154460-50154460 on Build GRCh38

View Full Product Details