Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113578580-113578839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HABP2
Entrez Gene ID
3026
Gene Name
hyaluronan binding protein 2
Gene Aliases
FSAP, HABP, HGFAL, NMTC5, PHBP
UniGene
Hs.422542
Gene Chromosome Location
Chr.10: 113550831-113589602 on Build GRCh38
Transcripts
NM_001177660.2 NM_004132.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4011684 c.641C>T p.A214V Chr.10: 113578699-113578699 on Build GRCh38
4011683 c.590G>A p.G197D Chr.10: 113578648-113578648 on Build GRCh38
4554126 c.612G>A p.G204G Chr.10: 113578670-113578670 on Build GRCh38
1703043 c.623G>A p.R208K Chr.10: 113578681-113578681 on Build GRCh38
1703042 c.618G>A p.M206I Chr.10: 113578676-113578676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030445013 Chr.10: 113578721-113578721 on Build GRCh38
rs1007282510 Chr.10: 113578600-113578600 on Build GRCh38
rs771594824 Chr.10: 113578790-113578790 on Build GRCh38
rs11575759 Chr.10: 113578691-113578691 on Build GRCh38
rs188451944 Chr.10: 113578695-113578695 on Build GRCh38
rs746446147 Chr.10: 113578618-113578618 on Build GRCh38
rs756569116 Chr.10: 113578759-113578759 on Build GRCh38
rs775761491 Chr.10: 113578623-113578623 on Build GRCh38
rs906853218 Chr.10: 113578653-113578653 on Build GRCh38
rs751837487 Chr.10: 113578626-113578626 on Build GRCh38
rs751212372 Chr.10: 113578732-113578732 on Build GRCh38
rs769188298 Chr.10: 113578627-113578627 on Build GRCh38
rs1045130468 Chr.10: 113578772-113578772 on Build GRCh38
rs963561602 Chr.10: 113578773-113578773 on Build GRCh38
rs757578844 Chr.10: 113578609-113578609 on Build GRCh38
rs577297383 Chr.10: 113578605-113578605 on Build GRCh38
rs140540118 Chr.10: 113578764-113578764 on Build GRCh38
rs774459136 Chr.10: 113578638-113578638 on Build GRCh38
rs370513627 Chr.10: 113578684-113578684 on Build GRCh38
rs141190297 Chr.10: 113578642-113578642 on Build GRCh38
rs751178357 Chr.10: 113578670-113578670 on Build GRCh38
rs767963374 Chr.10: 113578747-113578747 on Build GRCh38
rs769673422 Chr.10: 113578707-113578707 on Build GRCh38
rs768224706 Chr.10: 113578643-113578644 on Build GRCh38
rs781729169 Chr.10: 113578677-113578677 on Build GRCh38
rs533439977 Chr.10: 113578723-113578723 on Build GRCh38
rs200357151 Chr.10: 113578781-113578781 on Build GRCh38
rs201294830 Chr.10: 113578717-113578717 on Build GRCh38
rs769585478 Chr.10: 113578796-113578796 on Build GRCh38
rs967245190 Chr.10: 113578694-113578694 on Build GRCh38
rs184372424 Chr.10: 113578805-113578805 on Build GRCh38
rs750946584 Chr.10: 113578683-113578683 on Build GRCh38
rs748376634 Chr.10: 113578768-113578768 on Build GRCh38
rs761607979 Chr.10: 113578736-113578736 on Build GRCh38
rs749160931 Chr.10: 113578635-113578637 on Build GRCh38
rs145320448 Chr.10: 113578685-113578685 on Build GRCh38
rs909851850 Chr.10: 113578756-113578756 on Build GRCh38
rs764700677 Chr.10: 113578643-113578643 on Build GRCh38
rs41292628 Chr.10: 113578665-113578665 on Build GRCh38
rs540898574 Chr.10: 113578668-113578668 on Build GRCh38
rs772317550 Chr.10: 113578775-113578775 on Build GRCh38
rs755052978 Chr.10: 113578664-113578664 on Build GRCh38
rs529346597 Chr.10: 113578699-113578699 on Build GRCh38
rs775496528 Chr.10: 113578798-113578798 on Build GRCh38
rs986127763 Chr.10: 113578758-113578758 on Build GRCh38
rs572312483 Chr.10: 113578673-113578673 on Build GRCh38
rs769982239 Chr.10: 113578622-113578622 on Build GRCh38
rs750777674 Chr.10: 113578750-113578750 on Build GRCh38
rs781738576 Chr.10: 113578611-113578611 on Build GRCh38
rs192809610 Chr.10: 113578700-113578700 on Build GRCh38
rs778384779 Chr.10: 113578645-113578646 on Build GRCh38
rs768881945 Chr.10: 113578632-113578632 on Build GRCh38
rs755080087 Chr.10: 113578766-113578766 on Build GRCh38
rs780649236 Chr.10: 113578761-113578761 on Build GRCh38
rs759297516 Chr.10: 113578666-113578666 on Build GRCh38
rs776542230 Chr.10: 113578641-113578641 on Build GRCh38
rs770715419 Chr.10: 113578640-113578640 on Build GRCh38
rs144425723 Chr.10: 113578780-113578780 on Build GRCh38
rs776456898 Chr.10: 113578704-113578704 on Build GRCh38
rs545209433 Chr.10: 113578639-113578639 on Build GRCh38
rs201309699 Chr.10: 113578800-113578800 on Build GRCh38
rs747885404 Chr.10: 113578673-113578676 on Build GRCh38
rs749339659 Chr.10: 113578690-113578690 on Build GRCh38
rs150720516 Chr.10: 113578649-113578649 on Build GRCh38
rs551969304 Chr.10: 113578767-113578767 on Build GRCh38

View Full Product Details