Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40777165-40777406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A17
Entrez Gene ID
10478
Gene Name
solute carrier family 25 member 17
Gene Aliases
PMP34
UniGene
Hs.474938
Gene Chromosome Location
Chr.22: 40769630-40819399 on Build GRCh38
Transcripts
NM_001282726.1 NM_001282727.1 NM_006358.3 NR_104235.1 NR_104236.1 NR_104237.1 NR_104238.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4925229 c.568A>G p.K190E Chr.22: 40777257-40777257 on Build GRCh38
1416447 c.585G>T p.K195N Chr.22: 40777240-40777240 on Build GRCh38
3554853 c.578T>A p.L193H Chr.22: 40777247-40777247 on Build GRCh38
727047 c.474C>T p.R158R Chr.22: 40777351-40777351 on Build GRCh38
3357342 c.540C>A p.A180A Chr.22: 40777285-40777285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147363442 Chr.22: 40777252-40777252 on Build GRCh38
rs1033266015 Chr.22: 40777212-40777212 on Build GRCh38
rs200034270 Chr.22: 40777221-40777221 on Build GRCh38
rs1009274521 Chr.22: 40777334-40777334 on Build GRCh38
rs768147274 Chr.22: 40777236-40777236 on Build GRCh38
rs550917520 Chr.22: 40777196-40777196 on Build GRCh38
rs146405395 Chr.22: 40777299-40777299 on Build GRCh38
rs749337377 Chr.22: 40777312-40777312 on Build GRCh38
rs773140538 Chr.22: 40777311-40777311 on Build GRCh38
rs774570211 Chr.22: 40777337-40777337 on Build GRCh38
rs753200401 Chr.22: 40777241-40777241 on Build GRCh38
rs930460522 Chr.22: 40777245-40777245 on Build GRCh38
rs776207643 Chr.22: 40777348-40777348 on Build GRCh38
rs368228929 Chr.22: 40777339-40777339 on Build GRCh38
rs751210365 Chr.22: 40777357-40777357 on Build GRCh38
rs536724473 Chr.22: 40777350-40777350 on Build GRCh38
rs754832512 Chr.22: 40777364-40777364 on Build GRCh38
rs774597302 Chr.22: 40777234-40777234 on Build GRCh38
rs139416009 Chr.22: 40777353-40777353 on Build GRCh38
rs372825776 Chr.22: 40777352-40777352 on Build GRCh38
rs779708857 Chr.22: 40777287-40777287 on Build GRCh38
rs750132160 Chr.22: 40777253-40777253 on Build GRCh38
rs962467103 Chr.22: 40777351-40777351 on Build GRCh38
rs761996013 Chr.22: 40777251-40777252 on Build GRCh38
rs771085682 Chr.22: 40777332-40777332 on Build GRCh38
rs374275689 Chr.22: 40777229-40777229 on Build GRCh38
rs766812691 Chr.22: 40777233-40777233 on Build GRCh38
rs756888117 Chr.22: 40777247-40777247 on Build GRCh38
rs776484415 Chr.22: 40777193-40777193 on Build GRCh38
rs748129791 Chr.22: 40777370-40777370 on Build GRCh38
rs756065740 Chr.22: 40777375-40777375 on Build GRCh38
rs368390943 Chr.22: 40777220-40777220 on Build GRCh38
rs746785049 Chr.22: 40777292-40777292 on Build GRCh38
rs748072213 Chr.22: 40777309-40777309 on Build GRCh38
rs979787675 Chr.22: 40777276-40777276 on Build GRCh38
rs142349727 Chr.22: 40777254-40777254 on Build GRCh38
rs374468567 Chr.22: 40777356-40777356 on Build GRCh38
rs759909984 Chr.22: 40777235-40777235 on Build GRCh38
rs780932632 Chr.22: 40777365-40777365 on Build GRCh38

View Full Product Details