Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16847067-16847308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIN3B
Entrez Gene ID
23309
Gene Name
SIN3 transcription regulator family member B
Gene Aliases
-
UniGene
Hs.13999
Gene Chromosome Location
Chr.19: 16829387-16880355 on Build GRCh38
Transcripts
NM_001297595.1 NM_015260.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3530185 c.722C>T p.S241F Chr.19: 16847109-16847109 on Build GRCh38
992584 c.700T>G p.F234V Chr.19: 16847087-16847087 on Build GRCh38
6778625 c.688G>A p.E230K Chr.19: 16847075-16847075 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs978272608 Chr.19: 16847222-16847222 on Build GRCh38
rs532182362 Chr.19: 16847219-16847219 on Build GRCh38
rs200016524 Chr.19: 16847145-16847145 on Build GRCh38
rs200936139 Chr.19: 16847092-16847092 on Build GRCh38
rs371933736 Chr.19: 16847155-16847155 on Build GRCh38
rs78941635 Chr.19: 16847263-16847263 on Build GRCh38
rs143674853 Chr.19: 16847154-16847154 on Build GRCh38
rs751080871 Chr.19: 16847103-16847103 on Build GRCh38
rs758067163 Chr.19: 16847096-16847096 on Build GRCh38
rs371807771 Chr.19: 16847129-16847129 on Build GRCh38
rs551489883 Chr.19: 16847203-16847203 on Build GRCh38
rs1030676241 Chr.19: 16847173-16847173 on Build GRCh38
rs929820177 Chr.19: 16847167-16847167 on Build GRCh38
rs749567189 Chr.19: 16847143-16847143 on Build GRCh38
rs368387064 Chr.19: 16847202-16847202 on Build GRCh38
rs763419784 Chr.19: 16847143-16847145 on Build GRCh38
rs1031157556 Chr.19: 16847242-16847242 on Build GRCh38
rs752404902 Chr.19: 16847095-16847095 on Build GRCh38
rs773623461 Chr.19: 16847158-16847158 on Build GRCh38
rs989557149 Chr.19: 16847272-16847272 on Build GRCh38
rs1005242437 Chr.19: 16847191-16847191 on Build GRCh38
rs544341055 Chr.19: 16847193-16847193 on Build GRCh38
rs540045496 Chr.19: 16847101-16847101 on Build GRCh38
rs139915742 Chr.19: 16847204-16847204 on Build GRCh38
rs781013600 Chr.19: 16847105-16847105 on Build GRCh38
rs970939090 Chr.19: 16847209-16847209 on Build GRCh38
rs553355353 Chr.19: 16847123-16847123 on Build GRCh38
rs368818471 Chr.19: 16847122-16847122 on Build GRCh38
rs146827971 Chr.19: 16847160-16847160 on Build GRCh38
rs575750214 Chr.19: 16847187-16847187 on Build GRCh38
rs894128133 Chr.19: 16847200-16847200 on Build GRCh38
rs878987918 Chr.19: 16847190-16847190 on Build GRCh38
rs559147764 Chr.19: 16847252-16847252 on Build GRCh38
rs200744398 Chr.19: 16847142-16847142 on Build GRCh38
rs911265795 Chr.19: 16847279-16847279 on Build GRCh38
rs545997846 Chr.19: 16847221-16847221 on Build GRCh38

View Full Product Details