Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106842428-106842679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIC8B
Entrez Gene ID
55188
Gene Name
RIC8 guanine nucleotide exchange factor B
Gene Aliases
RIC8, hSyn
UniGene
Hs.131306
Gene Chromosome Location
Chr.12: 106774285-106889316 on Build GRCh38
Transcripts
NM_018157.2 XM_005268998.3 XM_011538526.2 XM_011538527.2 XM_011538528.2 XM_011538529.2 XM_017019548.1 XM_017019549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6268493 c.893G>T p.C298F Chr.12: 106842645-106842645 on Build GRCh38
6268494 c.773G>T p.C258F Chr.12: 106842645-106842645 on Build GRCh38
6766379 c.857G>A p.S286N Chr.12: 106842609-106842609 on Build GRCh38
5417561 c.849C>T p.N283N Chr.12: 106842601-106842601 on Build GRCh38
6766380 c.737G>A p.S246N Chr.12: 106842609-106842609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762493086 Chr.12: 106842549-106842549 on Build GRCh38
rs915481567 Chr.12: 106842472-106842472 on Build GRCh38
rs946703524 Chr.12: 106842522-106842522 on Build GRCh38
rs17038800 Chr.12: 106842619-106842619 on Build GRCh38
rs774038205 Chr.12: 106842554-106842554 on Build GRCh38
rs568370832 Chr.12: 106842478-106842478 on Build GRCh38
rs112286691 Chr.12: 106842625-106842625 on Build GRCh38
rs1050239086 Chr.12: 106842505-106842505 on Build GRCh38
rs899072265 Chr.12: 106842656-106842656 on Build GRCh38
rs772627912 Chr.12: 106842543-106842543 on Build GRCh38
rs752478686 Chr.12: 106842568-106842568 on Build GRCh38
rs1042772691 Chr.12: 106842529-106842529 on Build GRCh38
rs534237794 Chr.12: 106842530-106842530 on Build GRCh38
rs931929296 Chr.12: 106842503-106842503 on Build GRCh38
rs923203724 Chr.12: 106842483-106842483 on Build GRCh38
rs570933583 Chr.12: 106842575-106842575 on Build GRCh38
rs758485519 Chr.12: 106842622-106842622 on Build GRCh38
rs1057240635 Chr.12: 106842649-106842649 on Build GRCh38
rs751778418 Chr.12: 106842632-106842632 on Build GRCh38
rs200858563 Chr.12: 106842565-106842565 on Build GRCh38
rs941762679 Chr.12: 106842545-106842545 on Build GRCh38
rs909054761 Chr.12: 106842506-106842506 on Build GRCh38
rs895630233 Chr.12: 106842614-106842614 on Build GRCh38
rs770536112 Chr.12: 106842550-106842550 on Build GRCh38

View Full Product Details