Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137555444-137555677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPH7
Entrez Gene ID
92715
Gene Name
diphthamide biosynthesis 7
Gene Aliases
C9orf112, RRT2, WDR85
UniGene
Hs.292570
Gene Chromosome Location
Chr.9: 137554899-137578935 on Build GRCh38
Transcripts
NM_138778.3 XM_005266123.2 XM_005266126.3 XM_005266128.2 XM_005266129.2 XM_005266130.2 XM_011519188.1 XM_011519189.1 XM_011519191.2 XM_011519193.2 XM_011519194.1 XM_011519199.2 XM_011519200.1 XM_017015289.1 XM_017015290.1 XM_017015291.1 XM_017015292.1 XM_017015293.1 XM_017015294.1 XM_017015295.1 XM_017015296.1 XM_017015297.1 XM_017015298.1 XM_017015299.1 XM_017015300.1 XM_017015301.1 XM_017015302.1 XM_017015303.1 XM_017015304.1 XM_017015305.1 XM_017015306.1 XM_017015307.1 XM_017015308.1 XM_017015309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
316556 c.1069C>T p.P357S Chr.9: 137555529-137555529 on Build GRCh38
6851988 c.1153G>A p.D385N Chr.9: 137555445-137555445 on Build GRCh38
4393605 c.1048C>T p.R350W Chr.9: 137555550-137555550 on Build GRCh38
6902503 c.984C>T p.H328H Chr.9: 137555614-137555614 on Build GRCh38
3367659 c.1151A>C p.N384T Chr.9: 137555447-137555447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780698755 Chr.9: 137555539-137555539 on Build GRCh38
rs747419748 Chr.9: 137555618-137555618 on Build GRCh38
rs765339547 Chr.9: 137555515-137555515 on Build GRCh38
rs970904148 Chr.9: 137555528-137555528 on Build GRCh38
rs894344812 Chr.9: 137555558-137555558 on Build GRCh38
rs113942889 Chr.9: 137555602-137555602 on Build GRCh38
rs375207049 Chr.9: 137555611-137555611 on Build GRCh38
rs370507645 Chr.9: 137555485-137555485 on Build GRCh38
rs766505656 Chr.9: 137555511-137555511 on Build GRCh38
rs755014363 Chr.9: 137555549-137555550 on Build GRCh38
rs200029471 Chr.9: 137555591-137555591 on Build GRCh38
rs868377901 Chr.9: 137555629-137555629 on Build GRCh38
rs371186952 Chr.9: 137555529-137555529 on Build GRCh38
rs771901986 Chr.9: 137555640-137555641 on Build GRCh38
rs147900158 Chr.9: 137555507-137555507 on Build GRCh38
rs142602479 Chr.9: 137555562-137555562 on Build GRCh38
rs60900041 Chr.9: 137555520-137555520 on Build GRCh38
rs763115666 Chr.9: 137555642-137555642 on Build GRCh38
rs77855295 Chr.9: 137555632-137555632 on Build GRCh38
rs772885737 Chr.9: 137555575-137555575 on Build GRCh38
rs747882256 Chr.9: 137555566-137555566 on Build GRCh38
rs371780538 Chr.9: 137555604-137555604 on Build GRCh38
rs201005708 Chr.9: 137555544-137555544 on Build GRCh38
rs748820484 Chr.9: 137555550-137555550 on Build GRCh38
rs775918584 Chr.9: 137555636-137555636 on Build GRCh38
rs769355326 Chr.9: 137555494-137555494 on Build GRCh38
rs748767119 Chr.9: 137555610-137555610 on Build GRCh38
rs552023460 Chr.9: 137555633-137555633 on Build GRCh38
rs768728430 Chr.9: 137555652-137555652 on Build GRCh38
rs747420207 Chr.9: 137555476-137555476 on Build GRCh38
rs117223361 Chr.9: 137555630-137555630 on Build GRCh38
rs757710570 Chr.9: 137555538-137555538 on Build GRCh38
rs964037509 Chr.9: 137555567-137555567 on Build GRCh38
rs745369616 Chr.9: 137555492-137555492 on Build GRCh38
rs112061801 Chr.9: 137555584-137555584 on Build GRCh38
rs768560221 Chr.9: 137555561-137555561 on Build GRCh38
rs755544744 Chr.9: 137555563-137555563 on Build GRCh38
rs758977035 Chr.9: 137555472-137555472 on Build GRCh38
rs774996964 Chr.9: 137555498-137555498 on Build GRCh38
rs560027419 Chr.9: 137555605-137555605 on Build GRCh38
rs145337925 Chr.9: 137555542-137555542 on Build GRCh38
rs140026738 Chr.9: 137555513-137555513 on Build GRCh38
rs61000970 Chr.9: 137555568-137555568 on Build GRCh38
rs749158800 Chr.9: 137555499-137555499 on Build GRCh38
rs111387494 Chr.9: 137555540-137555540 on Build GRCh38
rs759690235 Chr.9: 137555514-137555514 on Build GRCh38
rs778235670 Chr.9: 137555590-137555590 on Build GRCh38
rs575002556 Chr.9: 137555484-137555484 on Build GRCh38
rs764742891 Chr.9: 137555536-137555536 on Build GRCh38
rs149407940 Chr.9: 137555549-137555549 on Build GRCh38
rs778299640 Chr.9: 137555616-137555616 on Build GRCh38
rs759762863 Chr.9: 137555583-137555583 on Build GRCh38
rs772822279 Chr.9: 137555656-137555657 on Build GRCh38
rs771842398 Chr.9: 137555569-137555569 on Build GRCh38
rs779011904 Chr.9: 137555550-137555551 on Build GRCh38
rs764689639 Chr.9: 137555586-137555586 on Build GRCh38
rs1055131999 Chr.9: 137555556-137555556 on Build GRCh38
rs753196193 Chr.9: 137555470-137555470 on Build GRCh38
rs149787260 Chr.9: 137555506-137555506 on Build GRCh38
rs748054982 Chr.9: 137555553-137555554 on Build GRCh38
rs768613421 Chr.9: 137555500-137555500 on Build GRCh38
rs767859766 Chr.9: 137555599-137555599 on Build GRCh38

View Full Product Details