Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 7999573-7999810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMX4
Entrez Gene ID
56255
Gene Name
thioredoxin related transmembrane protein 4
Gene Aliases
DJ971N18.2, PDIA14, TXNDC13
UniGene
Hs.169358
Gene Chromosome Location
Chr.20: 7977353-8019829 on Build GRCh38
Transcripts
NM_021156.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5005571 c.429C>T p.V143V Chr.20: 7999770-7999770 on Build GRCh38
3799791 c.421C>G p.Q141E Chr.20: 7999778-7999778 on Build GRCh38
3758795 c.456G>A p.P152P Chr.20: 7999743-7999743 on Build GRCh38
1029299 c.396G>T p.Q132H Chr.20: 7999803-7999803 on Build GRCh38
1713160 c.452C>T p.S151F Chr.20: 7999747-7999747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944437309 Chr.20: 7999636-7999636 on Build GRCh38
rs1012891 Chr.20: 7999743-7999743 on Build GRCh38
rs746078480 Chr.20: 7999622-7999622 on Build GRCh38
rs1012806744 Chr.20: 7999723-7999723 on Build GRCh38
rs552059538 Chr.20: 7999650-7999650 on Build GRCh38
rs756828345 Chr.20: 7999757-7999760 on Build GRCh38
rs760743812 Chr.20: 7999769-7999769 on Build GRCh38
rs765484923 Chr.20: 7999773-7999773 on Build GRCh38
rs1048954479 Chr.20: 7999652-7999652 on Build GRCh38
rs546277869 Chr.20: 7999706-7999706 on Build GRCh38
rs200533634 Chr.20: 7999731-7999731 on Build GRCh38
rs1003395010 Chr.20: 7999651-7999651 on Build GRCh38
rs570063622 Chr.20: 7999687-7999687 on Build GRCh38
rs779950183 Chr.20: 7999742-7999742 on Build GRCh38
rs940268665 Chr.20: 7999603-7999603 on Build GRCh38
rs751135998 Chr.20: 7999712-7999712 on Build GRCh38
rs994653287 Chr.20: 7999649-7999649 on Build GRCh38
rs760083195 Chr.20: 7999772-7999772 on Build GRCh38
rs753135799 Chr.20: 7999778-7999778 on Build GRCh38
rs932305 Chr.20: 7999643-7999643 on Build GRCh38
rs773120297 Chr.20: 7999765-7999765 on Build GRCh38
rs1023566030 Chr.20: 7999624-7999624 on Build GRCh38
rs368212898 Chr.20: 7999770-7999770 on Build GRCh38
rs746476405 Chr.20: 7999702-7999702 on Build GRCh38
rs752266807 Chr.20: 7999708-7999708 on Build GRCh38
rs772374070 Chr.20: 7999744-7999744 on Build GRCh38
rs200435865 Chr.20: 7999746-7999746 on Build GRCh38
rs113117051 Chr.20: 7999710-7999710 on Build GRCh38
rs368443047 Chr.20: 7999725-7999725 on Build GRCh38
rs898752748 Chr.20: 7999610-7999610 on Build GRCh38
rs897656421 Chr.20: 7999644-7999644 on Build GRCh38
rs200048630 Chr.20: 7999711-7999711 on Build GRCh38
rs202200332 Chr.20: 7999732-7999732 on Build GRCh38
rs375102033 Chr.20: 7999737-7999737 on Build GRCh38
rs200767534 Chr.20: 7999693-7999693 on Build GRCh38
rs759887035 Chr.20: 7999700-7999700 on Build GRCh38

View Full Product Details