Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 38895824-38896053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBDL2
Entrez Gene ID
54933
Gene Name
rhomboid like 2
Gene Aliases
RRP2
UniGene
Hs.524626
Gene Chromosome Location
Chr.1: 38885806-38941830 on Build GRCh38
Transcripts
NM_001304746.1 NM_017821.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3489304 c.824G>A p.G275E Chr.1: 38895994-38895994 on Build GRCh38
3489303 c.584G>A p.G195E Chr.1: 38895994-38895994 on Build GRCh38
6271567 c.557C>A p.A186D Chr.1: 38896021-38896021 on Build GRCh38
4819450 c.780C>G p.L260L Chr.1: 38896038-38896038 on Build GRCh38
680801 c.562G>A p.G188R Chr.1: 38896016-38896016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764014392 Chr.1: 38895984-38895984 on Build GRCh38
rs901690033 Chr.1: 38895947-38895947 on Build GRCh38
rs574407448 Chr.1: 38895900-38895900 on Build GRCh38
rs536192045 Chr.1: 38895891-38895891 on Build GRCh38
rs924242930 Chr.1: 38895881-38895881 on Build GRCh38
rs374046214 Chr.1: 38895942-38895942 on Build GRCh38
rs1040183987 Chr.1: 38895946-38895946 on Build GRCh38
rs770209539 Chr.1: 38895934-38895934 on Build GRCh38
rs977653610 Chr.1: 38895868-38895868 on Build GRCh38
rs111906633 Chr.1: 38896009-38896009 on Build GRCh38
rs766960969 Chr.1: 38895981-38895981 on Build GRCh38
rs971594787 Chr.1: 38895867-38895867 on Build GRCh38
rs12085567 Chr.1: 38895897-38895897 on Build GRCh38
rs753564928 Chr.1: 38896001-38896001 on Build GRCh38
rs758745269 Chr.1: 38896004-38896004 on Build GRCh38
rs112437076 Chr.1: 38896011-38896011 on Build GRCh38
rs763442431 Chr.1: 38896012-38896012 on Build GRCh38
rs964336206 Chr.1: 38895977-38895977 on Build GRCh38
rs371953902 Chr.1: 38895975-38895975 on Build GRCh38
rs567159311 Chr.1: 38895864-38895864 on Build GRCh38
rs751796323 Chr.1: 38896010-38896010 on Build GRCh38
rs773890339 Chr.1: 38895960-38895960 on Build GRCh38
rs929173125 Chr.1: 38895966-38895967 on Build GRCh38
rs9438984 Chr.1: 38895943-38895943 on Build GRCh38
rs576534526 Chr.1: 38895983-38895983 on Build GRCh38
rs543065636 Chr.1: 38895909-38895909 on Build GRCh38

View Full Product Details