Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 64152529-64152768 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX22
Entrez Gene ID
79856
Gene Name
sorting nexin 22
Gene Aliases
-
UniGene
Hs.744250
Gene Chromosome Location
Chr.15: 64151698-64157481 on Build GRCh38
Transcripts
NM_024798.2 NR_073534.1 XM_005254677.3 XM_017022581.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4056196 c.209A>G p.N70S Chr.15: 64152687-64152687 on Build GRCh38
326535 c.187T>G p.F63V Chr.15: 64152665-64152665 on Build GRCh38
5770952 c.262C>T p.Q88* Chr.15: 64152740-64152740 on Build GRCh38
4128330 c.247T>G p.L83V Chr.15: 64152725-64152725 on Build GRCh38
5633862 c.160-1G>C p.? Chr.15: 64152637-64152637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140236564 Chr.15: 64152673-64152673 on Build GRCh38
rs187502209 Chr.15: 64152573-64152573 on Build GRCh38
rs143180219 Chr.15: 64152703-64152703 on Build GRCh38
rs775189306 Chr.15: 64152658-64152658 on Build GRCh38
rs773080540 Chr.15: 64152624-64152624 on Build GRCh38
rs539984707 Chr.15: 64152699-64152699 on Build GRCh38
rs756358925 Chr.15: 64152697-64152697 on Build GRCh38
rs372493745 Chr.15: 64152616-64152616 on Build GRCh38
rs763430883 Chr.15: 64152742-64152742 on Build GRCh38
rs749257298 Chr.15: 64152628-64152628 on Build GRCh38
rs569929506 Chr.15: 64152551-64152551 on Build GRCh38
rs778968542 Chr.15: 64152705-64152705 on Build GRCh38
rs534410250 Chr.15: 64152604-64152604 on Build GRCh38
rs768336919 Chr.15: 64152727-64152727 on Build GRCh38
rs757676051 Chr.15: 64152554-64152554 on Build GRCh38
rs748380156 Chr.15: 64152609-64152609 on Build GRCh38
rs375736180 Chr.15: 64152631-64152631 on Build GRCh38
rs751994573 Chr.15: 64152672-64152672 on Build GRCh38
rs771468187 Chr.15: 64152653-64152653 on Build GRCh38
rs775513840 Chr.15: 64152702-64152702 on Build GRCh38
rs907758646 Chr.15: 64152563-64152563 on Build GRCh38
rs750747498 Chr.15: 64152665-64152665 on Build GRCh38
rs765581722 Chr.15: 64152559-64152559 on Build GRCh38
rs776241432 Chr.15: 64152731-64152731 on Build GRCh38
rs192651413 Chr.15: 64152661-64152661 on Build GRCh38
rs752985651 Chr.15: 64152691-64152691 on Build GRCh38
rs766961799 Chr.15: 64152670-64152670 on Build GRCh38
rs375272799 Chr.15: 64152707-64152707 on Build GRCh38
rs959837096 Chr.15: 64152558-64152558 on Build GRCh38
rs774931673 Chr.15: 64152714-64152714 on Build GRCh38
rs201844254 Chr.15: 64152602-64152602 on Build GRCh38
rs759133480 Chr.15: 64152649-64152649 on Build GRCh38
rs537063498 Chr.15: 64152561-64152561 on Build GRCh38
rs745725932 Chr.15: 64152656-64152656 on Build GRCh38
rs766644882 Chr.15: 64152747-64152747 on Build GRCh38
rs867884834 Chr.15: 64152663-64152663 on Build GRCh38
rs747154728 Chr.15: 64152603-64152603 on Build GRCh38
rs781350202 Chr.15: 64152687-64152687 on Build GRCh38
rs763198054 Chr.15: 64152667-64152667 on Build GRCh38
rs755436304 Chr.15: 64152731-64152732 on Build GRCh38
rs915154151 Chr.15: 64152560-64152581 on Build GRCh38
rs760351045 Chr.15: 64152719-64152719 on Build GRCh38
rs770859303 Chr.15: 64152630-64152630 on Build GRCh38

View Full Product Details