Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 101409439-101409700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF189
Entrez Gene ID
7743
Gene Name
zinc finger protein 189
Gene Aliases
-
UniGene
Hs.50123
Gene Chromosome Location
Chr.9: 101398830-101410660 on Build GRCh38
Transcripts
NM_001278231.1 NM_001278232.1 NM_001278240.1 NM_003452.3 NM_197977.2 NR_103480.1 XM_006717280.3 XM_006717281.3 XM_011518998.2 XM_011518999.2 XM_017015121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7135908 c.1779G>A p.V593V Chr.9: 101409547-101409547 on Build GRCh38
6487125 c.1816G>T p.E606* Chr.9: 101409584-101409584 on Build GRCh38
5570175 c.1784C>T p.T595I Chr.9: 101409552-101409552 on Build GRCh38
5410048 c.1754A>G p.N585S Chr.9: 101409522-101409522 on Build GRCh38
1103327 c.1865C>T p.T622I Chr.9: 101409633-101409633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951309327 Chr.9: 101409492-101409492 on Build GRCh38
rs551296072 Chr.9: 101409501-101409501 on Build GRCh38
rs141381160 Chr.9: 101409593-101409593 on Build GRCh38
rs561264739 Chr.9: 101409666-101409666 on Build GRCh38
rs777410356 Chr.9: 101409575-101409575 on Build GRCh38
rs753558896 Chr.9: 101409672-101409672 on Build GRCh38
rs749290504 Chr.9: 101409577-101409577 on Build GRCh38
rs777020223 Chr.9: 101409594-101409594 on Build GRCh38
rs61755102 Chr.9: 101409488-101409488 on Build GRCh38
rs898943543 Chr.9: 101409533-101409533 on Build GRCh38
rs767169646 Chr.9: 101409565-101409565 on Build GRCh38
rs374957338 Chr.9: 101409470-101409470 on Build GRCh38
rs200682925 Chr.9: 101409633-101409633 on Build GRCh38
rs1033574548 Chr.9: 101409635-101409635 on Build GRCh38
rs768990993 Chr.9: 101409514-101409514 on Build GRCh38
rs776940219 Chr.9: 101409515-101409515 on Build GRCh38
rs775403415 Chr.9: 101409510-101409510 on Build GRCh38
rs765953036 Chr.9: 101409556-101409556 on Build GRCh38
rs777719704 Chr.9: 101409498-101409498 on Build GRCh38
rs761028190 Chr.9: 101409512-101409512 on Build GRCh38
rs757140135 Chr.9: 101409582-101409582 on Build GRCh38
rs371845980 Chr.9: 101409589-101409589 on Build GRCh38
rs749132749 Chr.9: 101409503-101409503 on Build GRCh38
rs200753298 Chr.9: 101409471-101409471 on Build GRCh38
rs139400109 Chr.9: 101409476-101409476 on Build GRCh38
rs757016163 Chr.9: 101409673-101409673 on Build GRCh38
rs781030904 Chr.9: 101409487-101409487 on Build GRCh38
rs765848213 Chr.9: 101409463-101409463 on Build GRCh38
rs563902160 Chr.9: 101409507-101409507 on Build GRCh38
rs1003639996 Chr.9: 101409641-101409641 on Build GRCh38
rs751894244 Chr.9: 101409495-101409495 on Build GRCh38
rs112338244 Chr.9: 101409563-101409563 on Build GRCh38
rs374890233 Chr.9: 101409639-101409639 on Build GRCh38
rs759077787 Chr.9: 101409618-101409618 on Build GRCh38
rs770328593 Chr.9: 101409598-101409598 on Build GRCh38
rs765653896 Chr.9: 101409525-101409525 on Build GRCh38
rs772245213 Chr.9: 101409509-101409509 on Build GRCh38
rs764014416 Chr.9: 101409671-101409671 on Build GRCh38
rs200572031 Chr.9: 101409668-101409668 on Build GRCh38
rs975873200 Chr.9: 101409664-101409664 on Build GRCh38
rs145211802 Chr.9: 101409599-101409599 on Build GRCh38
rs151018933 Chr.9: 101409600-101409600 on Build GRCh38
rs536541759 Chr.9: 101409468-101409468 on Build GRCh38
rs17772514 Chr.9: 101409574-101409574 on Build GRCh38
rs748328852 Chr.9: 101409595-101409595 on Build GRCh38
rs903451820 Chr.9: 101409591-101409591 on Build GRCh38
rs147826896 Chr.9: 101409588-101409588 on Build GRCh38
rs769512658 Chr.9: 101409489-101409489 on Build GRCh38
rs773789220 Chr.9: 101409538-101409538 on Build GRCh38
rs942785328 Chr.9: 101409541-101409541 on Build GRCh38
rs766781706 Chr.9: 101409608-101409608 on Build GRCh38
rs567661929 Chr.9: 101409466-101409466 on Build GRCh38
rs143854569 Chr.9: 101409522-101409522 on Build GRCh38
rs763343823 Chr.9: 101409548-101409548 on Build GRCh38
rs186555374 Chr.9: 101409611-101409611 on Build GRCh38
rs370224206 Chr.9: 101409663-101409663 on Build GRCh38
rs1036597415 Chr.9: 101409532-101409532 on Build GRCh38

View Full Product Details