Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55483580-55483831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF628
Entrez Gene ID
89887
Gene Name
zinc finger protein 628
Gene Aliases
ZEC, Zfp628
UniGene
Hs.525209
Gene Chromosome Location
Chr.19: 55476332-55484487 on Build GRCh38
Transcripts
NM_033113.2 XM_005259371.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1396588 c.2580G>A p.T860T Chr.19: 55483785-55483785 on Build GRCh38
4336674 c.2415delG p.G807fs*9 Chr.19: 55483608-55483608 on Build GRCh38
4287668 c.2407C>T p.Q803* Chr.19: 55483600-55483600 on Build GRCh38
4744505 c.2560G>A p.V854I Chr.19: 55483753-55483753 on Build GRCh38
6579568 c.2425G>A p.E809K Chr.19: 55483618-55483618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774854473 Chr.19: 55483673-55483673 on Build GRCh38
rs73608718 Chr.19: 55483710-55483710 on Build GRCh38
rs536933453 Chr.19: 55483713-55483713 on Build GRCh38
rs767381145 Chr.19: 55483761-55483761 on Build GRCh38
rs780418169 Chr.19: 55483779-55483779 on Build GRCh38
rs776287825 Chr.19: 55483626-55483626 on Build GRCh38
rs769506560 Chr.19: 55483806-55483806 on Build GRCh38
rs769260821 Chr.19: 55483607-55483607 on Build GRCh38
rs756369050 Chr.19: 55483660-55483660 on Build GRCh38
rs764848720 Chr.19: 55483720-55483720 on Build GRCh38
rs778162010 Chr.19: 55483788-55483788 on Build GRCh38
rs759069851 Chr.19: 55483628-55483628 on Build GRCh38
rs776170068 Chr.19: 55483605-55483605 on Build GRCh38
rs779309683 Chr.19: 55483782-55483782 on Build GRCh38
rs747362459 Chr.19: 55483791-55483791 on Build GRCh38
rs150319373 Chr.19: 55483783-55483783 on Build GRCh38
rs781287394 Chr.19: 55483656-55483656 on Build GRCh38
rs765950605 Chr.19: 55483615-55483615 on Build GRCh38
rs764083798 Chr.19: 55483639-55483639 on Build GRCh38
rs755972905 Chr.19: 55483610-55483610 on Build GRCh38
rs894115463 Chr.19: 55483755-55483755 on Build GRCh38
rs746044467 Chr.19: 55483659-55483659 on Build GRCh38
rs995261941 Chr.19: 55483714-55483714 on Build GRCh38
rs376779424 Chr.19: 55483651-55483651 on Build GRCh38
rs751380394 Chr.19: 55483640-55483640 on Build GRCh38
rs760496368 Chr.19: 55483732-55483732 on Build GRCh38
rs764688690 Chr.19: 55483630-55483630 on Build GRCh38
rs769943031 Chr.19: 55483602-55483602 on Build GRCh38
rs200801147 Chr.19: 55483685-55483685 on Build GRCh38
rs752171791 Chr.19: 55483633-55483633 on Build GRCh38
rs772427940 Chr.19: 55483784-55483784 on Build GRCh38
rs756459119 Chr.19: 55483777-55483777 on Build GRCh38
rs1001275739 Chr.19: 55483753-55483753 on Build GRCh38
rs749847821 Chr.19: 55483606-55483606 on Build GRCh38
rs942769747 Chr.19: 55483623-55483623 on Build GRCh38
rs373922538 Chr.19: 55483703-55483703 on Build GRCh38
rs1010086901 Chr.19: 55483736-55483736 on Build GRCh38
rs368131720 Chr.19: 55483780-55483780 on Build GRCh38
rs568061373 Chr.19: 55483743-55483743 on Build GRCh38
rs112775775 Chr.19: 55483781-55483781 on Build GRCh38
rs771948704 Chr.19: 55483687-55483719 on Build GRCh38
rs770510539 Chr.19: 55483752-55483753 on Build GRCh38
rs750219385 Chr.19: 55483771-55483771 on Build GRCh38
rs757029887 Chr.19: 55483752-55483752 on Build GRCh38
rs201058799 Chr.19: 55483665-55483665 on Build GRCh38
rs912587130 Chr.19: 55483613-55483613 on Build GRCh38
rs567123998 Chr.19: 55483737-55483737 on Build GRCh38
rs572593368 Chr.19: 55483770-55483770 on Build GRCh38
rs769352712 Chr.19: 55483672-55483672 on Build GRCh38
rs746972880 Chr.19: 55483746-55483747 on Build GRCh38
rs371286782 Chr.19: 55483678-55483678 on Build GRCh38
rs758379873 Chr.19: 55483635-55483635 on Build GRCh38
rs780328005 Chr.19: 55483664-55483664 on Build GRCh38
rs114065657 Chr.19: 55483608-55483608 on Build GRCh38
rs138326099 Chr.19: 55483746-55483746 on Build GRCh38
rs151142470 Chr.19: 55483751-55483751 on Build GRCh38
rs777519969 Chr.19: 55483720-55483752 on Build GRCh38
rs762448718 Chr.19: 55483748-55483748 on Build GRCh38
rs557237868 Chr.19: 55483719-55483719 on Build GRCh38

View Full Product Details