Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 676229-676488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHBDL1
Entrez Gene ID
9028
Gene Name
rhomboid like 1
Gene Aliases
RHBDL, RRP
UniGene
Hs.137572
Gene Chromosome Location
Chr.16: 675666-678271 on Build GRCh38
Transcripts
NM_001278720.1 NM_001278721.1 NM_001318733.1 XM_017023849.1 XM_017023850.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5470564 c.70G>A p.G24S Chr.16: 676366-676366 on Build GRCh38
4789047 c.97C>T p.H33Y Chr.16: 676393-676393 on Build GRCh38
3795118 c.317C>T p.P106L Chr.16: 676418-676418 on Build GRCh38
6436519 c.163G>A p.E55K Chr.16: 676459-676459 on Build GRCh38
5669954 c.262A>T p.I88F Chr.16: 676363-676363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144739170 Chr.16: 676320-676320 on Build GRCh38
rs372057727 Chr.16: 676370-676370 on Build GRCh38
rs769572513 Chr.16: 676387-676387 on Build GRCh38
rs991633170 Chr.16: 676288-676288 on Build GRCh38
rs139776304 Chr.16: 676301-676301 on Build GRCh38
rs376628523 Chr.16: 676295-676295 on Build GRCh38
rs759274001 Chr.16: 676341-676341 on Build GRCh38
rs938146370 Chr.16: 676404-676404 on Build GRCh38
rs567095689 Chr.16: 676414-676414 on Build GRCh38
rs756689120 Chr.16: 676458-676458 on Build GRCh38
rs142178722 Chr.16: 676374-676374 on Build GRCh38
rs367618276 Chr.16: 676256-676256 on Build GRCh38
rs751610014 Chr.16: 676277-676277 on Build GRCh38
rs372353908 Chr.16: 676286-676286 on Build GRCh38
rs770752263 Chr.16: 676315-676315 on Build GRCh38
rs764279309 Chr.16: 676273-676273 on Build GRCh38
rs774507179 Chr.16: 676395-676395 on Build GRCh38
rs566759044 Chr.16: 676367-676367 on Build GRCh38
rs757317283 Chr.16: 676279-676279 on Build GRCh38
rs146745951 Chr.16: 676376-676376 on Build GRCh38
rs573526564 Chr.16: 676267-676267 on Build GRCh38
rs756671413 Chr.16: 676351-676351 on Build GRCh38
rs746998516 Chr.16: 676371-676371 on Build GRCh38
rs375143444 Chr.16: 676443-676443 on Build GRCh38
rs781118837 Chr.16: 676459-676459 on Build GRCh38
rs746553460 Chr.16: 676448-676448 on Build GRCh38
rs767000541 Chr.16: 676418-676418 on Build GRCh38
rs752329828 Chr.16: 676440-676440 on Build GRCh38
rs764251039 Chr.16: 676346-676346 on Build GRCh38
rs201853642 Chr.16: 676366-676366 on Build GRCh38
rs771550390 Chr.16: 676310-676310 on Build GRCh38
rs745664865 Chr.16: 676382-676382 on Build GRCh38
rs778588458 Chr.16: 676300-676300 on Build GRCh38
rs760801746 Chr.16: 676416-676416 on Build GRCh38
rs758043408 Chr.16: 676441-676441 on Build GRCh38
rs1021135609 Chr.16: 676294-676294 on Build GRCh38
rs775334989 Chr.16: 676467-676467 on Build GRCh38
rs759672799 Chr.16: 676295-676295 on Build GRCh38
rs773079364 Chr.16: 676311-676311 on Build GRCh38
rs200103024 Chr.16: 676347-676347 on Build GRCh38
rs537678830 Chr.16: 676422-676422 on Build GRCh38
rs200536876 Chr.16: 676290-676290 on Build GRCh38
rs143977565 Chr.16: 676460-676460 on Build GRCh38
rs752983076 Chr.16: 676264-676264 on Build GRCh38
rs754342602 Chr.16: 676426-676426 on Build GRCh38
rs148652480 Chr.16: 676368-676368 on Build GRCh38
rs149809924 Chr.16: 676313-676313 on Build GRCh38
rs533346109 Chr.16: 676365-676365 on Build GRCh38
rs140442350 Chr.16: 676431-676431 on Build GRCh38
rs751011486 Chr.16: 676350-676350 on Build GRCh38

View Full Product Details