Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 77480938-77481199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROBO2
Entrez Gene ID
6092
Gene Name
roundabout guidance receptor 2
Gene Aliases
SAX3
UniGene
Hs.13305
Gene Chromosome Location
Chr.3: 75906675-77649964 on Build GRCh38
Transcripts
NM_001128929.3 NM_001290039.1 NM_001290040.1 NM_001290065.1 NM_002942.4 XM_011533981.2 XM_011533982.1 XM_011533983.1 XM_011533984.1 XM_011533985.1 XM_017006986.1 XM_017006987.1 XM_017006988.1 XM_017006989.1 XM_017006990.1 XM_017006991.1 XM_017006992.1 XM_017006993.1 XM_017006994.1 XM_017006995.1 XM_017006996.1 XM_017006997.1 XM_017006998.1 XM_017006999.1 XM_017007000.1 XM_017007001.1 XM_017007002.1 XM_017007003.1 XM_017007004.1 XM_017007005.1 XM_017007006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7325447 c.664A>G p.T222A Chr.3: 77481168-77481168 on Build GRCh38
7116215 c.572C>A p.S191Y Chr.3: 77481124-77481124 on Build GRCh38
4120236 c.563T>C p.L188P Chr.3: 77481115-77481115 on Build GRCh38
4120238 c.634A>G p.R212G Chr.3: 77481186-77481186 on Build GRCh38
5742978 c.630G>T p.R210S Chr.3: 77481134-77481134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371101033 Chr.3: 77481073-77481073 on Build GRCh38
rs113680429 Chr.3: 77481096-77481096 on Build GRCh38
rs192994111 Chr.3: 77480984-77480984 on Build GRCh38
rs963810885 Chr.3: 77481144-77481144 on Build GRCh38
rs745573128 Chr.3: 77481170-77481170 on Build GRCh38
rs775901046 Chr.3: 77481179-77481179 on Build GRCh38
rs1032435655 Chr.3: 77481030-77481030 on Build GRCh38
rs778905542 Chr.3: 77481132-77481132 on Build GRCh38
rs752040148 Chr.3: 77481113-77481113 on Build GRCh38
rs763574051 Chr.3: 77481102-77481102 on Build GRCh38
rs760105370 Chr.3: 77481090-77481090 on Build GRCh38
rs200483677 Chr.3: 77481103-77481103 on Build GRCh38
rs575830317 Chr.3: 77480999-77480999 on Build GRCh38
rs770419777 Chr.3: 77481091-77481091 on Build GRCh38
rs770382531 Chr.3: 77481149-77481149 on Build GRCh38
rs542054744 Chr.3: 77480967-77480967 on Build GRCh38
rs1000585873 Chr.3: 77480996-77480996 on Build GRCh38
rs765634836 Chr.3: 77481100-77481100 on Build GRCh38
rs771830264 Chr.3: 77481135-77481135 on Build GRCh38
rs562914871 Chr.3: 77481126-77481126 on Build GRCh38
rs753038257 Chr.3: 77481101-77481101 on Build GRCh38
rs746918841 Chr.3: 77481140-77481140 on Build GRCh38
rs781379035 Chr.3: 77481119-77481119 on Build GRCh38
rs776091168 Chr.3: 77481161-77481161 on Build GRCh38
rs201584681 Chr.3: 77481117-77481117 on Build GRCh38
rs778819492 Chr.3: 77481057-77481057 on Build GRCh38
rs377490138 Chr.3: 77481048-77481048 on Build GRCh38
rs1026801097 Chr.3: 77481116-77481116 on Build GRCh38
rs777582973 Chr.3: 77481138-77481138 on Build GRCh38
rs2121816 Chr.3: 77481109-77481109 on Build GRCh38
rs375164221 Chr.3: 77481122-77481122 on Build GRCh38
rs768602825 Chr.3: 77481051-77481051 on Build GRCh38
rs564036431 Chr.3: 77481043-77481043 on Build GRCh38
rs374693254 Chr.3: 77481095-77481095 on Build GRCh38
rs772843473 Chr.3: 77481084-77481084 on Build GRCh38
rs769423024 Chr.3: 77481175-77481175 on Build GRCh38
rs1053412408 Chr.3: 77480989-77480989 on Build GRCh38
rs902261873 Chr.3: 77480972-77480972 on Build GRCh38
rs747948687 Chr.3: 77481134-77481134 on Build GRCh38
rs892118988 Chr.3: 77481038-77481038 on Build GRCh38
rs886058876 Chr.3: 77481110-77481110 on Build GRCh38
rs771886885 Chr.3: 77481078-77481078 on Build GRCh38

View Full Product Details