Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 128814724-128814991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC15A4
Entrez Gene ID
121260
Gene Name
solute carrier family 15 member 4
Gene Aliases
FP12591, PHT1, PTR4
UniGene
Hs.507260
Gene Chromosome Location
Chr.12: 128793194-128823996 on Build GRCh38
Transcripts
NM_145648.3 XM_011537895.1 XM_017018791.1 XM_017018792.1 XM_017018793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4726535 c.699C>T p.V233V Chr.12: 128814918-128814918 on Build GRCh38
3376172 c.645T>C p.Y215Y Chr.12: 128814972-128814972 on Build GRCh38
5021778 c.716T>C p.V239A Chr.12: 128814901-128814901 on Build GRCh38
367107 c.635G>T p.G212V Chr.12: 128814982-128814982 on Build GRCh38
7154265 c.693C>T p.V231V Chr.12: 128814924-128814924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751202723 Chr.12: 128814932-128814932 on Build GRCh38
rs35439692 Chr.12: 128814781-128814781 on Build GRCh38
rs201371924 Chr.12: 128814842-128814842 on Build GRCh38
rs756744535 Chr.12: 128814934-128814934 on Build GRCh38
rs33990080 Chr.12: 128814901-128814901 on Build GRCh38
rs779979815 Chr.12: 128814952-128814952 on Build GRCh38
rs746920247 Chr.12: 128814786-128814786 on Build GRCh38
rs199661368 Chr.12: 128814787-128814787 on Build GRCh38
rs377171343 Chr.12: 128814882-128814882 on Build GRCh38
rs370092029 Chr.12: 128814916-128814916 on Build GRCh38
rs756449348 Chr.12: 128814830-128814830 on Build GRCh38
rs754716226 Chr.12: 128814881-128814881 on Build GRCh38
rs10847697 Chr.12: 128814840-128814840 on Build GRCh38
rs772892319 Chr.12: 128814783-128814783 on Build GRCh38
rs369480906 Chr.12: 128814767-128814767 on Build GRCh38
rs35580712 Chr.12: 128814890-128814890 on Build GRCh38
rs780617143 Chr.12: 128814936-128814936 on Build GRCh38
rs919577174 Chr.12: 128814946-128814946 on Build GRCh38
rs201786635 Chr.12: 128814959-128814959 on Build GRCh38
rs745816903 Chr.12: 128814875-128814875 on Build GRCh38
rs141893441 Chr.12: 128814809-128814809 on Build GRCh38
rs774440827 Chr.12: 128814892-128814892 on Build GRCh38
rs372436878 Chr.12: 128814841-128814841 on Build GRCh38
rs767996998 Chr.12: 128814826-128814826 on Build GRCh38
rs750920352 Chr.12: 128814827-128814827 on Build GRCh38
rs774580370 Chr.12: 128814960-128814960 on Build GRCh38
rs942446529 Chr.12: 128814920-128814920 on Build GRCh38
rs751719646 Chr.12: 128814814-128814814 on Build GRCh38
rs749972447 Chr.12: 128814822-128814822 on Build GRCh38
rs376967004 Chr.12: 128814937-128814937 on Build GRCh38
rs769779173 Chr.12: 128814944-128814944 on Build GRCh38
rs763146072 Chr.12: 128814888-128814888 on Build GRCh38
rs781144726 Chr.12: 128814870-128814870 on Build GRCh38
rs929610161 Chr.12: 128814961-128814961 on Build GRCh38
rs770352171 Chr.12: 128814866-128814866 on Build GRCh38
rs749014801 Chr.12: 128814957-128814957 on Build GRCh38
rs746473520 Chr.12: 128814865-128814865 on Build GRCh38
rs373549893 Chr.12: 128814918-128814918 on Build GRCh38
rs201775464 Chr.12: 128814788-128814788 on Build GRCh38
rs201921716 Chr.12: 128814921-128814921 on Build GRCh38
rs767266484 Chr.12: 128814861-128814861 on Build GRCh38
rs767108406 Chr.12: 128814811-128814811 on Build GRCh38
rs964143980 Chr.12: 128814823-128814823 on Build GRCh38
rs757297305 Chr.12: 128814816-128814816 on Build GRCh38
rs763201114 Chr.12: 128814807-128814807 on Build GRCh38
rs765047849 Chr.12: 128814797-128814797 on Build GRCh38
rs755410968 Chr.12: 128814923-128814923 on Build GRCh38
rs756437219 Chr.12: 128814768-128814768 on Build GRCh38
rs1031312276 Chr.12: 128814864-128814864 on Build GRCh38
rs761909134 Chr.12: 128814893-128814893 on Build GRCh38
rs938097004 Chr.12: 128814925-128814925 on Build GRCh38
rs536892230 Chr.12: 128814843-128814843 on Build GRCh38
rs955593396 Chr.12: 128814868-128814868 on Build GRCh38
rs775641409 Chr.12: 128814799-128814799 on Build GRCh38
rs777421504 Chr.12: 128814929-128814929 on Build GRCh38
rs573021477 Chr.12: 128814770-128814770 on Build GRCh38
rs767512302 Chr.12: 128814895-128814895 on Build GRCh38

View Full Product Details