Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 45748261-45748532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC14A1
Entrez Gene ID
6563
Gene Name
solute carrier family 14 member 1 (Kidd blood group)
Gene Aliases
HUT11, HsT1341, JK, RACH1, RACH2, UT-B1, UT1, UTE
UniGene
Hs.101307
Gene Chromosome Location
Chr.18: 45724123-45752520 on Build GRCh38
Transcripts
NM_001128588.3 NM_001146036.2 NM_001146037.1 NM_001308278.1 NM_001308279.1 NM_015865.6 XM_005258329.1 XM_005258333.1 XM_006722526.3 XM_011526142.1 XM_011526144.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4072277 c.979G>A p.A327T Chr.18: 45748408-45748408 on Build GRCh38
2877492 c.996G>A p.E332E Chr.18: 45748425-45748425 on Build GRCh38
3796419 c.994G>C p.E332Q Chr.18: 45748423-45748423 on Build GRCh38
6465405 c.1115-4G>A p.? Chr.18: 45748372-45748372 on Build GRCh38
6465406 c.947-4G>A p.? Chr.18: 45748372-45748372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751697666 Chr.18: 45748360-45748361 on Build GRCh38
rs757527142 Chr.18: 45748361-45748361 on Build GRCh38
rs1019935777 Chr.18: 45748321-45748321 on Build GRCh38
rs778842969 Chr.18: 45748360-45748360 on Build GRCh38
rs200167236 Chr.18: 45748334-45748334 on Build GRCh38
rs188063122 Chr.18: 45748434-45748434 on Build GRCh38
rs746135759 Chr.18: 45748378-45748378 on Build GRCh38
rs911488140 Chr.18: 45748380-45748380 on Build GRCh38
rs753669513 Chr.18: 45748345-45748345 on Build GRCh38
rs140320419 Chr.18: 45748402-45748402 on Build GRCh38
rs934902019 Chr.18: 45748505-45748505 on Build GRCh38
rs751076758 Chr.18: 45748326-45748326 on Build GRCh38
rs374156279 Chr.18: 45748443-45748443 on Build GRCh38
rs781490241 Chr.18: 45748431-45748431 on Build GRCh38
rs769240583 Chr.18: 45748396-45748396 on Build GRCh38
rs750106254 Chr.18: 45748335-45748335 on Build GRCh38
rs202010103 Chr.18: 45748339-45748339 on Build GRCh38
rs776501225 Chr.18: 45748401-45748401 on Build GRCh38
rs767610085 Chr.18: 45748389-45748389 on Build GRCh38
rs474270 Chr.18: 45748330-45748330 on Build GRCh38
rs377318084 Chr.18: 45748458-45748458 on Build GRCh38
rs754816074 Chr.18: 45748475-45748475 on Build GRCh38
rs28398419 Chr.18: 45748325-45748325 on Build GRCh38
rs771040146 Chr.18: 45748368-45748368 on Build GRCh38
rs997294167 Chr.18: 45748332-45748332 on Build GRCh38
rs753525998 Chr.18: 45748467-45748467 on Build GRCh38
rs565898944 Chr.18: 45748385-45748385 on Build GRCh38
rs372565028 Chr.18: 45748340-45748340 on Build GRCh38
rs376375507 Chr.18: 45748386-45748386 on Build GRCh38
rs986026630 Chr.18: 45748349-45748349 on Build GRCh38
rs537252659 Chr.18: 45748496-45748496 on Build GRCh38
rs1011242124 Chr.18: 45748302-45748302 on Build GRCh38
rs747068460 Chr.18: 45748365-45748365 on Build GRCh38
rs981580189 Chr.18: 45748479-45748479 on Build GRCh38
rs147114544 Chr.18: 45748408-45748408 on Build GRCh38
rs767190566 Chr.18: 45748414-45748414 on Build GRCh38
rs754869097 Chr.18: 45748346-45748346 on Build GRCh38
rs781473333 Chr.18: 45748376-45748376 on Build GRCh38

View Full Product Details