Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56525380-56525637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF471
Entrez Gene ID
57573
Gene Name
zinc finger protein 471
Gene Aliases
ERP1, Z1971
UniGene
Hs.230188
Gene Chromosome Location
Chr.19: 56507843-56530214 on Build GRCh38
Transcripts
NM_001321768.1 NM_020813.3 XM_011527148.1 XM_017027032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1396831 c.1479A>C p.G493G Chr.19: 56525546-56525546 on Build GRCh38
4931257 c.1346A>C p.H449P Chr.19: 56525413-56525413 on Build GRCh38
1396832 c.1534G>T p.E512* Chr.19: 56525601-56525601 on Build GRCh38
7149396 c.1408C>G p.Q470E Chr.19: 56525475-56525475 on Build GRCh38
6017743 c.1373C>T p.P458L Chr.19: 56525440-56525440 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777528841 Chr.19: 56525461-56525461 on Build GRCh38
rs752713599 Chr.19: 56525556-56525556 on Build GRCh38
rs746665208 Chr.19: 56525479-56525479 on Build GRCh38
rs555600680 Chr.19: 56525533-56525533 on Build GRCh38
rs369270810 Chr.19: 56525501-56525501 on Build GRCh38
rs765040103 Chr.19: 56525555-56525555 on Build GRCh38
rs771272067 Chr.19: 56525486-56525486 on Build GRCh38
rs754940350 Chr.19: 56525438-56525438 on Build GRCh38
rs775617412 Chr.19: 56525525-56525525 on Build GRCh38
rs866891164 Chr.19: 56525440-56525440 on Build GRCh38
rs751798855 Chr.19: 56525417-56525417 on Build GRCh38
rs758005517 Chr.19: 56525459-56525459 on Build GRCh38
rs113529011 Chr.19: 56525513-56525513 on Build GRCh38
rs762033462 Chr.19: 56525545-56525545 on Build GRCh38
rs757915725 Chr.19: 56525575-56525575 on Build GRCh38
rs781196948 Chr.19: 56525552-56525553 on Build GRCh38
rs375130699 Chr.19: 56525429-56525429 on Build GRCh38
rs191965415 Chr.19: 56525423-56525423 on Build GRCh38
rs758777065 Chr.19: 56525433-56525459 on Build GRCh38
rs778172247 Chr.19: 56525436-56525436 on Build GRCh38
rs8108270 Chr.19: 56525548-56525548 on Build GRCh38
rs200784127 Chr.19: 56525442-56525442 on Build GRCh38
rs1012708346 Chr.19: 56525577-56525577 on Build GRCh38
rs1046506533 Chr.19: 56525563-56525563 on Build GRCh38
rs142901193 Chr.19: 56525495-56525495 on Build GRCh38
rs777439262 Chr.19: 56525581-56525581 on Build GRCh38
rs776399463 Chr.19: 56525550-56525550 on Build GRCh38
rs186228630 Chr.19: 56525509-56525509 on Build GRCh38
rs199918463 Chr.19: 56525552-56525552 on Build GRCh38
rs764347946 Chr.19: 56525408-56525408 on Build GRCh38
rs138670329 Chr.19: 56525490-56525490 on Build GRCh38
rs771471502 Chr.19: 56525541-56525542 on Build GRCh38
rs756925379 Chr.19: 56525603-56525603 on Build GRCh38
rs747514348 Chr.19: 56525537-56525538 on Build GRCh38
rs372782062 Chr.19: 56525582-56525582 on Build GRCh38
rs752308979 Chr.19: 56525451-56525451 on Build GRCh38
rs372005205 Chr.19: 56525413-56525413 on Build GRCh38
rs745988727 Chr.19: 56525585-56525598 on Build GRCh38
rs774487781 Chr.19: 56525539-56525539 on Build GRCh38
rs768750703 Chr.19: 56525535-56525535 on Build GRCh38
rs79222237 Chr.19: 56525420-56525420 on Build GRCh38
rs150139976 Chr.19: 56525441-56525441 on Build GRCh38
rs541658230 Chr.19: 56525606-56525606 on Build GRCh38
rs752937306 Chr.19: 56525416-56525417 on Build GRCh38

View Full Product Details