Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 78137978-78138217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF9B
Entrez Gene ID
51616
Gene Name
TATA-box binding protein associated factor 9b
Gene Aliases
DN-7, DN7, TAF9L, TAFII31L, TFIID-31
UniGene
Hs.592248
Gene Chromosome Location
Chr.X: 78129748-78139682 on Build GRCh38
Transcripts
NM_015975.4

SNP Detail

SNP ID Chromosome Location
rs781860580 Chr.X: 78138144-78138144 on Build GRCh38
rs781842827 Chr.X: 78138081-78138081 on Build GRCh38
rs149136745 Chr.X: 78138051-78138051 on Build GRCh38
rs782793081 Chr.X: 78138008-78138008 on Build GRCh38
rs782268006 Chr.X: 78138072-78138072 on Build GRCh38
rs782575339 Chr.X: 78138043-78138043 on Build GRCh38
rs782694960 Chr.X: 78138121-78138121 on Build GRCh38
rs375706397 Chr.X: 78138053-78138053 on Build GRCh38
rs950337689 Chr.X: 78138184-78138184 on Build GRCh38
rs979179220 Chr.X: 78138128-78138128 on Build GRCh38
rs781956732 Chr.X: 78138095-78138095 on Build GRCh38
rs782109706 Chr.X: 78138100-78138100 on Build GRCh38
rs782483788 Chr.X: 78138129-78138129 on Build GRCh38
rs917533164 Chr.X: 78138180-78138180 on Build GRCh38
rs966336184 Chr.X: 78138032-78138032 on Build GRCh38
rs782707275 Chr.X: 78138089-78138089 on Build GRCh38
rs781899334 Chr.X: 78138027-78138027 on Build GRCh38
rs782643274 Chr.X: 78138071-78138071 on Build GRCh38
rs782322290 Chr.X: 78138093-78138093 on Build GRCh38
rs782181450 Chr.X: 78138092-78138092 on Build GRCh38
rs782410167 Chr.X: 78138073-78138073 on Build GRCh38
rs992212404 Chr.X: 78138165-78138165 on Build GRCh38
rs1018879704 Chr.X: 78138024-78138024 on Build GRCh38
rs782017595 Chr.X: 78138102-78138102 on Build GRCh38
rs782493957 Chr.X: 78138059-78138059 on Build GRCh38
rs200232048 Chr.X: 78138134-78138134 on Build GRCh38
rs782201154 Chr.X: 78138058-78138058 on Build GRCh38
rs782034254 Chr.X: 78138087-78138087 on Build GRCh38
rs782160975 Chr.X: 78138108-78138108 on Build GRCh38
rs369109622 Chr.X: 78138101-78138101 on Build GRCh38

View Full Product Details